Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5343344-5343577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR51B5
Entrez Gene ID
282763
Gene Name
olfactory receptor family 51 subfamily B member 5
Gene Aliases
HOR5'Beta5, OR11-37
UniGene
Hs.690458
Gene Chromosome Location
Chr.11: 5340883-5505652 on Build GRCh38
Transcripts
NM_001005567.2 NR_038321.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
226249 c.77C>T p.S26F Chr.11: 5343448-5343448 on Build GRCh38
223437 c.160C>T p.H54Y Chr.11: 5343365-5343365 on Build GRCh38
3448643 c.21C>T p.S7S Chr.11: 5343504-5343504 on Build GRCh38
3666374 c.3G>C p.M1I Chr.11: 5343522-5343522 on Build GRCh38
4033542 c.110T>C p.L37P Chr.11: 5343415-5343415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771546661 Chr.11: 5343416-5343416 on Build GRCh38
rs149637851 Chr.11: 5343521-5343521 on Build GRCh38
rs199512163 Chr.11: 5343494-5343494 on Build GRCh38
rs141129878 Chr.11: 5343399-5343399 on Build GRCh38
rs147069555 Chr.11: 5343495-5343495 on Build GRCh38
rs397830523 Chr.11: 5343549-5343550 on Build GRCh38
rs138291126 Chr.11: 5343519-5343519 on Build GRCh38
rs767555437 Chr.11: 5343547-5343548 on Build GRCh38
rs775093256 Chr.11: 5343479-5343480 on Build GRCh38
rs763471851 Chr.11: 5343413-5343413 on Build GRCh38
rs759271625 Chr.11: 5343458-5343478 on Build GRCh38
rs763534331 Chr.11: 5343511-5343511 on Build GRCh38
rs746334115 Chr.11: 5343537-5343537 on Build GRCh38
rs770781282 Chr.11: 5343388-5343390 on Build GRCh38
rs749588708 Chr.11: 5343477-5343477 on Build GRCh38
rs529505176 Chr.11: 5343516-5343516 on Build GRCh38
rs577566074 Chr.11: 5343485-5343485 on Build GRCh38
rs748373536 Chr.11: 5343404-5343404 on Build GRCh38
rs754413438 Chr.11: 5343550-5343555 on Build GRCh38
rs146946914 Chr.11: 5343446-5343446 on Build GRCh38
rs781385753 Chr.11: 5343403-5343403 on Build GRCh38
rs768849113 Chr.11: 5343491-5343491 on Build GRCh38
rs200657676 Chr.11: 5343478-5343478 on Build GRCh38
rs775039177 Chr.11: 5343418-5343418 on Build GRCh38
rs776034792 Chr.11: 5343500-5343500 on Build GRCh38
rs752700199 Chr.11: 5343520-5343520 on Build GRCh38
rs554096970 Chr.11: 5343503-5343503 on Build GRCh38
rs774986351 Chr.11: 5343488-5343488 on Build GRCh38
rs777853171 Chr.11: 5343523-5343523 on Build GRCh38
rs267602946 Chr.11: 5343380-5343380 on Build GRCh38
rs11036913 Chr.11: 5343512-5343512 on Build GRCh38
rs534026223 Chr.11: 5343462-5343462 on Build GRCh38
rs555677622 Chr.11: 5343464-5343464 on Build GRCh38
rs773484190 Chr.11: 5343412-5343412 on Build GRCh38
rs574030611 Chr.11: 5343473-5343473 on Build GRCh38
rs765130324 Chr.11: 5343467-5343468 on Build GRCh38
rs746503842 Chr.11: 5343551-5343551 on Build GRCh38
rs779427713 Chr.11: 5343533-5343533 on Build GRCh38
rs78253695 Chr.11: 5343546-5343547 on Build GRCh38
rs760258239 Chr.11: 5343422-5343422 on Build GRCh38
rs201816284 Chr.11: 5343396-5343396 on Build GRCh38
rs12801245 Chr.11: 5343545-5343545 on Build GRCh38
rs770131750 Chr.11: 5343406-5343406 on Build GRCh38
rs370052288 Chr.11: 5343401-5343401 on Build GRCh38
rs138029050 Chr.11: 5343447-5343447 on Build GRCh38
rs141505911 Chr.11: 5343476-5343476 on Build GRCh38
rs772953127 Chr.11: 5343507-5343507 on Build GRCh38
rs566514307 Chr.11: 5343461-5343461 on Build GRCh38
rs544747302 Chr.11: 5343486-5343486 on Build GRCh38
rs571418177 Chr.11: 5343370-5343370 on Build GRCh38
rs985219587 Chr.11: 5343395-5343395 on Build GRCh38
rs957861990 Chr.11: 5343393-5343393 on Build GRCh38
rs776450308 Chr.11: 5343451-5343452 on Build GRCh38
rs761374346 Chr.11: 5343430-5343430 on Build GRCh38
rs762662467 Chr.11: 5343510-5343512 on Build GRCh38
rs758298388 Chr.11: 5343376-5343376 on Build GRCh38
rs995797416 Chr.11: 5343391-5343391 on Build GRCh38
rs376277471 Chr.11: 5343471-5343471 on Build GRCh38
rs969503389 Chr.11: 5343499-5343499 on Build GRCh38
rs763756903 Chr.11: 5343424-5343424 on Build GRCh38
rs761477133 Chr.11: 5343502-5343502 on Build GRCh38
rs201732422 Chr.11: 5343513-5343513 on Build GRCh38
rs753485829 Chr.11: 5343427-5343427 on Build GRCh38
rs199973004 Chr.11: 5343498-5343498 on Build GRCh38
rs766817329 Chr.11: 5343550-5343550 on Build GRCh38
rs12800375 Chr.11: 5343525-5343525 on Build GRCh38

View Full Product Details