Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33303896-33304163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAPBP
Entrez Gene ID
6892
Gene Name
TAP binding protein (tapasin)
Gene Aliases
NGS17, TAPA, TPN, TPSN
UniGene
Hs.370937
Gene Chromosome Location
Chr.6: 33299694-33314387 on Build GRCh38
Transcripts
NM_003190.4 NM_172208.2 NM_172209.2 XM_011514828.2 XM_017011227.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1621516 c.1300+4A>C p.? Chr.6: 33304124-33304124 on Build GRCh38
6586145 c.1281_1300+6del26 p.? Chr.6: 33304122-33304147 on Build GRCh38
6409109 c.1297G>A p.A433T Chr.6: 33304131-33304131 on Build GRCh38
6403560 c.1276C>G p.L426V Chr.6: 33304152-33304152 on Build GRCh38
1158245 c.1371C>G p.V457V Chr.6: 33303919-33303919 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749036026 Chr.6: 33304083-33304083 on Build GRCh38
rs778241930 Chr.6: 33304010-33304010 on Build GRCh38
rs753985919 Chr.6: 33303965-33303965 on Build GRCh38
rs746600048 Chr.6: 33304020-33304020 on Build GRCh38
rs924449358 Chr.6: 33304075-33304075 on Build GRCh38
rs755140017 Chr.6: 33303979-33303979 on Build GRCh38
rs372291038 Chr.6: 33304085-33304085 on Build GRCh38
rs368035810 Chr.6: 33304064-33304064 on Build GRCh38
rs759758236 Chr.6: 33304112-33304112 on Build GRCh38
rs752989371 Chr.6: 33304004-33304004 on Build GRCh38
rs749130678 Chr.6: 33303942-33303942 on Build GRCh38
rs775432139 Chr.6: 33304081-33304081 on Build GRCh38
rs753120114 Chr.6: 33304132-33304132 on Build GRCh38
rs758918099 Chr.6: 33304008-33304008 on Build GRCh38
rs770150271 Chr.6: 33304122-33304125 on Build GRCh38
rs766663522 Chr.6: 33303943-33303943 on Build GRCh38
rs949862956 Chr.6: 33304058-33304058 on Build GRCh38
rs774124089 Chr.6: 33304092-33304092 on Build GRCh38
rs966303595 Chr.6: 33304120-33304120 on Build GRCh38
rs780950801 Chr.6: 33304028-33304028 on Build GRCh38
rs981234947 Chr.6: 33304074-33304074 on Build GRCh38
rs200105727 Chr.6: 33303925-33303925 on Build GRCh38
rs144810851 Chr.6: 33304128-33304128 on Build GRCh38
rs760853660 Chr.6: 33304099-33304099 on Build GRCh38
rs768778713 Chr.6: 33304089-33304089 on Build GRCh38
rs766392504 Chr.6: 33304108-33304108 on Build GRCh38
rs770319484 Chr.6: 33304026-33304026 on Build GRCh38
rs568174690 Chr.6: 33303936-33303936 on Build GRCh38
rs758758961 Chr.6: 33304139-33304139 on Build GRCh38
rs974359476 Chr.6: 33303930-33303930 on Build GRCh38
rs769532741 Chr.6: 33304078-33304078 on Build GRCh38
rs760665415 Chr.6: 33303940-33303940 on Build GRCh38
rs73741554 Chr.6: 33304003-33304003 on Build GRCh38
rs140890714 Chr.6: 33304040-33304040 on Build GRCh38
rs1027705232 Chr.6: 33303967-33303967 on Build GRCh38
rs771709981 Chr.6: 33304111-33304111 on Build GRCh38
rs765383234 Chr.6: 33304131-33304131 on Build GRCh38
rs112348926 Chr.6: 33303991-33303991 on Build GRCh38
rs568571603 Chr.6: 33304067-33304067 on Build GRCh38
rs914537869 Chr.6: 33304142-33304142 on Build GRCh38

View Full Product Details