Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3088963-3089224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSCAN10
Entrez Gene ID
84891
Gene Name
zinc finger and SCAN domain containing 10
Gene Aliases
ZNF206
UniGene
Hs.334515
Gene Chromosome Location
Chr.16: 3088890-3099317 on Build GRCh38
Transcripts
NM_001282415.1 NM_001282416.1 NM_032805.2 XM_017023791.1 XM_017023792.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4391783 c.2140C>T p.R714C Chr.16: 3089129-3089129 on Build GRCh38
6391737 c.2054T>C p.L685P Chr.16: 3089215-3089215 on Build GRCh38
6391828 c.2104T>C p.C702R Chr.16: 3089165-3089165 on Build GRCh38
6834224 c.2075delC p.T692fs*>34 Chr.16: 3089194-3089194 on Build GRCh38
6393972 c.2075C>T p.T692M Chr.16: 3089194-3089194 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577693683 Chr.16: 3089112-3089112 on Build GRCh38
rs749178024 Chr.16: 3089048-3089048 on Build GRCh38
rs778605954 Chr.16: 3089086-3089086 on Build GRCh38
rs752262251 Chr.16: 3089071-3089071 on Build GRCh38
rs1034384387 Chr.16: 3088999-3088999 on Build GRCh38
rs764365956 Chr.16: 3089055-3089055 on Build GRCh38
rs149187078 Chr.16: 3089035-3089035 on Build GRCh38
rs752102720 Chr.16: 3089185-3089185 on Build GRCh38
rs973081248 Chr.16: 3089033-3089033 on Build GRCh38
rs755684702 Chr.16: 3089075-3089075 on Build GRCh38
rs753350310 Chr.16: 3089191-3089191 on Build GRCh38
rs540219705 Chr.16: 3089184-3089184 on Build GRCh38
rs767775089 Chr.16: 3089082-3089083 on Build GRCh38
rs779961203 Chr.16: 3089111-3089111 on Build GRCh38
rs768699536 Chr.16: 3089120-3089120 on Build GRCh38
rs766937073 Chr.16: 3089069-3089069 on Build GRCh38
rs761968967 Chr.16: 3089159-3089159 on Build GRCh38
rs141059976 Chr.16: 3089108-3089108 on Build GRCh38
rs1012270016 Chr.16: 3089142-3089142 on Build GRCh38
rs776947172 Chr.16: 3089132-3089132 on Build GRCh38
rs903221467 Chr.16: 3089195-3089195 on Build GRCh38
rs773459938 Chr.16: 3089169-3089169 on Build GRCh38
rs757987581 Chr.16: 3088993-3088993 on Build GRCh38
rs756884410 Chr.16: 3089192-3089192 on Build GRCh38
rs915832700 Chr.16: 3089014-3089014 on Build GRCh38
rs765482884 Chr.16: 3089062-3089062 on Build GRCh38
rs772007968 Chr.16: 3089050-3089050 on Build GRCh38
rs75291351 Chr.16: 3089077-3089077 on Build GRCh38
rs376167359 Chr.16: 3089065-3089065 on Build GRCh38
rs1040080986 Chr.16: 3089118-3089118 on Build GRCh38
rs200764017 Chr.16: 3089047-3089047 on Build GRCh38
rs897538463 Chr.16: 3089183-3089183 on Build GRCh38
rs759695813 Chr.16: 3089083-3089083 on Build GRCh38
rs758848477 Chr.16: 3089066-3089066 on Build GRCh38
rs778572083 Chr.16: 3089193-3089193 on Build GRCh38
rs941666952 Chr.16: 3089027-3089027 on Build GRCh38
rs377718588 Chr.16: 3089049-3089049 on Build GRCh38
rs763312640 Chr.16: 3089170-3089170 on Build GRCh38
rs566855016 Chr.16: 3089187-3089187 on Build GRCh38
rs770006908 Chr.16: 3089160-3089160 on Build GRCh38
rs776820091 Chr.16: 3089059-3089059 on Build GRCh38
rs755846680 Chr.16: 3089046-3089046 on Build GRCh38
rs185364182 Chr.16: 3089189-3089189 on Build GRCh38
rs910483646 Chr.16: 3089105-3089105 on Build GRCh38
rs891262329 Chr.16: 3089056-3089056 on Build GRCh38
rs745629189 Chr.16: 3089094-3089094 on Build GRCh38
rs555768061 Chr.16: 3089207-3089207 on Build GRCh38
rs535883956 Chr.16: 3089188-3089188 on Build GRCh38
rs750251089 Chr.16: 3089202-3089202 on Build GRCh38
rs757116832 Chr.16: 3089082-3089082 on Build GRCh38

View Full Product Details