Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 48100923-48101148 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC12
Entrez Gene ID
94160
Gene Name
ATP binding cassette subfamily C member 12
Gene Aliases
MRP9
UniGene
Hs.410111
Gene Chromosome Location
Chr.16: 48082973-48146770 on Build GRCh38
Transcripts
NM_033226.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6016168 c.2960G>A p.W987* Chr.16: 48100950-48100950 on Build GRCh38
6522809 c.2968C>T p.H990Y Chr.16: 48100942-48100942 on Build GRCh38
6144549 c.2964C>T p.F988F Chr.16: 48100946-48100946 on Build GRCh38
4260400 c.2950C>T p.R984W Chr.16: 48100960-48100960 on Build GRCh38
7353610 c.2938G>A p.E980K Chr.16: 48100972-48100972 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745357141 Chr.16: 48101052-48101052 on Build GRCh38
rs769332846 Chr.16: 48101055-48101055 on Build GRCh38
rs375381289 Chr.16: 48101028-48101028 on Build GRCh38
rs548362111 Chr.16: 48100987-48100987 on Build GRCh38
rs374564911 Chr.16: 48101011-48101011 on Build GRCh38
rs770413564 Chr.16: 48100953-48100953 on Build GRCh38
rs897576954 Chr.16: 48101121-48101121 on Build GRCh38
rs764416997 Chr.16: 48101047-48101047 on Build GRCh38
rs774895053 Chr.16: 48100943-48100943 on Build GRCh38
rs373868982 Chr.16: 48101003-48101003 on Build GRCh38
rs370623509 Chr.16: 48100958-48100958 on Build GRCh38
rs547827994 Chr.16: 48101037-48101037 on Build GRCh38
rs1016909905 Chr.16: 48101068-48101068 on Build GRCh38
rs751500861 Chr.16: 48100999-48100999 on Build GRCh38
rs1014088338 Chr.16: 48101030-48101030 on Build GRCh38
rs780403881 Chr.16: 48101024-48101024 on Build GRCh38
rs568390871 Chr.16: 48101033-48101033 on Build GRCh38
rs750502026 Chr.16: 48101012-48101012 on Build GRCh38
rs534363642 Chr.16: 48101095-48101095 on Build GRCh38
rs769195586 Chr.16: 48100942-48100942 on Build GRCh38
rs202003321 Chr.16: 48101034-48101034 on Build GRCh38
rs756358542 Chr.16: 48101021-48101021 on Build GRCh38
rs775170627 Chr.16: 48100960-48100960 on Build GRCh38
rs755392633 Chr.16: 48101029-48101029 on Build GRCh38
rs6500305 Chr.16: 48100945-48100945 on Build GRCh38
rs1056143768 Chr.16: 48101110-48101110 on Build GRCh38
rs866552854 Chr.16: 48100967-48100967 on Build GRCh38
rs772483072 Chr.16: 48101043-48101043 on Build GRCh38
rs763862061 Chr.16: 48100993-48100993 on Build GRCh38
rs531444167 Chr.16: 48100959-48100959 on Build GRCh38
rs1003297065 Chr.16: 48101066-48101066 on Build GRCh38

View Full Product Details