Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 47344085-47344352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB13
Entrez Gene ID
124989
Gene Name
EF-hand calcium binding domain 13
Gene Aliases
C17orf57
UniGene
Hs.463303
Gene Chromosome Location
Chr.17: 47323961-47441312 on Build GRCh38
Transcripts
NM_001195192.1 NM_152347.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4916665 c.378T>A p.N126K Chr.17: 47344236-47344236 on Build GRCh38
5462654 c.406C>T p.P136S Chr.17: 47344264-47344264 on Build GRCh38
980439 c.434G>A p.R145Q Chr.17: 47344292-47344292 on Build GRCh38
3518538 c.394A>C p.K132Q Chr.17: 47344252-47344252 on Build GRCh38
1479683 c.419C>G p.S140C Chr.17: 47344277-47344277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750341759 Chr.17: 47344262-47344262 on Build GRCh38
rs775817590 Chr.17: 47344233-47344233 on Build GRCh38
rs750288576 Chr.17: 47344192-47344192 on Build GRCh38
rs752954769 Chr.17: 47344134-47344134 on Build GRCh38
rs757091075 Chr.17: 47344171-47344171 on Build GRCh38
rs764046159 Chr.17: 47344150-47344150 on Build GRCh38
rs748861760 Chr.17: 47344198-47344198 on Build GRCh38
rs768245889 Chr.17: 47344201-47344201 on Build GRCh38
rs751543643 Chr.17: 47344151-47344151 on Build GRCh38
rs781776880 Chr.17: 47344309-47344309 on Build GRCh38
rs78561706 Chr.17: 47344197-47344197 on Build GRCh38
rs147519392 Chr.17: 47344209-47344209 on Build GRCh38
rs758846715 Chr.17: 47344315-47344315 on Build GRCh38
rs766123779 Chr.17: 47344272-47344272 on Build GRCh38
rs771495756 Chr.17: 47344216-47344216 on Build GRCh38
rs140181680 Chr.17: 47344220-47344220 on Build GRCh38
rs113208705 Chr.17: 47344293-47344293 on Build GRCh38
rs775664888 Chr.17: 47344329-47344329 on Build GRCh38
rs761950286 Chr.17: 47344246-47344246 on Build GRCh38
rs754532197 Chr.17: 47344291-47344291 on Build GRCh38
rs757967112 Chr.17: 47344304-47344304 on Build GRCh38
rs759841251 Chr.17: 47344127-47344127 on Build GRCh38
rs763138951 Chr.17: 47344142-47344142 on Build GRCh38
rs757961619 Chr.17: 47344256-47344256 on Build GRCh38
rs760035327 Chr.17: 47344222-47344222 on Build GRCh38
rs753404813 Chr.17: 47344157-47344160 on Build GRCh38
rs879085629 Chr.17: 47344306-47344306 on Build GRCh38
rs777077005 Chr.17: 47344118-47344118 on Build GRCh38
rs900977510 Chr.17: 47344124-47344124 on Build GRCh38
rs747566876 Chr.17: 47344210-47344210 on Build GRCh38
rs774234656 Chr.17: 47344245-47344245 on Build GRCh38
rs202126404 Chr.17: 47344292-47344292 on Build GRCh38
rs267604922 Chr.17: 47344167-47344167 on Build GRCh38
rs138567967 Chr.17: 47344234-47344234 on Build GRCh38
rs778550478 Chr.17: 47344194-47344194 on Build GRCh38
rs753552730 Chr.17: 47344277-47344277 on Build GRCh38
rs770260947 Chr.17: 47344231-47344231 on Build GRCh38
rs755943359 Chr.17: 47344264-47344264 on Build GRCh38
rs765621224 Chr.17: 47344128-47344128 on Build GRCh38
rs199897750 Chr.17: 47344183-47344183 on Build GRCh38
rs746342069 Chr.17: 47344312-47344312 on Build GRCh38
rs767504556 Chr.17: 47344250-47344250 on Build GRCh38
rs556813827 Chr.17: 47344314-47344314 on Build GRCh38

View Full Product Details