Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 21167480-21167721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF431
Entrez Gene ID
170959
Gene Name
zinc finger protein 431
Gene Aliases
-
UniGene
Hs.656394
Gene Chromosome Location
Chr.19: 21142009-21191367 on Build GRCh38
Transcripts
NM_001319124.1 NM_001319126.1 NM_001319127.1 NM_133473.2 XM_011527750.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
299645 c.224-4G>A p.? Chr.19: 21167567-21167567 on Build GRCh38
4076164 c.283A>G p.N95D Chr.19: 21167630-21167630 on Build GRCh38
5855220 c.309T>A p.D103E Chr.19: 21167656-21167656 on Build GRCh38
6277914 c.269A>G p.E90G Chr.19: 21167616-21167616 on Build GRCh38
1685022 c.310G>A p.E104K Chr.19: 21167657-21167657 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963866208 Chr.19: 21167631-21167631 on Build GRCh38
rs1019406309 Chr.19: 21167628-21167628 on Build GRCh38
rs142355676 Chr.19: 21167611-21167611 on Build GRCh38
rs552681934 Chr.19: 21167585-21167585 on Build GRCh38
rs16997591 Chr.19: 21167527-21167527 on Build GRCh38
rs761370746 Chr.19: 21167664-21167664 on Build GRCh38
rs767143470 Chr.19: 21167682-21167682 on Build GRCh38
rs779915220 Chr.19: 21167608-21167608 on Build GRCh38
rs367939367 Chr.19: 21167648-21167648 on Build GRCh38
rs755800917 Chr.19: 21167600-21167600 on Build GRCh38
rs964820698 Chr.19: 21167687-21167687 on Build GRCh38
rs770416008 Chr.19: 21167679-21167680 on Build GRCh38
rs776793511 Chr.19: 21167626-21167626 on Build GRCh38
rs1025285190 Chr.19: 21167511-21167511 on Build GRCh38
rs780947521 Chr.19: 21167579-21167579 on Build GRCh38
rs764661029 Chr.19: 21167653-21167653 on Build GRCh38
rs765658315 Chr.19: 21167534-21167534 on Build GRCh38
rs572616048 Chr.19: 21167647-21167647 on Build GRCh38
rs771020067 Chr.19: 21167624-21167624 on Build GRCh38
rs753076297 Chr.19: 21167548-21167548 on Build GRCh38
rs923335347 Chr.19: 21167656-21167656 on Build GRCh38
rs759780027 Chr.19: 21167529-21167529 on Build GRCh38
rs774312871 Chr.19: 21167618-21167618 on Build GRCh38
rs752139278 Chr.19: 21167660-21167660 on Build GRCh38
rs766564654 Chr.19: 21167524-21167524 on Build GRCh38
rs751106970 Chr.19: 21167571-21167571 on Build GRCh38
rs768636309 Chr.19: 21167614-21167614 on Build GRCh38
rs747068487 Chr.19: 21167623-21167623 on Build GRCh38
rs759807857 Chr.19: 21167634-21167634 on Build GRCh38
rs375796628 Chr.19: 21167646-21167646 on Build GRCh38
rs758955541 Chr.19: 21167563-21167563 on Build GRCh38
rs996294730 Chr.19: 21167691-21167691 on Build GRCh38
rs764752892 Chr.19: 21167569-21167569 on Build GRCh38

View Full Product Details