Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43981246-43981489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALT2
Entrez Gene ID
8704
Gene Name
beta-1,4-galactosyltransferase 2
Gene Aliases
B4Gal-T2, B4Gal-T3, beta4Gal-T2
UniGene
Hs.632403
Gene Chromosome Location
Chr.1: 43979202-43991171 on Build GRCh38
Transcripts
NM_001005417.2 NM_003780.4 NM_030587.2 XM_017002716.1 XM_017002717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6501664 c.218G>A p.R73H Chr.1: 43981291-43981291 on Build GRCh38
6609434 c.96T>A p.F32L Chr.1: 43981256-43981256 on Build GRCh38
4610960 c.175_177delAGC p.S65delS Chr.1: 43981335-43981337 on Build GRCh38
4610959 c.262_264delAGC p.S94delS Chr.1: 43981335-43981337 on Build GRCh38
6609433 c.183T>A p.F61L Chr.1: 43981256-43981256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569291036 Chr.1: 43981311-43981311 on Build GRCh38
rs752759378 Chr.1: 43981335-43981337 on Build GRCh38
rs928961448 Chr.1: 43981399-43981399 on Build GRCh38
rs1047306273 Chr.1: 43981435-43981435 on Build GRCh38
rs912110603 Chr.1: 43981416-43981416 on Build GRCh38
rs757787915 Chr.1: 43981323-43981323 on Build GRCh38
rs764277669 Chr.1: 43981335-43981343 on Build GRCh38
rs769733983 Chr.1: 43981371-43981371 on Build GRCh38
rs781072642 Chr.1: 43981430-43981430 on Build GRCh38
rs767545341 Chr.1: 43981335-43981340 on Build GRCh38
rs755229393 Chr.1: 43981429-43981429 on Build GRCh38
rs762532434 Chr.1: 43981291-43981291 on Build GRCh38
rs774201196 Chr.1: 43981459-43981459 on Build GRCh38
rs367580012 Chr.1: 43981422-43981422 on Build GRCh38
rs148339623 Chr.1: 43981392-43981392 on Build GRCh38
rs767080506 Chr.1: 43981317-43981317 on Build GRCh38
rs754260512 Chr.1: 43981322-43981322 on Build GRCh38
rs758687714 Chr.1: 43981351-43981351 on Build GRCh38
rs751687755 Chr.1: 43981427-43981427 on Build GRCh38
rs199512234 Chr.1: 43981365-43981365 on Build GRCh38
rs769533543 Chr.1: 43981290-43981290 on Build GRCh38
rs779365326 Chr.1: 43981324-43981324 on Build GRCh38
rs1030999156 Chr.1: 43981372-43981372 on Build GRCh38
rs770551686 Chr.1: 43981377-43981377 on Build GRCh38
rs762398729 Chr.1: 43981403-43981403 on Build GRCh38
rs902610690 Chr.1: 43981460-43981460 on Build GRCh38
rs138683109 Chr.1: 43981380-43981380 on Build GRCh38
rs1022750383 Chr.1: 43981359-43981359 on Build GRCh38
rs765910641 Chr.1: 43981411-43981411 on Build GRCh38
rs371421881 Chr.1: 43981391-43981391 on Build GRCh38
rs766089438 Chr.1: 43981302-43981302 on Build GRCh38
rs551490191 Chr.1: 43981334-43981335 on Build GRCh38
rs980986533 Chr.1: 43981395-43981395 on Build GRCh38
rs776480818 Chr.1: 43981276-43981276 on Build GRCh38
rs747964707 Chr.1: 43981367-43981367 on Build GRCh38
rs748315206 Chr.1: 43981431-43981431 on Build GRCh38
rs882961 Chr.1: 43981412-43981412 on Build GRCh38
rs940311738 Chr.1: 43981463-43981463 on Build GRCh38
rs150890298 Chr.1: 43981438-43981438 on Build GRCh38
rs201975391 Chr.1: 43981432-43981432 on Build GRCh38
rs375270042 Chr.1: 43981441-43981441 on Build GRCh38
rs1018673467 Chr.1: 43981333-43981333 on Build GRCh38
rs759264772 Chr.1: 43981390-43981390 on Build GRCh38
rs368130720 Chr.1: 43981446-43981446 on Build GRCh38
rs987553231 Chr.1: 43981414-43981414 on Build GRCh38
rs780462896 Chr.1: 43981363-43981363 on Build GRCh38
rs761829356 Chr.1: 43981281-43981281 on Build GRCh38
rs200716449 Chr.1: 43981373-43981373 on Build GRCh38
rs149892509 Chr.1: 43981335-43981335 on Build GRCh38
rs374651243 Chr.1: 43981421-43981421 on Build GRCh38
rs756246445 Chr.1: 43981268-43981268 on Build GRCh38
rs749123997 Chr.1: 43981374-43981374 on Build GRCh38
rs145753168 Chr.1: 43981366-43981366 on Build GRCh38
rs2353008 Chr.1: 43981384-43981384 on Build GRCh38

View Full Product Details