Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3091849-3092108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSCAN10
Entrez Gene ID
84891
Gene Name
zinc finger and SCAN domain containing 10
Gene Aliases
ZNF206
UniGene
Hs.334515
Gene Chromosome Location
Chr.16: 3088890-3099317 on Build GRCh38
Transcripts
NM_001282415.1 NM_001282416.1 NM_032805.2 XM_017023791.1 XM_017023792.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1749565 c.472A>G p.K158E Chr.16: 3092076-3092076 on Build GRCh38
7257238 c.465delG p.Q157fs*35 Chr.16: 3092083-3092083 on Build GRCh38
3509210 c.471G>A p.Q157Q Chr.16: 3092077-3092077 on Build GRCh38
399325 c.463G>T p.G155W Chr.16: 3092085-3092085 on Build GRCh38
5510774 c.462G>A p.P154P Chr.16: 3092086-3092086 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs908304296 Chr.16: 3092083-3092083 on Build GRCh38
rs72772563 Chr.16: 3092008-3092008 on Build GRCh38
rs111987520 Chr.16: 3091952-3091952 on Build GRCh38
rs771523800 Chr.16: 3092026-3092026 on Build GRCh38
rs567095403 Chr.16: 3091904-3091911 on Build GRCh38
rs988395268 Chr.16: 3091916-3091916 on Build GRCh38
rs115852910 Chr.16: 3091947-3091947 on Build GRCh38
rs752756761 Chr.16: 3092087-3092087 on Build GRCh38
rs915140870 Chr.16: 3091933-3091933 on Build GRCh38
rs559893386 Chr.16: 3092006-3092006 on Build GRCh38
rs781614089 Chr.16: 3091999-3091999 on Build GRCh38
rs758685684 Chr.16: 3091908-3091908 on Build GRCh38
rs374595812 Chr.16: 3092009-3092009 on Build GRCh38
rs773120329 Chr.16: 3092072-3092072 on Build GRCh38
rs1053839721 Chr.16: 3092018-3092018 on Build GRCh38
rs930987546 Chr.16: 3091907-3091907 on Build GRCh38
rs983812337 Chr.16: 3091939-3091939 on Build GRCh38
rs537231551 Chr.16: 3091885-3091885 on Build GRCh38
rs933581124 Chr.16: 3091964-3091964 on Build GRCh38
rs749555825 Chr.16: 3092011-3092011 on Build GRCh38
rs200755766 Chr.16: 3092007-3092007 on Build GRCh38
rs1051130334 Chr.16: 3091973-3091973 on Build GRCh38
rs199609947 Chr.16: 3092027-3092027 on Build GRCh38
rs370197363 Chr.16: 3092085-3092085 on Build GRCh38
rs746565574 Chr.16: 3092032-3092032 on Build GRCh38
rs766381388 Chr.16: 3092081-3092081 on Build GRCh38
rs923475422 Chr.16: 3091958-3091958 on Build GRCh38
rs141587079 Chr.16: 3092055-3092055 on Build GRCh38
rs939802874 Chr.16: 3091948-3091948 on Build GRCh38
rs768128520 Chr.16: 3092033-3092033 on Build GRCh38
rs150895149 Chr.16: 3092065-3092065 on Build GRCh38
rs747807940 Chr.16: 3092061-3092061 on Build GRCh38
rs774134341 Chr.16: 3092082-3092082 on Build GRCh38
rs1031742164 Chr.16: 3091892-3091892 on Build GRCh38
rs201889093 Chr.16: 3092086-3092086 on Build GRCh38
rs762653280 Chr.16: 3092080-3092080 on Build GRCh38
rs546270126 Chr.16: 3091967-3091967 on Build GRCh38

View Full Product Details