Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 72974554-72974819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFYVE1
Entrez Gene ID
53349
Gene Name
zinc finger FYVE-type containing 1
Gene Aliases
DFCP1, PPP1R172, SR3, TAFF1, ZNFN2A1
UniGene
Hs.335106
Gene Chromosome Location
Chr.14: 72969445-73027212 on Build GRCh38
Transcripts
NM_001281734.1 NM_001281735.1 NM_021260.3 NM_178441.2 XM_017021373.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4052219 c.1987+1G>C p.? Chr.14: 72974778-72974778 on Build GRCh38
5832812 c.1967_1968insT p.E656fs*17 Chr.14: 72974798-72974799 on Build GRCh38
957566 c.1977C>T p.N659N Chr.14: 72974789-72974789 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs182521851 Chr.14: 72974601-72974601 on Build GRCh38
rs772894152 Chr.14: 72974774-72974774 on Build GRCh38
rs1056952853 Chr.14: 72974669-72974669 on Build GRCh38
rs540199986 Chr.14: 72974621-72974621 on Build GRCh38
rs775701799 Chr.14: 72974762-72974762 on Build GRCh38
rs777952568 Chr.14: 72974735-72974735 on Build GRCh38
rs753471508 Chr.14: 72974788-72974788 on Build GRCh38
rs770136019 Chr.14: 72974760-72974760 on Build GRCh38
rs115033268 Chr.14: 72974775-72974775 on Build GRCh38
rs766158995 Chr.14: 72974783-72974783 on Build GRCh38
rs577016043 Chr.14: 72974693-72974693 on Build GRCh38
rs953143386 Chr.14: 72974798-72974798 on Build GRCh38
rs759101141 Chr.14: 72974789-72974789 on Build GRCh38
rs959456076 Chr.14: 72974592-72974592 on Build GRCh38
rs369264081 Chr.14: 72974773-72974773 on Build GRCh38
rs544573576 Chr.14: 72974657-72974657 on Build GRCh38
rs531184214 Chr.14: 72974622-72974622 on Build GRCh38
rs562532149 Chr.14: 72974715-72974715 on Build GRCh38
rs907132480 Chr.14: 72974786-72974786 on Build GRCh38
rs35189121 Chr.14: 72974784-72974785 on Build GRCh38
rs564958686 Chr.14: 72974654-72974654 on Build GRCh38
rs117850671 Chr.14: 72974701-72974701 on Build GRCh38
rs371331449 Chr.14: 72974796-72974796 on Build GRCh38
rs747070212 Chr.14: 72974746-72974746 on Build GRCh38
rs910860061 Chr.14: 72974710-72974710 on Build GRCh38
rs924228804 Chr.14: 72974680-72974680 on Build GRCh38
rs763584103 Chr.14: 72974772-72974772 on Build GRCh38
rs770895509 Chr.14: 72974625-72974625 on Build GRCh38
rs376709641 Chr.14: 72974776-72974776 on Build GRCh38
rs771164912 Chr.14: 72974753-72974753 on Build GRCh38
rs572885746 Chr.14: 72974609-72974609 on Build GRCh38
rs990892410 Chr.14: 72974593-72974593 on Build GRCh38
rs188388322 Chr.14: 72974757-72974757 on Build GRCh38
rs758267961 Chr.14: 72974731-72974731 on Build GRCh38

View Full Product Details