Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 15678693-15678964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC171
Entrez Gene ID
203238
Gene Name
coiled-coil domain containing 171
Gene Aliases
C9orf93, bA536D16.1, bA778P13.1
UniGene
Hs.17267
Gene Chromosome Location
Chr.9: 15552874-16062431 on Build GRCh38
Transcripts
NM_173550.2 XM_005251397.2 XM_005251398.3 XM_005251399.4 XM_011517788.2 XM_011517791.2 XM_011517796.2 XM_017014431.1 XM_017014432.1 XM_017014433.1 XM_017014434.1 XM_017014435.1 XM_017014436.1 XM_017014437.1 XM_017014438.1 XM_017014439.1 XM_017014440.1 XM_017014441.1 XM_017014442.1 XM_017014443.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1283861 c.1141G>C p.V381L Chr.9: 15678822-15678822 on Build GRCh38
3906288 c.1172A>G p.Y391C Chr.9: 15678853-15678853 on Build GRCh38
3906289 c.1204G>A p.E402K Chr.9: 15678885-15678885 on Build GRCh38
3906287 c.1117G>T p.E373* Chr.9: 15678798-15678798 on Build GRCh38
6054979 c.1133A>C p.Q378P Chr.9: 15678814-15678814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758588500 Chr.9: 15678784-15678784 on Build GRCh38
rs896004580 Chr.9: 15678738-15678738 on Build GRCh38
rs564678608 Chr.9: 15678844-15678844 on Build GRCh38
rs367869682 Chr.9: 15678917-15678917 on Build GRCh38
rs191503435 Chr.9: 15678891-15678891 on Build GRCh38
rs771261283 Chr.9: 15678934-15678934 on Build GRCh38
rs760083621 Chr.9: 15678855-15678855 on Build GRCh38
rs1008769190 Chr.9: 15678846-15678846 on Build GRCh38
rs1010380798 Chr.9: 15678767-15678767 on Build GRCh38
rs766740699 Chr.9: 15678757-15678757 on Build GRCh38
rs574212746 Chr.9: 15678923-15678923 on Build GRCh38
rs752729635 Chr.9: 15678871-15678871 on Build GRCh38
rs764161319 Chr.9: 15678878-15678878 on Build GRCh38
rs780162778 Chr.9: 15678897-15678897 on Build GRCh38
rs768144600 Chr.9: 15678839-15678839 on Build GRCh38
rs746729978 Chr.9: 15678803-15678803 on Build GRCh38
rs887329470 Chr.9: 15678920-15678920 on Build GRCh38
rs756082815 Chr.9: 15678893-15678893 on Build GRCh38
rs756991077 Chr.9: 15678886-15678886 on Build GRCh38
rs370605119 Chr.9: 15678807-15678807 on Build GRCh38
rs543500235 Chr.9: 15678818-15678818 on Build GRCh38
rs149253983 Chr.9: 15678823-15678823 on Build GRCh38
rs765356612 Chr.9: 15678772-15678772 on Build GRCh38
rs370235683 Chr.9: 15678822-15678822 on Build GRCh38
rs372876267 Chr.9: 15678931-15678931 on Build GRCh38
rs765380667 Chr.9: 15678798-15678798 on Build GRCh38
rs1057366997 Chr.9: 15678736-15678736 on Build GRCh38
rs996158614 Chr.9: 15678744-15678744 on Build GRCh38
rs1039827869 Chr.9: 15678864-15678864 on Build GRCh38
rs765270352 Chr.9: 15678870-15678870 on Build GRCh38
rs138368957 Chr.9: 15678790-15678790 on Build GRCh38
rs755045737 Chr.9: 15678762-15678762 on Build GRCh38
rs781185148 Chr.9: 15678888-15678888 on Build GRCh38
rs200523149 Chr.9: 15678806-15678806 on Build GRCh38
rs371974266 Chr.9: 15678927-15678927 on Build GRCh38
rs951014562 Chr.9: 15678770-15678770 on Build GRCh38
rs752011926 Chr.9: 15678937-15678937 on Build GRCh38
rs758524253 Chr.9: 15678875-15678875 on Build GRCh38
rs761124321 Chr.9: 15678735-15678735 on Build GRCh38
rs116499856 Chr.9: 15678848-15678848 on Build GRCh38
rs1027273260 Chr.9: 15678760-15678760 on Build GRCh38
rs920370987 Chr.9: 15678910-15678910 on Build GRCh38
rs560190847 Chr.9: 15678852-15678852 on Build GRCh38
rs1041669633 Chr.9: 15678851-15678851 on Build GRCh38
rs777107054 Chr.9: 15678853-15678853 on Build GRCh38
rs200026340 Chr.9: 15678915-15678915 on Build GRCh38
rs945310882 Chr.9: 15678850-15678850 on Build GRCh38
rs752893392 Chr.9: 15678778-15678778 on Build GRCh38
rs546941505 Chr.9: 15678887-15678887 on Build GRCh38
rs751809583 Chr.9: 15678879-15678879 on Build GRCh38

View Full Product Details