Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160841341-160841588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD244
Entrez Gene ID
51744
Gene Name
CD244 molecule
Gene Aliases
2B4, NAIL, NKR2B4, Nmrk, SLAMF4
UniGene
Hs.157872
Gene Chromosome Location
Chr.1: 160830160-160862902 on Build GRCh38
Transcripts
NM_001166663.1 NM_001166664.1 NM_016382.3 XM_011509621.2 XM_011509622.2 XM_011509623.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5668906 c.380-2A>T p.? Chr.1: 160841487-160841487 on Build GRCh38
898382 c.397C>T p.R133C Chr.1: 160841468-160841468 on Build GRCh38
7138354 c.379G>A p.E127K Chr.1: 160841584-160841584 on Build GRCh38
4973176 c.482C>G p.S161C Chr.1: 160841383-160841383 on Build GRCh38
4973175 c.482C>G p.S161C Chr.1: 160841383-160841383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373331921 Chr.1: 160841468-160841468 on Build GRCh38
rs771876926 Chr.1: 160841516-160841516 on Build GRCh38
rs573600543 Chr.1: 160841389-160841389 on Build GRCh38
rs770643711 Chr.1: 160841508-160841508 on Build GRCh38
rs756376571 Chr.1: 160841485-160841485 on Build GRCh38
rs755036224 Chr.1: 160841467-160841467 on Build GRCh38
rs745765975 Chr.1: 160841513-160841513 on Build GRCh38
rs760448230 Chr.1: 160841398-160841398 on Build GRCh38
rs752048374 Chr.1: 160841463-160841463 on Build GRCh38
rs750877965 Chr.1: 160841410-160841410 on Build GRCh38
rs370978919 Chr.1: 160841471-160841471 on Build GRCh38
rs559404051 Chr.1: 160841403-160841403 on Build GRCh38
rs574301343 Chr.1: 160841448-160841448 on Build GRCh38
rs771548413 Chr.1: 160841392-160841392 on Build GRCh38
rs920073430 Chr.1: 160841391-160841391 on Build GRCh38
rs542223030 Chr.1: 160841394-160841394 on Build GRCh38
rs965814280 Chr.1: 160841472-160841472 on Build GRCh38
rs572603366 Chr.1: 160841512-160841512 on Build GRCh38
rs775118207 Chr.1: 160841535-160841535 on Build GRCh38
rs774339579 Chr.1: 160841368-160841368 on Build GRCh38
rs771045132 Chr.1: 160841550-160841553 on Build GRCh38
rs746507662 Chr.1: 160841536-160841536 on Build GRCh38
rs200475714 Chr.1: 160841441-160841441 on Build GRCh38
rs770911534 Chr.1: 160841364-160841364 on Build GRCh38
rs374641198 Chr.1: 160841501-160841501 on Build GRCh38

View Full Product Details