Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2250350-2250601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMH
Entrez Gene ID
268
Gene Name
anti-Mullerian hormone
Gene Aliases
MIF, MIS
UniGene
Hs.112432
Gene Chromosome Location
Chr.19: 2249114-2252073 on Build GRCh38
Transcripts
NM_000479.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5691309 c.451C>T p.P151S Chr.19: 2250375-2250375 on Build GRCh38
5660849 c.475C>T p.P159S Chr.19: 2250399-2250399 on Build GRCh38
5706940 c.484G>A p.A162T Chr.19: 2250408-2250408 on Build GRCh38
1392112 c.509C>A p.P170H Chr.19: 2250433-2250433 on Build GRCh38
3756561 c.546G>A p.P182P Chr.19: 2250470-2250470 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547180609 Chr.19: 2250524-2250524 on Build GRCh38
rs773178911 Chr.19: 2250385-2250385 on Build GRCh38
rs17854573 Chr.19: 2250470-2250470 on Build GRCh38
rs936428725 Chr.19: 2250397-2250397 on Build GRCh38
rs372744500 Chr.19: 2250494-2250494 on Build GRCh38
rs761857057 Chr.19: 2250484-2250485 on Build GRCh38
rs753191322 Chr.19: 2250399-2250399 on Build GRCh38
rs866519862 Chr.19: 2250414-2250414 on Build GRCh38
rs8112524 Chr.19: 2250529-2250529 on Build GRCh38
rs775588008 Chr.19: 2250489-2250489 on Build GRCh38
rs768585333 Chr.19: 2250370-2250370 on Build GRCh38
rs774430982 Chr.19: 2250480-2250480 on Build GRCh38
rs749049690 Chr.19: 2250472-2250472 on Build GRCh38
rs748063248 Chr.19: 2250454-2250454 on Build GRCh38
rs567844561 Chr.19: 2250490-2250490 on Build GRCh38
rs754233174 Chr.19: 2250409-2250409 on Build GRCh38
rs762070367 Chr.19: 2250449-2250450 on Build GRCh38
rs747222239 Chr.19: 2250546-2250546 on Build GRCh38
rs907222158 Chr.19: 2250432-2250432 on Build GRCh38
rs371874189 Chr.19: 2250424-2250424 on Build GRCh38
rs539338966 Chr.19: 2250538-2250538 on Build GRCh38
rs765703485 Chr.19: 2250513-2250513 on Build GRCh38
rs764611221 Chr.19: 2250491-2250491 on Build GRCh38
rs187807834 Chr.19: 2250396-2250396 on Build GRCh38
rs374588581 Chr.19: 2250390-2250390 on Build GRCh38
rs763192254 Chr.19: 2250388-2250388 on Build GRCh38
rs756662915 Chr.19: 2250402-2250402 on Build GRCh38
rs941604570 Chr.19: 2250435-2250435 on Build GRCh38
rs867455699 Chr.19: 2250400-2250400 on Build GRCh38
rs780894513 Chr.19: 2250436-2250436 on Build GRCh38
rs942147657 Chr.19: 2250523-2250523 on Build GRCh38
rs557221831 Chr.19: 2250541-2250541 on Build GRCh38
rs774592796 Chr.19: 2250388-2250401 on Build GRCh38
rs757570984 Chr.19: 2250507-2250507 on Build GRCh38
rs975692513 Chr.19: 2250444-2250444 on Build GRCh38
rs1051486104 Chr.19: 2250488-2250488 on Build GRCh38
rs751597455 Chr.19: 2250394-2250394 on Build GRCh38
rs767563790 Chr.19: 2250483-2250483 on Build GRCh38
rs529706314 Chr.19: 2250461-2250461 on Build GRCh38
rs778082272 Chr.19: 2250404-2250404 on Build GRCh38
rs144093812 Chr.19: 2250500-2250500 on Build GRCh38
rs367783341 Chr.19: 2250395-2250395 on Build GRCh38
rs768050453 Chr.19: 2250426-2250426 on Build GRCh38
rs370532523 Chr.19: 2250375-2250375 on Build GRCh38
rs779588404 Chr.19: 2250425-2250425 on Build GRCh38
rs1045987276 Chr.19: 2250432-2250432 on Build GRCh38
rs767836187 Chr.19: 2250475-2250475 on Build GRCh38
rs558873107 Chr.19: 2250553-2250553 on Build GRCh38
rs371965734 Chr.19: 2250379-2250379 on Build GRCh38
rs759859203 Chr.19: 2250481-2250481 on Build GRCh38
rs200523942 Chr.19: 2250477-2250477 on Build GRCh38

View Full Product Details