Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42226721-42226994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM87A
Entrez Gene ID
25963
Gene Name
transmembrane protein 87A
Gene Aliases
-
UniGene
Hs.511138
Gene Chromosome Location
Chr.15: 42210452-42273584 on Build GRCh38
Transcripts
NM_001286487.1 NM_015497.4 XM_005254287.1 XM_006720468.2 XM_011521443.2 XM_017022067.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5769113 c.1300-7A>G p.? Chr.15: 42226916-42226916 on Build GRCh38
4994650 c.1382G>A p.R461Q Chr.15: 42226827-42226827 on Build GRCh38
6015718 c.1364T>G p.V455G Chr.15: 42226845-42226845 on Build GRCh38
4564677 c.1349_1350CC>TT p.S450F Chr.15: 42226859-42226860 on Build GRCh38
6019194 c.1319T>G p.V440G Chr.15: 42226890-42226890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773002976 Chr.15: 42226940-42226940 on Build GRCh38
rs866710996 Chr.15: 42226853-42226853 on Build GRCh38
rs946442951 Chr.15: 42226960-42226960 on Build GRCh38
rs747668054 Chr.15: 42226927-42226927 on Build GRCh38
rs539628249 Chr.15: 42226865-42226865 on Build GRCh38
rs1041515699 Chr.15: 42226928-42226928 on Build GRCh38
rs763424097 Chr.15: 42226857-42226857 on Build GRCh38
rs781184005 Chr.15: 42226925-42226925 on Build GRCh38
rs772984430 Chr.15: 42226812-42226812 on Build GRCh38
rs888327370 Chr.15: 42226966-42226966 on Build GRCh38
rs746794420 Chr.15: 42226908-42226908 on Build GRCh38
rs1028113884 Chr.15: 42226957-42226957 on Build GRCh38
rs752140251 Chr.15: 42226867-42226867 on Build GRCh38
rs137984068 Chr.15: 42226902-42226902 on Build GRCh38
rs377475368 Chr.15: 42226879-42226879 on Build GRCh38
rs538440452 Chr.15: 42226773-42226773 on Build GRCh38
rs756184652 Chr.15: 42226886-42226886 on Build GRCh38
rs779660502 Chr.15: 42226903-42226903 on Build GRCh38
rs751579474 Chr.15: 42226791-42226791 on Build GRCh38
rs199851830 Chr.15: 42226774-42226774 on Build GRCh38
rs765047427 Chr.15: 42226798-42226798 on Build GRCh38
rs963864445 Chr.15: 42226842-42226842 on Build GRCh38
rs749458668 Chr.15: 42226899-42226899 on Build GRCh38
rs113004234 Chr.15: 42226807-42226807 on Build GRCh38
rs373502445 Chr.15: 42226872-42226872 on Build GRCh38
rs374676715 Chr.15: 42226764-42226764 on Build GRCh38
rs761351432 Chr.15: 42226790-42226790 on Build GRCh38
rs778029392 Chr.15: 42226890-42226890 on Build GRCh38
rs369913019 Chr.15: 42226777-42226777 on Build GRCh38
rs993489146 Chr.15: 42226742-42226742 on Build GRCh38
rs768512518 Chr.15: 42226916-42226916 on Build GRCh38
rs762655659 Chr.15: 42226950-42226950 on Build GRCh38
rs1678988 Chr.15: 42226938-42226938 on Build GRCh38

View Full Product Details