Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128178238-128178509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIZ1
Entrez Gene ID
25792
Gene Name
CDKN1A interacting zinc finger protein 1
Gene Aliases
LSFR1, NP94, ZNF356
UniGene
Hs.212395
Gene Chromosome Location
Chr.9: 128166065-128204383 on Build GRCh38
Transcripts
NM_001131015.1 NM_001131016.1 NM_001131017.1 NM_001131018.1 NM_001257975.1 NM_001257976.1 NM_012127.2 XM_005251888.3 XM_005251891.3 XM_005251892.3 XM_005251893.4 XM_006717039.3 XM_011518481.1 XM_011518482.2 XM_011518483.1 XM_017014594.1 XM_017014595.1 XM_017014596.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7150114 c.1587_1591delAGAAT p.E530fs*1 Chr.9: 128178398-128178402 on Build GRCh38
7150113 c.1587_1591delAGAAT p.E530fs*1 Chr.9: 128178398-128178402 on Build GRCh38
4812140 c.1586G>C p.G529A Chr.9: 128178403-128178403 on Build GRCh38
4907571 c.1605G>A p.A535A Chr.9: 128178384-128178384 on Build GRCh38
3326490 c.1560T>G p.N520K Chr.9: 128178429-128178429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758712379 Chr.9: 128178479-128178479 on Build GRCh38
rs911820183 Chr.9: 128178286-128178286 on Build GRCh38
rs935823276 Chr.9: 128178341-128178341 on Build GRCh38
rs767099878 Chr.9: 128178423-128178423 on Build GRCh38
rs1013117113 Chr.9: 128178346-128178346 on Build GRCh38
rs866370286 Chr.9: 128178424-128178424 on Build GRCh38
rs977364710 Chr.9: 128178323-128178323 on Build GRCh38
rs778098389 Chr.9: 128178487-128178487 on Build GRCh38
rs781714721 Chr.9: 128178264-128178264 on Build GRCh38
rs773721220 Chr.9: 128178375-128178375 on Build GRCh38
rs971527949 Chr.9: 128178378-128178378 on Build GRCh38
rs755827271 Chr.9: 128178448-128178448 on Build GRCh38
rs986507950 Chr.9: 128178260-128178260 on Build GRCh38
rs555555042 Chr.9: 128178415-128178415 on Build GRCh38
rs915639548 Chr.9: 128178398-128178398 on Build GRCh38
rs753772317 Chr.9: 128178462-128178462 on Build GRCh38
rs1017158261 Chr.9: 128178290-128178290 on Build GRCh38
rs1004474800 Chr.9: 128178266-128178266 on Build GRCh38
rs754934873 Chr.9: 128178466-128178466 on Build GRCh38
rs45497192 Chr.9: 128178351-128178351 on Build GRCh38
rs748443356 Chr.9: 128178335-128178335 on Build GRCh38
rs760065353 Chr.9: 128178385-128178385 on Build GRCh38
rs773847029 Chr.9: 128178403-128178403 on Build GRCh38
rs750241334 Chr.9: 128178438-128178438 on Build GRCh38
rs746348226 Chr.9: 128178316-128178316 on Build GRCh38
rs886076503 Chr.9: 128178261-128178261 on Build GRCh38
rs550697095 Chr.9: 128178384-128178384 on Build GRCh38
rs535553816 Chr.9: 128178379-128178379 on Build GRCh38
rs924474245 Chr.9: 128178338-128178338 on Build GRCh38
rs948405233 Chr.9: 128178413-128178413 on Build GRCh38
rs763568377 Chr.9: 128178395-128178395 on Build GRCh38
rs966318056 Chr.9: 128178288-128178288 on Build GRCh38
rs766167859 Chr.9: 128178457-128178457 on Build GRCh38
rs1031642422 Chr.9: 128178459-128178459 on Build GRCh38
rs779205828 Chr.9: 128178478-128178478 on Build GRCh38

View Full Product Details