Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 114628056-114628329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM268
Entrez Gene ID
203197
Gene Name
transmembrane protein 268
Gene Aliases
C9orf91
UniGene
Hs.522357
Gene Chromosome Location
Chr.9: 114603897-114646423 on Build GRCh38
Transcripts
NM_153045.3 XM_006716997.1 XM_011518350.1 XM_011518351.2 XM_011518352.1 XM_011518353.1 XM_011518354.1 XM_011518355.1 XM_017014429.1 XM_017014430.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
356249 c.385A>C p.N129H Chr.9: 114628161-114628161 on Build GRCh38
6209821 c.325-10C>T p.? Chr.9: 114628091-114628091 on Build GRCh38
5626331 c.457G>A p.E153K Chr.9: 114628233-114628233 on Build GRCh38
319158 c.393G>T p.W131C Chr.9: 114628169-114628169 on Build GRCh38
1104426 c.451G>A p.V151I Chr.9: 114628227-114628227 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540092762 Chr.9: 114628163-114628163 on Build GRCh38
rs143980128 Chr.9: 114628199-114628199 on Build GRCh38
rs148625500 Chr.9: 114628249-114628249 on Build GRCh38
rs147884610 Chr.9: 114628280-114628281 on Build GRCh38
rs925830089 Chr.9: 114628193-114628193 on Build GRCh38
rs752097380 Chr.9: 114628223-114628223 on Build GRCh38
rs764121914 Chr.9: 114628295-114628295 on Build GRCh38
rs972516517 Chr.9: 114628116-114628116 on Build GRCh38
rs201972863 Chr.9: 114628081-114628081 on Build GRCh38
rs767552985 Chr.9: 114628198-114628198 on Build GRCh38
rs763876815 Chr.9: 114628227-114628227 on Build GRCh38
rs756013892 Chr.9: 114628288-114628288 on Build GRCh38
rs760336432 Chr.9: 114628294-114628294 on Build GRCh38
rs773356275 Chr.9: 114628191-114628191 on Build GRCh38
rs756714077 Chr.9: 114628244-114628244 on Build GRCh38
rs1049939070 Chr.9: 114628301-114628301 on Build GRCh38
rs752496400 Chr.9: 114628278-114628279 on Build GRCh38
rs578122296 Chr.9: 114628114-114628114 on Build GRCh38
rs563856909 Chr.9: 114628256-114628256 on Build GRCh38
rs750515493 Chr.9: 114628226-114628226 on Build GRCh38
rs544080323 Chr.9: 114628140-114628140 on Build GRCh38
rs746155666 Chr.9: 114628290-114628290 on Build GRCh38
rs528126436 Chr.9: 114628302-114628302 on Build GRCh38
rs755987374 Chr.9: 114628088-114628088 on Build GRCh38
rs928680995 Chr.9: 114628264-114628264 on Build GRCh38
rs369630298 Chr.9: 114628190-114628190 on Build GRCh38
rs781398677 Chr.9: 114628181-114628181 on Build GRCh38
rs747405443 Chr.9: 114628162-114628162 on Build GRCh38
rs753407916 Chr.9: 114628229-114628229 on Build GRCh38
rs1019297669 Chr.9: 114628078-114628079 on Build GRCh38
rs778105415 Chr.9: 114628267-114628267 on Build GRCh38
rs759814908 Chr.9: 114628213-114628213 on Build GRCh38
rs749786039 Chr.9: 114628268-114628268 on Build GRCh38
rs777405472 Chr.9: 114628101-114628101 on Build GRCh38
rs767474362 Chr.9: 114628216-114628216 on Build GRCh38
rs143983393 Chr.9: 114628185-114628185 on Build GRCh38
rs767392872 Chr.9: 114628078-114628078 on Build GRCh38
rs781624654 Chr.9: 114628253-114628253 on Build GRCh38
rs115691052 Chr.9: 114628293-114628293 on Build GRCh38
rs751873443 Chr.9: 114628173-114628173 on Build GRCh38
rs146075825 Chr.9: 114628274-114628274 on Build GRCh38
rs1041386561 Chr.9: 114628184-114628184 on Build GRCh38
rs758042204 Chr.9: 114628251-114628251 on Build GRCh38
rs769048740 Chr.9: 114628168-114628168 on Build GRCh38
rs372528111 Chr.9: 114628196-114628196 on Build GRCh38
rs778486904 Chr.9: 114628159-114628159 on Build GRCh38

View Full Product Details