Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 81017215-81017452 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPN
Entrez Gene ID
55839
Gene Name
centromere protein N
Gene Aliases
BM039, C16orf60, CENP-N, ICEN32
UniGene
Hs.726537
Gene Chromosome Location
Chr.16: 81006498-81033107 on Build GRCh38
Transcripts
NM_001100624.2 NM_001100625.2 NM_001270473.1 NM_001270474.1 NM_018455.5 XM_006721236.3 XM_017023456.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6626663 c.248G>T p.W83L Chr.16: 81017356-81017356 on Build GRCh38
6626664 c.248G>T p.W83L Chr.16: 81017356-81017356 on Build GRCh38
4416329 c.225A>G p.Q75Q Chr.16: 81017333-81017333 on Build GRCh38
5013054 c.230A>G p.H77R Chr.16: 81017338-81017338 on Build GRCh38
6626665 c.248G>T p.W83L Chr.16: 81017356-81017356 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774349900 Chr.16: 81017429-81017429 on Build GRCh38
rs948864527 Chr.16: 81017321-81017321 on Build GRCh38
rs767148583 Chr.16: 81017359-81017359 on Build GRCh38
rs759209838 Chr.16: 81017294-81017294 on Build GRCh38
rs1044157265 Chr.16: 81017331-81017331 on Build GRCh38
rs776364668 Chr.16: 81017308-81017308 on Build GRCh38
rs200572330 Chr.16: 81017398-81017398 on Build GRCh38
rs765047210 Chr.16: 81017354-81017354 on Build GRCh38
rs778922505 Chr.16: 81017279-81017279 on Build GRCh38
rs929566271 Chr.16: 81017258-81017258 on Build GRCh38
rs769290478 Chr.16: 81017290-81017290 on Build GRCh38
rs1013437864 Chr.16: 81017309-81017309 on Build GRCh38
rs778243677 Chr.16: 81017396-81017396 on Build GRCh38
rs117418344 Chr.16: 81017333-81017333 on Build GRCh38
rs764092622 Chr.16: 81017343-81017343 on Build GRCh38
rs532597905 Chr.16: 81017381-81017381 on Build GRCh38
rs748190925 Chr.16: 81017372-81017372 on Build GRCh38
rs371472854 Chr.16: 81017427-81017427 on Build GRCh38
rs771333609 Chr.16: 81017403-81017403 on Build GRCh38
rs764873691 Chr.16: 81017329-81017329 on Build GRCh38
rs748673336 Chr.16: 81017281-81017281 on Build GRCh38
rs763638658 Chr.16: 81017428-81017428 on Build GRCh38
rs1057503665 Chr.16: 81017274-81017274 on Build GRCh38
rs769549703 Chr.16: 81017411-81017411 on Build GRCh38
rs761093434 Chr.16: 81017293-81017293 on Build GRCh38
rs376874847 Chr.16: 81017299-81017299 on Build GRCh38
rs759046186 Chr.16: 81017326-81017326 on Build GRCh38
rs143205259 Chr.16: 81017361-81017361 on Build GRCh38
rs768783741 Chr.16: 81017276-81017276 on Build GRCh38
rs751378474 Chr.16: 81017345-81017345 on Build GRCh38
rs368992165 Chr.16: 81017406-81017406 on Build GRCh38
rs779010537 Chr.16: 81017368-81017368 on Build GRCh38
rs935939 Chr.16: 81017360-81017360 on Build GRCh38
rs757088255 Chr.16: 81017353-81017353 on Build GRCh38
rs148364313 Chr.16: 81017288-81017288 on Build GRCh38
rs775180407 Chr.16: 81017416-81017416 on Build GRCh38
rs773044631 Chr.16: 81017424-81017424 on Build GRCh38

View Full Product Details