Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133424280-133424545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS13
Entrez Gene ID
11093
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 13
Gene Aliases
ADAM-TS13, ADAMTS-13, C9orf8, VWFCP, vWF-CP
UniGene
Hs.131433
Gene Chromosome Location
Chr.9: 133414339-133459403 on Build GRCh38
Transcripts
NM_139025.4 NM_139026.4 NM_139027.4 NR_024514.2 XM_017014232.1 XM_017014233.1 XM_017014235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5548088 c.173-10delC p.? Chr.9: 133424311-133424311 on Build GRCh38
4394420 c.186C>T p.S62S Chr.9: 133424334-133424334 on Build GRCh38
1106426 c.330C>T p.I110I Chr.9: 133424478-133424478 on Build GRCh38
1106424 c.176G>A p.R59H Chr.9: 133424324-133424324 on Build GRCh38
5166607 c.261C>T p.A87A Chr.9: 133424409-133424409 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782149039 Chr.9: 133424420-133424420 on Build GRCh38
rs782677352 Chr.9: 133424327-133424327 on Build GRCh38
rs970382362 Chr.9: 133424489-133424489 on Build GRCh38
rs781838309 Chr.9: 133424303-133424314 on Build GRCh38
rs142107133 Chr.9: 133424478-133424478 on Build GRCh38
rs756898709 Chr.9: 133424418-133424418 on Build GRCh38
rs587712720 Chr.9: 133424380-133424380 on Build GRCh38
rs368684532 Chr.9: 133424486-133424486 on Build GRCh38
rs781788692 Chr.9: 133424438-133424438 on Build GRCh38
rs148644959 Chr.9: 133424389-133424389 on Build GRCh38
rs782456900 Chr.9: 133424331-133424332 on Build GRCh38
rs782049664 Chr.9: 133424514-133424514 on Build GRCh38
rs782158346 Chr.9: 133424305-133424305 on Build GRCh38
rs782716712 Chr.9: 133424453-133424453 on Build GRCh38
rs781864642 Chr.9: 133424454-133424454 on Build GRCh38
rs387906345 Chr.9: 133424439-133424467 on Build GRCh38
rs782681039 Chr.9: 133424465-133424465 on Build GRCh38
rs782576304 Chr.9: 133424350-133424350 on Build GRCh38
rs781795441 Chr.9: 133424315-133424315 on Build GRCh38
rs1055586573 Chr.9: 133424405-133424405 on Build GRCh38
rs139735640 Chr.9: 133424323-133424323 on Build GRCh38
rs782215999 Chr.9: 133424490-133424490 on Build GRCh38
rs782188171 Chr.9: 133424381-133424381 on Build GRCh38
rs782680569 Chr.9: 133424329-133424329 on Build GRCh38
rs782220406 Chr.9: 133424361-133424361 on Build GRCh38
rs782744028 Chr.9: 133424515-133424515 on Build GRCh38
rs1029207345 Chr.9: 133424306-133424306 on Build GRCh38
rs587611213 Chr.9: 133424395-133424395 on Build GRCh38
rs782453359 Chr.9: 133424341-133424341 on Build GRCh38
rs782548827 Chr.9: 133424344-133424349 on Build GRCh38
rs281875302 Chr.9: 133424410-133424410 on Build GRCh38
rs782777667 Chr.9: 133424409-133424409 on Build GRCh38
rs370929256 Chr.9: 133424324-133424324 on Build GRCh38
rs375370257 Chr.9: 133424333-133424333 on Build GRCh38
rs370263552 Chr.9: 133424432-133424432 on Build GRCh38
rs200841843 Chr.9: 133424386-133424386 on Build GRCh38
rs377683598 Chr.9: 133424314-133424314 on Build GRCh38
rs782483941 Chr.9: 133424445-133424445 on Build GRCh38
rs142366230 Chr.9: 133424372-133424372 on Build GRCh38
rs782203933 Chr.9: 133424451-133424452 on Build GRCh38
rs35410697 Chr.9: 133424303-133424303 on Build GRCh38
rs781984753 Chr.9: 133424492-133424492 on Build GRCh38
rs782488906 Chr.9: 133424461-133424461 on Build GRCh38
rs782291909 Chr.9: 133424506-133424506 on Build GRCh38
rs782316384 Chr.9: 133424480-133424481 on Build GRCh38
rs781948066 Chr.9: 133424524-133424524 on Build GRCh38
rs782302107 Chr.9: 133424375-133424375 on Build GRCh38
rs1036424000 Chr.9: 133424300-133424300 on Build GRCh38
rs373857849 Chr.9: 133424513-133424513 on Build GRCh38
rs143856697 Chr.9: 133424373-133424373 on Build GRCh38
rs901814856 Chr.9: 133424302-133424302 on Build GRCh38
rs281875297 Chr.9: 133424385-133424385 on Build GRCh38
rs781853881 Chr.9: 133424344-133424355 on Build GRCh38
rs121908469 Chr.9: 133424452-133424452 on Build GRCh38
rs121908467 Chr.9: 133424434-133424434 on Build GRCh38
rs782397762 Chr.9: 133424491-133424491 on Build GRCh38
rs980887226 Chr.9: 133424496-133424496 on Build GRCh38
rs375415632 Chr.9: 133424479-133424479 on Build GRCh38
rs370611753 Chr.9: 133424326-133424326 on Build GRCh38
rs367627378 Chr.9: 133424379-133424379 on Build GRCh38
rs782035751 Chr.9: 133424415-133424415 on Build GRCh38
rs782400931 Chr.9: 133424511-133424511 on Build GRCh38
rs781958663 Chr.9: 133424390-133424390 on Build GRCh38
rs782101639 Chr.9: 133424320-133424320 on Build GRCh38
rs782306107 Chr.9: 133424343-133424344 on Build GRCh38

View Full Product Details