Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 4082831-4083076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STIM1
Entrez Gene ID
6786
Gene Name
stromal interaction molecule 1
Gene Aliases
D11S4896E, GOK, IMD10, STRMK, TAM, TAM1
UniGene
Hs.501735
Gene Chromosome Location
Chr.11: 3855703-4093210 on Build GRCh38
Transcripts
NM_001277961.1 NM_001277962.1 NM_003156.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1604535 c.1185C>G p.H395Q Chr.11: 4082929-4082929 on Build GRCh38
6791839 c.1143G>T p.E381D Chr.11: 4082887-4082887 on Build GRCh38
5838518 c.1138G>A p.A380T Chr.11: 4082882-4082882 on Build GRCh38
6444229 c.1229T>C p.L410P Chr.11: 4082973-4082973 on Build GRCh38
926855 c.1186G>A p.V396M Chr.11: 4082930-4082930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs10767828 Chr.11: 4082857-4082857 on Build GRCh38
rs533834581 Chr.11: 4082858-4082858 on Build GRCh38
rs750360122 Chr.11: 4082874-4082874 on Build GRCh38
rs1014925401 Chr.11: 4082887-4082887 on Build GRCh38
rs748912734 Chr.11: 4082892-4082892 on Build GRCh38
rs374941454 Chr.11: 4083025-4083025 on Build GRCh38
rs368128270 Chr.11: 4083020-4083020 on Build GRCh38
rs200343814 Chr.11: 4083010-4083010 on Build GRCh38
rs200232554 Chr.11: 4082974-4082974 on Build GRCh38
rs575778741 Chr.11: 4083012-4083012 on Build GRCh38
rs761951593 Chr.11: 4083001-4083001 on Build GRCh38
rs200103292 Chr.11: 4082868-4082868 on Build GRCh38
rs981013850 Chr.11: 4083036-4083036 on Build GRCh38
rs763198159 Chr.11: 4082956-4082956 on Build GRCh38
rs913528464 Chr.11: 4083011-4083011 on Build GRCh38
rs761654472 Chr.11: 4082863-4082863 on Build GRCh38
rs201788011 Chr.11: 4083004-4083004 on Build GRCh38
rs1027836784 Chr.11: 4082929-4082929 on Build GRCh38
rs375604403 Chr.11: 4082991-4082991 on Build GRCh38
rs771694025 Chr.11: 4082924-4082924 on Build GRCh38
rs921124197 Chr.11: 4083051-4083051 on Build GRCh38
rs202226168 Chr.11: 4082953-4082953 on Build GRCh38
rs1018096047 Chr.11: 4082855-4082855 on Build GRCh38
rs767550909 Chr.11: 4082862-4082863 on Build GRCh38
rs201116081 Chr.11: 4082992-4082992 on Build GRCh38
rs753704247 Chr.11: 4083022-4083022 on Build GRCh38
rs58461583 Chr.11: 4082873-4082873 on Build GRCh38
rs143609147 Chr.11: 4082914-4082914 on Build GRCh38
rs960940124 Chr.11: 4082959-4082959 on Build GRCh38
rs969938570 Chr.11: 4082946-4082946 on Build GRCh38
rs751701355 Chr.11: 4082860-4082860 on Build GRCh38
rs542166361 Chr.11: 4082962-4082962 on Build GRCh38
rs767523017 Chr.11: 4083005-4083005 on Build GRCh38
rs542114519 Chr.11: 4083039-4083039 on Build GRCh38
rs201091587 Chr.11: 4082948-4082948 on Build GRCh38
rs757306058 Chr.11: 4082862-4082862 on Build GRCh38
rs756033597 Chr.11: 4083018-4083018 on Build GRCh38
rs201669026 Chr.11: 4082902-4082902 on Build GRCh38
rs755910491 Chr.11: 4082882-4082882 on Build GRCh38
rs146779524 Chr.11: 4083015-4083015 on Build GRCh38
rs768311695 Chr.11: 4082898-4082898 on Build GRCh38
rs760051786 Chr.11: 4082950-4082950 on Build GRCh38
rs59178256 Chr.11: 4083042-4083042 on Build GRCh38
rs779843723 Chr.11: 4082890-4082890 on Build GRCh38
rs992178341 Chr.11: 4082915-4082915 on Build GRCh38

View Full Product Details