Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39364192-39364465 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNGR1
Entrez Gene ID
9145
Gene Name
synaptogyrin 1
Gene Aliases
-
UniGene
Hs.216226
Gene Chromosome Location
Chr.22: 39349949-39385588 on Build GRCh38
Transcripts
NM_004711.4 NM_145731.3 NM_145738.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4582413 c.20G>A p.G7E Chr.22: 39364214-39364214 on Build GRCh38
3151946 c.33C>A p.F11L Chr.22: 39364227-39364227 on Build GRCh38
3964297 c.93C>T p.P31P Chr.22: 39364287-39364287 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761120394 Chr.22: 39364336-39364336 on Build GRCh38
rs531879160 Chr.22: 39364441-39364441 on Build GRCh38
rs769915273 Chr.22: 39364227-39364227 on Build GRCh38
rs527390350 Chr.22: 39364256-39364256 on Build GRCh38
rs962818692 Chr.22: 39364369-39364369 on Build GRCh38
rs772426656 Chr.22: 39364322-39364322 on Build GRCh38
rs751499112 Chr.22: 39364308-39364308 on Build GRCh38
rs964690529 Chr.22: 39364443-39364443 on Build GRCh38
rs531162330 Chr.22: 39364288-39364288 on Build GRCh38
rs987701835 Chr.22: 39364402-39364402 on Build GRCh38
rs773101579 Chr.22: 39364228-39364228 on Build GRCh38
rs993926221 Chr.22: 39364371-39364371 on Build GRCh38
rs200429015 Chr.22: 39364262-39364262 on Build GRCh38
rs75692937 Chr.22: 39364309-39364310 on Build GRCh38
rs754758729 Chr.22: 39364281-39364281 on Build GRCh38
rs201937056 Chr.22: 39364339-39364339 on Build GRCh38
rs151233936 Chr.22: 39364248-39364248 on Build GRCh38
rs368937127 Chr.22: 39364267-39364267 on Build GRCh38
rs774746830 Chr.22: 39364249-39364249 on Build GRCh38
rs938278293 Chr.22: 39364409-39364410 on Build GRCh38
rs762343115 Chr.22: 39364346-39364346 on Build GRCh38
rs771230013 Chr.22: 39364230-39364230 on Build GRCh38
rs759904371 Chr.22: 39364321-39364321 on Build GRCh38
rs751267002 Chr.22: 39364276-39364276 on Build GRCh38
rs536347065 Chr.22: 39364417-39364417 on Build GRCh38
rs779646356 Chr.22: 39364275-39364275 on Build GRCh38
rs77424082 Chr.22: 39364270-39364270 on Build GRCh38
rs541992751 Chr.22: 39364368-39364368 on Build GRCh38
rs769563265 Chr.22: 39364292-39364292 on Build GRCh38
rs568379142 Chr.22: 39364325-39364325 on Build GRCh38
rs777231093 Chr.22: 39364340-39364340 on Build GRCh38
rs759949316 Chr.22: 39364253-39364253 on Build GRCh38
rs749851166 Chr.22: 39364263-39364263 on Build GRCh38
rs979663233 Chr.22: 39364352-39364352 on Build GRCh38
rs774702049 Chr.22: 39364312-39364312 on Build GRCh38
rs73887483 Chr.22: 39364384-39364384 on Build GRCh38
rs547583746 Chr.22: 39364380-39364380 on Build GRCh38
rs781035075 Chr.22: 39364287-39364287 on Build GRCh38
rs549387307 Chr.22: 39364310-39364310 on Build GRCh38
rs370437345 Chr.22: 39364251-39364251 on Build GRCh38
rs777790134 Chr.22: 39364304-39364304 on Build GRCh38

View Full Product Details