Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 119026537-119026766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNPO2
Entrez Gene ID
171024
Gene Name
synaptopodin 2
Gene Aliases
-
UniGene
Hs.655519
Gene Chromosome Location
Chr.4: 118850688-119061247 on Build GRCh38
Transcripts
NM_001128933.2 NM_001128934.2 NM_001286754.1 NM_001286755.1 NM_133477.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1050385 c.361C>T p.R121* Chr.4: 119026730-119026730 on Build GRCh38
7095216 c.292G>A p.E98K Chr.4: 119026661-119026661 on Build GRCh38
5439161 c.258-1G>C p.? Chr.4: 119026626-119026626 on Build GRCh38
6795312 c.290C>A p.S97Y Chr.4: 119026659-119026659 on Build GRCh38
1050383 c.313G>A p.E105K Chr.4: 119026682-119026682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773028849 Chr.4: 119026701-119026701 on Build GRCh38
rs376661222 Chr.4: 119026583-119026583 on Build GRCh38
rs778341529 Chr.4: 119026643-119026643 on Build GRCh38
rs577375744 Chr.4: 119026674-119026674 on Build GRCh38
rs745513035 Chr.4: 119026655-119026655 on Build GRCh38
rs1054543231 Chr.4: 119026720-119026720 on Build GRCh38
rs369836756 Chr.4: 119026738-119026738 on Build GRCh38
rs1002184829 Chr.4: 119026656-119026656 on Build GRCh38
rs373931978 Chr.4: 119026678-119026678 on Build GRCh38
rs769570941 Chr.4: 119026697-119026697 on Build GRCh38
rs763334728 Chr.4: 119026689-119026689 on Build GRCh38
rs764524011 Chr.4: 119026744-119026744 on Build GRCh38
rs368999641 Chr.4: 119026635-119026635 on Build GRCh38
rs887876477 Chr.4: 119026636-119026636 on Build GRCh38
rs143593795 Chr.4: 119026669-119026669 on Build GRCh38
rs746555337 Chr.4: 119026673-119026673 on Build GRCh38
rs755387686 Chr.4: 119026598-119026598 on Build GRCh38
rs766607102 Chr.4: 119026721-119026721 on Build GRCh38
rs751705732 Chr.4: 119026584-119026584 on Build GRCh38
rs763135726 Chr.4: 119026717-119026717 on Build GRCh38
rs752937103 Chr.4: 119026618-119026618 on Build GRCh38
rs546298905 Chr.4: 119026734-119026734 on Build GRCh38
rs781738065 Chr.4: 119026609-119026609 on Build GRCh38
rs757805899 Chr.4: 119026606-119026607 on Build GRCh38
rs779618624 Chr.4: 119026695-119026696 on Build GRCh38
rs147426423 Chr.4: 119026735-119026735 on Build GRCh38
rs758165192 Chr.4: 119026667-119026667 on Build GRCh38
rs112703735 Chr.4: 119026700-119026700 on Build GRCh38
rs1014285920 Chr.4: 119026709-119026709 on Build GRCh38
rs747883620 Chr.4: 119026682-119026682 on Build GRCh38
rs376873485 Chr.4: 119026730-119026730 on Build GRCh38
rs949858137 Chr.4: 119026611-119026611 on Build GRCh38

View Full Product Details