Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26268430-26268703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP85
Entrez Gene ID
64793
Gene Name
centrosomal protein 85
Gene Aliases
CCDC21
UniGene
Hs.109051
Gene Chromosome Location
Chr.1: 26234153-26278810 on Build GRCh38
Transcripts
NM_001281517.2 NM_001319944.1 NM_022778.4 XM_017002105.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5649734 c.1399G>C p.E467Q Chr.1: 26268540-26268540 on Build GRCh38
4671725 c.1366A>G p.N456D Chr.1: 26268507-26268507 on Build GRCh38
680200 c.1386C>T p.C462C Chr.1: 26268527-26268527 on Build GRCh38
2235028 c.1363A>G p.I455V Chr.1: 26268504-26268504 on Build GRCh38
7151013 c.1442G>A p.R481H Chr.1: 26268583-26268583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528156145 Chr.1: 26268508-26268508 on Build GRCh38
rs749075472 Chr.1: 26268549-26268549 on Build GRCh38
rs904552152 Chr.1: 26268505-26268505 on Build GRCh38
rs962713850 Chr.1: 26268659-26268659 on Build GRCh38
rs762262272 Chr.1: 26268636-26268636 on Build GRCh38
rs932809281 Chr.1: 26268604-26268604 on Build GRCh38
rs753394861 Chr.1: 26268619-26268619 on Build GRCh38
rs777474666 Chr.1: 26268544-26268544 on Build GRCh38
rs763772158 Chr.1: 26268610-26268610 on Build GRCh38
rs780039203 Chr.1: 26268515-26268515 on Build GRCh38
rs776546287 Chr.1: 26268666-26268666 on Build GRCh38
rs571259846 Chr.1: 26268565-26268565 on Build GRCh38
rs373834084 Chr.1: 26268583-26268583 on Build GRCh38
rs752991127 Chr.1: 26268466-26268466 on Build GRCh38
rs773770372 Chr.1: 26268456-26268456 on Build GRCh38
rs761056355 Chr.1: 26268679-26268679 on Build GRCh38
rs752405102 Chr.1: 26268490-26268490 on Build GRCh38
rs747297368 Chr.1: 26268528-26268528 on Build GRCh38
rs750882317 Chr.1: 26268507-26268507 on Build GRCh38
rs369268663 Chr.1: 26268487-26268487 on Build GRCh38
rs537012743 Chr.1: 26268605-26268605 on Build GRCh38
rs764427141 Chr.1: 26268496-26268496 on Build GRCh38
rs758902052 Chr.1: 26268657-26268657 on Build GRCh38
rs371019921 Chr.1: 26268513-26268513 on Build GRCh38
rs376750508 Chr.1: 26268643-26268643 on Build GRCh38
rs768908451 Chr.1: 26268533-26268533 on Build GRCh38
rs371564484 Chr.1: 26268493-26268493 on Build GRCh38
rs758484239 Chr.1: 26268511-26268511 on Build GRCh38
rs780165225 Chr.1: 26268668-26268668 on Build GRCh38
rs201534703 Chr.1: 26268503-26268503 on Build GRCh38
rs759144881 Chr.1: 26268457-26268457 on Build GRCh38
rs774364440 Chr.1: 26268558-26268558 on Build GRCh38
rs551503509 Chr.1: 26268550-26268550 on Build GRCh38
rs376096659 Chr.1: 26268494-26268494 on Build GRCh38
rs975122562 Chr.1: 26268661-26268661 on Build GRCh38
rs754209054 Chr.1: 26268502-26268502 on Build GRCh38
rs770875992 Chr.1: 26268455-26268455 on Build GRCh38
rs750933219 Chr.1: 26268645-26268645 on Build GRCh38
rs771779625 Chr.1: 26268582-26268582 on Build GRCh38

View Full Product Details