Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 119990024-119990279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BICDL1
Entrez Gene ID
92558
Gene Name
BICD family like cargo adaptor 1
Gene Aliases
BICDR-1, BICDR1, CCDC64, H_267D11.1
UniGene
Hs.369763
Gene Chromosome Location
Chr.12: 119989806-120094495 on Build GRCh38
Transcripts
NM_207311.2 XM_005253991.4 XM_006719694.3 XM_006719696.3 XM_006719697.3 XM_011538999.2 XM_011539001.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4428629 c.229C>G p.Q77E Chr.12: 119990097-119990097 on Build GRCh38
3703891 c.356C>A p.A119E Chr.12: 119990224-119990224 on Build GRCh38
5739825 c.302C>T p.S101L Chr.12: 119990170-119990170 on Build GRCh38
7241151 c.360G>C p.L120L Chr.12: 119990228-119990228 on Build GRCh38
6437894 c.272C>G p.P91R Chr.12: 119990140-119990140 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868060652 Chr.12: 119990208-119990208 on Build GRCh38
rs776556645 Chr.12: 119990129-119990129 on Build GRCh38
rs764750075 Chr.12: 119990181-119990181 on Build GRCh38
rs573768778 Chr.12: 119990225-119990225 on Build GRCh38
rs201470199 Chr.12: 119990113-119990113 on Build GRCh38
rs753285073 Chr.12: 119990174-119990174 on Build GRCh38
rs757487906 Chr.12: 119990201-119990201 on Build GRCh38
rs560592174 Chr.12: 119990186-119990186 on Build GRCh38
rs756408889 Chr.12: 119990179-119990179 on Build GRCh38
rs1007121921 Chr.12: 119990136-119990136 on Build GRCh38
rs540732478 Chr.12: 119990159-119990159 on Build GRCh38
rs542779128 Chr.12: 119990228-119990228 on Build GRCh38
rs1046134059 Chr.12: 119990111-119990111 on Build GRCh38
rs771846007 Chr.12: 119990104-119990104 on Build GRCh38
rs779095775 Chr.12: 119990077-119990077 on Build GRCh38
rs867210299 Chr.12: 119990231-119990231 on Build GRCh38
rs756565876 Chr.12: 119990054-119990054 on Build GRCh38
rs768546649 Chr.12: 119990117-119990117 on Build GRCh38
rs771297213 Chr.12: 119990061-119990061 on Build GRCh38
rs761415586 Chr.12: 119990142-119990142 on Build GRCh38
rs948658982 Chr.12: 119990226-119990226 on Build GRCh38
rs999953767 Chr.12: 119990063-119990063 on Build GRCh38
rs777824310 Chr.12: 119990056-119990056 on Build GRCh38
rs868637098 Chr.12: 119990045-119990045 on Build GRCh38
rs1015843569 Chr.12: 119990158-119990158 on Build GRCh38
rs772259396 Chr.12: 119990247-119990247 on Build GRCh38
rs917065830 Chr.12: 119990213-119990213 on Build GRCh38
rs925611858 Chr.12: 119990249-119990249 on Build GRCh38
rs954423253 Chr.12: 119990115-119990115 on Build GRCh38
rs776434283 Chr.12: 119990130-119990130 on Build GRCh38
rs749881243 Chr.12: 119990150-119990150 on Build GRCh38
rs745946640 Chr.12: 119990078-119990078 on Build GRCh38
rs752994089 Chr.12: 119990051-119990051 on Build GRCh38
rs892004401 Chr.12: 119990094-119990094 on Build GRCh38
rs749566747 Chr.12: 119990057-119990057 on Build GRCh38
rs778146805 Chr.12: 119990180-119990180 on Build GRCh38
rs949180274 Chr.12: 119990075-119990075 on Build GRCh38
rs762585214 Chr.12: 119990155-119990155 on Build GRCh38
rs567765976 Chr.12: 119990147-119990147 on Build GRCh38
rs1044300350 Chr.12: 119990246-119990246 on Build GRCh38
rs865910809 Chr.12: 119990050-119990050 on Build GRCh38
rs1052814293 Chr.12: 119990090-119990090 on Build GRCh38
rs993050261 Chr.12: 119990207-119990207 on Build GRCh38
rs781730862 Chr.12: 119990049-119990049 on Build GRCh38

View Full Product Details