Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 48046405-48046658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf57
Entrez Gene ID
136288
Gene Name
chromosome 7 open reading frame 57
Gene Aliases
-
UniGene
Hs.258357
Gene Chromosome Location
Chr.7: 48035511-48061297 on Build GRCh38
Transcripts
NM_001100159.2 NM_001267865.1 NM_001267866.1 XM_006715649.3 XM_006715650.2 XM_011515121.1 XM_011515122.2 XM_011515123.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3762706 c.371C>T p.P124L Chr.7: 48046480-48046480 on Build GRCh38
453155 c.406C>G p.Q136E Chr.7: 48046515-48046515 on Build GRCh38
3715744 c.366C>T p.S122S Chr.7: 48046475-48046475 on Build GRCh38
4411556 c.439C>T p.P147S Chr.7: 48046548-48046548 on Build GRCh38
3663226 c.367A>G p.M123V Chr.7: 48046476-48046476 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369582125 Chr.7: 48046499-48046499 on Build GRCh38
rs1028445620 Chr.7: 48046565-48046565 on Build GRCh38
rs758011369 Chr.7: 48046511-48046511 on Build GRCh38
rs201788289 Chr.7: 48046509-48046509 on Build GRCh38
rs971940133 Chr.7: 48046489-48046489 on Build GRCh38
rs372814501 Chr.7: 48046621-48046621 on Build GRCh38
rs768558403 Chr.7: 48046448-48046448 on Build GRCh38
rs368961481 Chr.7: 48046605-48046605 on Build GRCh38
rs768127170 Chr.7: 48046488-48046488 on Build GRCh38
rs758104715 Chr.7: 48046615-48046616 on Build GRCh38
rs753841415 Chr.7: 48046428-48046429 on Build GRCh38
rs150041609 Chr.7: 48046640-48046640 on Build GRCh38
rs758103183 Chr.7: 48046613-48046613 on Build GRCh38
rs201149144 Chr.7: 48046465-48046465 on Build GRCh38
rs781116234 Chr.7: 48046534-48046534 on Build GRCh38
rs778407612 Chr.7: 48046607-48046608 on Build GRCh38
rs1056623485 Chr.7: 48046537-48046537 on Build GRCh38
rs765383564 Chr.7: 48046458-48046458 on Build GRCh38
rs748080242 Chr.7: 48046624-48046624 on Build GRCh38
rs749911913 Chr.7: 48046608-48046608 on Build GRCh38
rs748191370 Chr.7: 48046549-48046549 on Build GRCh38
rs374647757 Chr.7: 48046441-48046441 on Build GRCh38
rs553478065 Chr.7: 48046567-48046567 on Build GRCh38
rs746988343 Chr.7: 48046446-48046446 on Build GRCh38
rs777246386 Chr.7: 48046641-48046646 on Build GRCh38
rs10233232 Chr.7: 48046564-48046564 on Build GRCh38
rs776630651 Chr.7: 48046454-48046454 on Build GRCh38
rs769588265 Chr.7: 48046631-48046631 on Build GRCh38
rs923265166 Chr.7: 48046610-48046610 on Build GRCh38
rs573627346 Chr.7: 48046615-48046615 on Build GRCh38
rs746793015 Chr.7: 48046515-48046515 on Build GRCh38
rs1010121370 Chr.7: 48046541-48046541 on Build GRCh38
rs111480809 Chr.7: 48046554-48046554 on Build GRCh38
rs868833355 Chr.7: 48046559-48046559 on Build GRCh38
rs753195208 Chr.7: 48046596-48046596 on Build GRCh38
rs369752964 Chr.7: 48046560-48046560 on Build GRCh38
rs1017233916 Chr.7: 48046434-48046434 on Build GRCh38
rs781013700 Chr.7: 48046622-48046622 on Build GRCh38
rs764839883 Chr.7: 48046607-48046607 on Build GRCh38
rs954012510 Chr.7: 48046603-48046603 on Build GRCh38
rs377348619 Chr.7: 48046572-48046572 on Build GRCh38
rs754897945 Chr.7: 48046599-48046604 on Build GRCh38
rs779873173 Chr.7: 48046512-48046512 on Build GRCh38
rs2708912 Chr.7: 48046480-48046480 on Build GRCh38
rs1025293633 Chr.7: 48046455-48046455 on Build GRCh38
rs773427716 Chr.7: 48046464-48046464 on Build GRCh38
rs747591930 Chr.7: 48046608-48046608 on Build GRCh38
rs761946232 Chr.7: 48046456-48046456 on Build GRCh38
rs189439693 Chr.7: 48046485-48046485 on Build GRCh38
rs768646082 Chr.7: 48046532-48046532 on Build GRCh38
rs1049327150 Chr.7: 48046508-48046508 on Build GRCh38
rs376223715 Chr.7: 48046492-48046492 on Build GRCh38
rs377253797 Chr.7: 48046639-48046639 on Build GRCh38
rs200430273 Chr.7: 48046553-48046553 on Build GRCh38
rs767919671 Chr.7: 48046593-48046593 on Build GRCh38
rs896762198 Chr.7: 48046538-48046538 on Build GRCh38
rs751359233 Chr.7: 48046616-48046616 on Build GRCh38
rs756730366 Chr.7: 48046496-48046496 on Build GRCh38
rs774918710 Chr.7: 48046562-48046562 on Build GRCh38
rs751945237 Chr.7: 48046481-48046481 on Build GRCh38
rs937708076 Chr.7: 48046527-48046527 on Build GRCh38
rs202087899 Chr.7: 48046436-48046436 on Build GRCh38

View Full Product Details