Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36461528-36461795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN14
Entrez Gene ID
23562
Gene Name
claudin 14
Gene Aliases
DFNB29
UniGene
Hs.660278
Gene Chromosome Location
Chr.21: 36460621-36576569 on Build GRCh38
Transcripts
NM_001146077.1 NM_001146078.2 NM_001146079.1 NM_012130.3 NM_144492.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5897228 c.145G>A p.G49R Chr.21: 36461551-36461551 on Build GRCh38
3405385 c.106G>A p.V36M Chr.21: 36461590-36461590 on Build GRCh38
6026662 c.129C>A p.A43A Chr.21: 36461567-36461567 on Build GRCh38
6526261 c.82C>T p.P28S Chr.21: 36461614-36461614 on Build GRCh38
3972766 c.167G>A p.W56* Chr.21: 36461529-36461529 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150731625 Chr.21: 36461567-36461567 on Build GRCh38
rs140177046 Chr.21: 36461570-36461570 on Build GRCh38
rs117560775 Chr.21: 36461633-36461633 on Build GRCh38
rs557122405 Chr.21: 36461771-36461771 on Build GRCh38
rs899830471 Chr.21: 36461597-36461597 on Build GRCh38
rs387907417 Chr.21: 36461678-36461678 on Build GRCh38
rs779937857 Chr.21: 36461622-36461622 on Build GRCh38
rs764358289 Chr.21: 36461684-36461684 on Build GRCh38
rs868529657 Chr.21: 36461576-36461576 on Build GRCh38
rs768411939 Chr.21: 36461628-36461628 on Build GRCh38
rs1052311905 Chr.21: 36461742-36461742 on Build GRCh38
rs537529014 Chr.21: 36461770-36461770 on Build GRCh38
rs1041031415 Chr.21: 36461604-36461604 on Build GRCh38
rs188479887 Chr.21: 36461752-36461752 on Build GRCh38
rs1044825412 Chr.21: 36461562-36461562 on Build GRCh38
rs150040476 Chr.21: 36461645-36461645 on Build GRCh38
rs141139613 Chr.21: 36461566-36461566 on Build GRCh38
rs532176130 Chr.21: 36461634-36461634 on Build GRCh38
rs769320772 Chr.21: 36461641-36461641 on Build GRCh38
rs886057051 Chr.21: 36461749-36461749 on Build GRCh38
rs202117396 Chr.21: 36461680-36461680 on Build GRCh38
rs911817843 Chr.21: 36461749-36461749 on Build GRCh38
rs560763542 Chr.21: 36461591-36461591 on Build GRCh38
rs368231049 Chr.21: 36461700-36461700 on Build GRCh38
rs886057050 Chr.21: 36461600-36461600 on Build GRCh38
rs387907416 Chr.21: 36461681-36461681 on Build GRCh38
rs146395322 Chr.21: 36461594-36461594 on Build GRCh38
rs533194857 Chr.21: 36461571-36461571 on Build GRCh38
rs780834768 Chr.21: 36461629-36461629 on Build GRCh38
rs374578221 Chr.21: 36461686-36461686 on Build GRCh38
rs762948927 Chr.21: 36461630-36461632 on Build GRCh38
rs758426367 Chr.21: 36461721-36461721 on Build GRCh38
rs750364874 Chr.21: 36461720-36461720 on Build GRCh38
rs549639383 Chr.21: 36461624-36461624 on Build GRCh38
rs754032374 Chr.21: 36461595-36461595 on Build GRCh38
rs757426764 Chr.21: 36461605-36461605 on Build GRCh38
rs758081638 Chr.21: 36461614-36461614 on Build GRCh38
rs982935102 Chr.21: 36461637-36461637 on Build GRCh38
rs548165835 Chr.21: 36461748-36461748 on Build GRCh38
rs149049781 Chr.21: 36461732-36461732 on Build GRCh38
rs752639706 Chr.21: 36461578-36461580 on Build GRCh38
rs947811327 Chr.21: 36461553-36461553 on Build GRCh38
rs956380527 Chr.21: 36461701-36461701 on Build GRCh38
rs778927942 Chr.21: 36461612-36461612 on Build GRCh38
rs387907415 Chr.21: 36461636-36461636 on Build GRCh38
rs142205038 Chr.21: 36461590-36461590 on Build GRCh38
rs113831133 Chr.21: 36461685-36461685 on Build GRCh38
rs980535943 Chr.21: 36461719-36461719 on Build GRCh38
rs369883669 Chr.21: 36461644-36461644 on Build GRCh38
rs759652223 Chr.21: 36461753-36461753 on Build GRCh38
rs1023845285 Chr.21: 36461677-36461677 on Build GRCh38
rs772295911 Chr.21: 36461668-36461668 on Build GRCh38
rs727504989 Chr.21: 36461613-36461613 on Build GRCh38
rs759850341 Chr.21: 36461587-36461587 on Build GRCh38
rs779699092 Chr.21: 36461741-36461741 on Build GRCh38

View Full Product Details