Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 32311866-32312103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZAR1L
Entrez Gene ID
646799
Gene Name
zygote arrest 1-like
Gene Aliases
Z3CXXC7, ZAR2
UniGene
Hs.569254
Gene Chromosome Location
Chr.13: 32303771-32311954 on Build GRCh38
Transcripts
NM_001136571.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4611401 c.12_13insT p.V5fs*53 Chr.13: 32311913-32311914 on Build GRCh38
5376637 c.48G>A p.G16G Chr.13: 32311878-32311878 on Build GRCh38
3981995 c.39G>A p.Q13Q Chr.13: 32311887-32311887 on Build GRCh38
5694086 c.8G>T p.R3L Chr.13: 32311918-32311918 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1040715502 Chr.13: 32311933-32311933 on Build GRCh38
rs1052309722 Chr.13: 32311912-32311912 on Build GRCh38
rs779506896 Chr.13: 32311975-32311975 on Build GRCh38
rs768338220 Chr.13: 32312034-32312034 on Build GRCh38
rs769341314 Chr.13: 32311972-32311972 on Build GRCh38
rs1004878019 Chr.13: 32312064-32312064 on Build GRCh38
rs1015830675 Chr.13: 32312066-32312066 on Build GRCh38
rs746498594 Chr.13: 32311942-32311942 on Build GRCh38
rs766777323 Chr.13: 32312047-32312047 on Build GRCh38
rs906518845 Chr.13: 32311907-32311907 on Build GRCh38
rs571400021 Chr.13: 32311931-32311931 on Build GRCh38
rs538708911 Chr.13: 32311993-32311993 on Build GRCh38
rs376522727 Chr.13: 32311966-32311966 on Build GRCh38
rs143985973 Chr.13: 32312003-32312003 on Build GRCh38
rs375519711 Chr.13: 32311927-32311927 on Build GRCh38
rs915811918 Chr.13: 32311890-32311890 on Build GRCh38
rs998407613 Chr.13: 32312035-32312035 on Build GRCh38
rs952873170 Chr.13: 32312060-32312060 on Build GRCh38
rs9534154 Chr.13: 32312076-32312076 on Build GRCh38
rs201833883 Chr.13: 32311901-32311901 on Build GRCh38
rs950022307 Chr.13: 32311900-32311900 on Build GRCh38
rs772747864 Chr.13: 32311936-32311936 on Build GRCh38
rs903656091 Chr.13: 32311889-32311889 on Build GRCh38
rs75257539 Chr.13: 32311893-32311893 on Build GRCh38
rs753381961 Chr.13: 32311961-32311962 on Build GRCh38

View Full Product Details