Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 116719682-116719953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf49
Entrez Gene ID
79794
Gene Name
chromosome 12 open reading frame 49
Gene Aliases
-
UniGene
Hs.592011
Gene Chromosome Location
Chr.12: 116713320-116738070 on Build GRCh38
Transcripts
NM_024738.2 XM_005253937.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4494268 c.339C>T p.L113L Chr.12: 116719868-116719868 on Build GRCh38
5652260 c.396_397delCG p.E133fs*4 Chr.12: 116719810-116719811 on Build GRCh38
5739817 c.365C>T p.A122V Chr.12: 116719842-116719842 on Build GRCh38
5497236 c.519C>G p.C173W Chr.12: 116719778-116719778 on Build GRCh38
4621577 c.351C>T p.F117F Chr.12: 116719856-116719856 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770784378 Chr.12: 116719861-116719861 on Build GRCh38
rs114256458 Chr.12: 116719851-116719851 on Build GRCh38
rs745725961 Chr.12: 116719784-116719784 on Build GRCh38
rs746926171 Chr.12: 116719804-116719804 on Build GRCh38
rs753909676 Chr.12: 116719734-116719734 on Build GRCh38
rs770571265 Chr.12: 116719918-116719918 on Build GRCh38
rs202158268 Chr.12: 116719812-116719812 on Build GRCh38
rs749582789 Chr.12: 116719830-116719830 on Build GRCh38
rs896305779 Chr.12: 116719923-116719923 on Build GRCh38
rs777265313 Chr.12: 116719927-116719927 on Build GRCh38
rs181497058 Chr.12: 116719811-116719811 on Build GRCh38
rs747710312 Chr.12: 116719904-116719904 on Build GRCh38
rs1001738495 Chr.12: 116719912-116719912 on Build GRCh38
rs772835759 Chr.12: 116719821-116719821 on Build GRCh38
rs767773934 Chr.12: 116719837-116719837 on Build GRCh38
rs374656191 Chr.12: 116719894-116719894 on Build GRCh38
rs138632621 Chr.12: 116719810-116719810 on Build GRCh38
rs779249114 Chr.12: 116719769-116719769 on Build GRCh38
rs758581865 Chr.12: 116719885-116719885 on Build GRCh38
rs781184202 Chr.12: 116719898-116719898 on Build GRCh38
rs746794597 Chr.12: 116719893-116719893 on Build GRCh38
rs199666897 Chr.12: 116719834-116719834 on Build GRCh38
rs560355233 Chr.12: 116719735-116719735 on Build GRCh38
rs779734088 Chr.12: 116719886-116719886 on Build GRCh38
rs771325476 Chr.12: 116719860-116719860 on Build GRCh38
rs371336657 Chr.12: 116719722-116719722 on Build GRCh38
rs1043673227 Chr.12: 116719910-116719910 on Build GRCh38
rs781280193 Chr.12: 116719711-116719711 on Build GRCh38
rs756573910 Chr.12: 116719721-116719721 on Build GRCh38
rs150320799 Chr.12: 116719848-116719848 on Build GRCh38
rs758256986 Chr.12: 116719789-116719789 on Build GRCh38
rs765226688 Chr.12: 116719861-116719861 on Build GRCh38
rs149484642 Chr.12: 116719853-116719853 on Build GRCh38
rs142722974 Chr.12: 116719841-116719841 on Build GRCh38
rs185336717 Chr.12: 116719906-116719906 on Build GRCh38
rs150275614 Chr.12: 116719793-116719793 on Build GRCh38
rs761073433 Chr.12: 116719924-116719924 on Build GRCh38
rs770078946 Chr.12: 116719764-116719764 on Build GRCh38
rs201890321 Chr.12: 116719849-116719849 on Build GRCh38
rs189446899 Chr.12: 116719913-116719913 on Build GRCh38
rs753153840 Chr.12: 116719714-116719714 on Build GRCh38
rs769500444 Chr.12: 116719820-116719820 on Build GRCh38
rs372795724 Chr.12: 116719922-116719922 on Build GRCh38
rs199940069 Chr.12: 116719880-116719880 on Build GRCh38
rs144009690 Chr.12: 116719874-116719874 on Build GRCh38
rs761804514 Chr.12: 116719781-116719786 on Build GRCh38
rs764580049 Chr.12: 116719925-116719925 on Build GRCh38
rs748967611 Chr.12: 116719917-116719917 on Build GRCh38
rs748265648 Chr.12: 116719743-116719743 on Build GRCh38

View Full Product Details