Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127815861-127816088 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENG
Entrez Gene ID
2022
Gene Name
endoglin
Gene Aliases
END, HHT1, ORW1
UniGene
Hs.76753
Gene Chromosome Location
Chr.9: 127815012-127854773 on Build GRCh38
Transcripts
NM_000118.3 NM_001114753.2 NM_001278138.1 NR_136302.1

Gene Symbol
LOC102723566
Entrez Gene ID
102723566
Gene Name
uncharacterized LOC102723566
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.9: 127816066-127822520 on Build GRCh38
Transcripts
NM_000118.3 NM_001114753.2 NM_001278138.1 NR_136302.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5467605 c.1790T>C p.F597S Chr.9: 127816005-127816005 on Build GRCh38
6306459 c.1863C>A p.P621P Chr.9: 127815932-127815932 on Build GRCh38
6370313 c.1780G>T p.G594C Chr.9: 127816015-127816015 on Build GRCh38
5472663 c.1845G>A p.S615S Chr.9: 127815950-127815950 on Build GRCh38
5467606 c.1790T>C p.F597S Chr.9: 127816005-127816005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776826819 Chr.9: 127816040-127816040 on Build GRCh38
rs779115954 Chr.9: 127815909-127815909 on Build GRCh38
rs756018069 Chr.9: 127815900-127815900 on Build GRCh38
rs201031046 Chr.9: 127815945-127815945 on Build GRCh38
rs747196026 Chr.9: 127816036-127816036 on Build GRCh38
rs748191815 Chr.9: 127816029-127816029 on Build GRCh38
rs748422950 Chr.9: 127815913-127815913 on Build GRCh38
rs41358947 Chr.9: 127816001-127816001 on Build GRCh38
rs370943570 Chr.9: 127816025-127816025 on Build GRCh38
rs371104611 Chr.9: 127815891-127815891 on Build GRCh38
rs753536186 Chr.9: 127816069-127816069 on Build GRCh38
rs748184867 Chr.9: 127815886-127815887 on Build GRCh38
rs1021923141 Chr.9: 127815935-127815935 on Build GRCh38
rs756003321 Chr.9: 127816021-127816021 on Build GRCh38
rs374591551 Chr.9: 127816045-127816045 on Build GRCh38
rs546872552 Chr.9: 127816034-127816034 on Build GRCh38
rs148002300 Chr.9: 127815951-127815951 on Build GRCh38
rs759950184 Chr.9: 127815950-127815950 on Build GRCh38
rs766073909 Chr.9: 127816022-127816022 on Build GRCh38
rs747344646 Chr.9: 127815944-127815944 on Build GRCh38
rs201678228 Chr.9: 127816068-127816068 on Build GRCh38
rs767008006 Chr.9: 127816065-127816065 on Build GRCh38
rs34792295 Chr.9: 127815899-127815900 on Build GRCh38
rs373611252 Chr.9: 127816066-127816066 on Build GRCh38
rs201768056 Chr.9: 127816033-127816033 on Build GRCh38
rs147188969 Chr.9: 127815922-127815922 on Build GRCh38
rs373002544 Chr.9: 127815989-127815989 on Build GRCh38
rs200080694 Chr.9: 127816024-127816024 on Build GRCh38
rs768836999 Chr.9: 127815906-127815906 on Build GRCh38
rs779974705 Chr.9: 127815901-127815901 on Build GRCh38
rs373626498 Chr.9: 127815892-127815892 on Build GRCh38
rs150566847 Chr.9: 127816037-127816037 on Build GRCh38
rs868296721 Chr.9: 127816014-127816014 on Build GRCh38
rs750181050 Chr.9: 127815898-127815898 on Build GRCh38
rs749176478 Chr.9: 127815905-127815905 on Build GRCh38
rs761244549 Chr.9: 127816063-127816063 on Build GRCh38
rs562814353 Chr.9: 127815896-127815896 on Build GRCh38
rs891615717 Chr.9: 127815910-127815910 on Build GRCh38
rs760221843 Chr.9: 127816067-127816067 on Build GRCh38
rs773861569 Chr.9: 127816050-127816050 on Build GRCh38
rs746414124 Chr.9: 127816044-127816044 on Build GRCh38
rs750187963 Chr.9: 127816002-127816002 on Build GRCh38
rs143524008 Chr.9: 127815923-127815923 on Build GRCh38
rs753864390 Chr.9: 127816023-127816023 on Build GRCh38

View Full Product Details