Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161118384-161118647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NIT1
Entrez Gene ID
4817
Gene Name
nitrilase 1
Gene Aliases
-
UniGene
Hs.146406
Gene Chromosome Location
Chr.1: 161118072-161125446 on Build GRCh38
Transcripts
NM_001185092.1 NM_001185093.1 NM_001185094.1 NM_005600.2 XM_005245214.2 XM_005245215.4 XM_005245216.3 XM_006711341.3 XM_017001371.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5713567 c.29_30insAG p.E12fs*7 Chr.1: 161118527-161118528 on Build GRCh38
1196767 c.4A>G p.T2A Chr.1: 161118502-161118502 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772516440 Chr.1: 161118452-161118452 on Build GRCh38
rs775929267 Chr.1: 161118459-161118459 on Build GRCh38
rs551456324 Chr.1: 161118467-161118467 on Build GRCh38
rs752905690 Chr.1: 161118486-161118486 on Build GRCh38
rs571394571 Chr.1: 161118469-161118469 on Build GRCh38
rs765423397 Chr.1: 161118599-161118599 on Build GRCh38
rs1030343397 Chr.1: 161118509-161118509 on Build GRCh38
rs971633236 Chr.1: 161118619-161118619 on Build GRCh38
rs947572001 Chr.1: 161118413-161118414 on Build GRCh38
rs777711028 Chr.1: 161118520-161118520 on Build GRCh38
rs532639132 Chr.1: 161118453-161118453 on Build GRCh38
rs1012637413 Chr.1: 161118598-161118598 on Build GRCh38
rs567539661 Chr.1: 161118611-161118611 on Build GRCh38
rs757100174 Chr.1: 161118596-161118596 on Build GRCh38
rs1006809650 Chr.1: 161118590-161118590 on Build GRCh38
rs541894223 Chr.1: 161118605-161118605 on Build GRCh38
rs138523655 Chr.1: 161118502-161118502 on Build GRCh38
rs761276637 Chr.1: 161118460-161118460 on Build GRCh38
rs776174998 Chr.1: 161118503-161118503 on Build GRCh38
rs769269150 Chr.1: 161118490-161118490 on Build GRCh38
rs191022081 Chr.1: 161118575-161118575 on Build GRCh38
rs748841515 Chr.1: 161118451-161118451 on Build GRCh38
rs1041904123 Chr.1: 161118430-161118430 on Build GRCh38
rs778405677 Chr.1: 161118504-161118505 on Build GRCh38
rs902877449 Chr.1: 161118444-161118444 on Build GRCh38
rs539801933 Chr.1: 161118466-161118466 on Build GRCh38

View Full Product Details