Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57029384-57029627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO1A
Entrez Gene ID
4640
Gene Name
myosin IA
Gene Aliases
BBMI, DFNA48, MIHC, MYHL
UniGene
Hs.5394
Gene Chromosome Location
Chr.12: 57028517-57050765 on Build GRCh38
Transcripts
NM_001256041.1 NM_005379.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3463618 c.2748G>A p.L916L Chr.12: 57029564-57029564 on Build GRCh38
7114930 c.2732C>T p.S911F Chr.12: 57029580-57029580 on Build GRCh38
6720128 c.2760T>C p.H920H Chr.12: 57029552-57029552 on Build GRCh38
300236 c.2833G>A p.V945I Chr.12: 57029479-57029479 on Build GRCh38
3676503 c.2850T>C p.D950D Chr.12: 57029462-57029462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202005897 Chr.12: 57029462-57029462 on Build GRCh38
rs759579125 Chr.12: 57029436-57029436 on Build GRCh38
rs780443212 Chr.12: 57029579-57029581 on Build GRCh38
rs573190348 Chr.12: 57029578-57029578 on Build GRCh38
rs760874091 Chr.12: 57029447-57029447 on Build GRCh38
rs201397880 Chr.12: 57029560-57029560 on Build GRCh38
rs774383209 Chr.12: 57029435-57029435 on Build GRCh38
rs375656169 Chr.12: 57029595-57029595 on Build GRCh38
rs771897400 Chr.12: 57029496-57029496 on Build GRCh38
rs148980496 Chr.12: 57029589-57029589 on Build GRCh38
rs866518742 Chr.12: 57029541-57029541 on Build GRCh38
rs772061719 Chr.12: 57029596-57029596 on Build GRCh38
rs777268952 Chr.12: 57029477-57029477 on Build GRCh38
rs755531233 Chr.12: 57029581-57029581 on Build GRCh38
rs772987859 Chr.12: 57029422-57029422 on Build GRCh38
rs774295122 Chr.12: 57029482-57029482 on Build GRCh38
rs370491058 Chr.12: 57029597-57029597 on Build GRCh38
rs748970956 Chr.12: 57029479-57029479 on Build GRCh38
rs758868062 Chr.12: 57029576-57029576 on Build GRCh38
rs984572306 Chr.12: 57029551-57029551 on Build GRCh38
rs750893525 Chr.12: 57029547-57029548 on Build GRCh38
rs759004707 Chr.12: 57029551-57029553 on Build GRCh38
rs760467546 Chr.12: 57029426-57029426 on Build GRCh38
rs764252617 Chr.12: 57029504-57029504 on Build GRCh38
rs773165131 Chr.12: 57029577-57029577 on Build GRCh38
rs747781087 Chr.12: 57029408-57029408 on Build GRCh38
rs762023364 Chr.12: 57029557-57029557 on Build GRCh38
rs765400327 Chr.12: 57029546-57029547 on Build GRCh38
rs372760228 Chr.12: 57029508-57029508 on Build GRCh38
rs754139259 Chr.12: 57029461-57029461 on Build GRCh38
rs958915780 Chr.12: 57029565-57029567 on Build GRCh38
rs770898901 Chr.12: 57029432-57029432 on Build GRCh38
rs747610908 Chr.12: 57029475-57029475 on Build GRCh38
rs764190058 Chr.12: 57029459-57029459 on Build GRCh38
rs750859877 Chr.12: 57029471-57029471 on Build GRCh38
rs755694024 Chr.12: 57029476-57029476 on Build GRCh38
rs750775990 Chr.12: 57029567-57029567 on Build GRCh38
rs752161080 Chr.12: 57029579-57029579 on Build GRCh38
rs184810732 Chr.12: 57029440-57029440 on Build GRCh38
rs866842692 Chr.12: 57029553-57029553 on Build GRCh38
rs121909306 Chr.12: 57029584-57029584 on Build GRCh38
rs1034626773 Chr.12: 57029562-57029562 on Build GRCh38
rs780569775 Chr.12: 57029407-57029407 on Build GRCh38
rs56128678 Chr.12: 57029413-57029413 on Build GRCh38
rs758918100 Chr.12: 57029472-57029472 on Build GRCh38
rs868837386 Chr.12: 57029569-57029569 on Build GRCh38
rs200448145 Chr.12: 57029593-57029593 on Build GRCh38
rs770810873 Chr.12: 57029481-57029481 on Build GRCh38
rs760649045 Chr.12: 57029501-57029501 on Build GRCh38
rs202090975 Chr.12: 57029484-57029484 on Build GRCh38
rs765618377 Chr.12: 57029467-57029467 on Build GRCh38
rs754081858 Chr.12: 57029495-57029495 on Build GRCh38
rs1020910578 Chr.12: 57029547-57029547 on Build GRCh38
rs1054353924 Chr.12: 57029483-57029483 on Build GRCh38
rs756986110 Chr.12: 57029592-57029592 on Build GRCh38
rs764261858 Chr.12: 57029427-57029428 on Build GRCh38
rs775417473 Chr.12: 57029443-57029443 on Build GRCh38
rs762814449 Chr.12: 57029428-57029428 on Build GRCh38
rs780672432 Chr.12: 57029474-57029474 on Build GRCh38
rs777375497 Chr.12: 57029587-57029587 on Build GRCh38
rs761861596 Chr.12: 57029536-57029539 on Build GRCh38

View Full Product Details