Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110517592-110517863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNA10
Entrez Gene ID
3744
Gene Name
potassium voltage-gated channel subfamily A member 10
Gene Aliases
Kcn1, Kv1.8
UniGene
Hs.248140
Gene Chromosome Location
Chr.1: 110517217-110519175 on Build GRCh38
Transcripts
NM_005549.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3471070 c.1195G>A p.D399N Chr.1: 110517593-110517593 on Build GRCh38
6024775 c.1110G>A p.A370A Chr.1: 110517678-110517678 on Build GRCh38
6056357 c.1163T>C p.L388P Chr.1: 110517625-110517625 on Build GRCh38
3471071 c.1068C>T p.R356R Chr.1: 110517720-110517720 on Build GRCh38
6009410 c.1129T>C p.L377L Chr.1: 110517659-110517659 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs930506374 Chr.1: 110517626-110517626 on Build GRCh38
rs968278633 Chr.1: 110517823-110517823 on Build GRCh38
rs566195238 Chr.1: 110517723-110517723 on Build GRCh38
rs373170268 Chr.1: 110517622-110517622 on Build GRCh38
rs760343284 Chr.1: 110517728-110517728 on Build GRCh38
rs376109106 Chr.1: 110517675-110517675 on Build GRCh38
rs187179669 Chr.1: 110517679-110517679 on Build GRCh38
rs897182272 Chr.1: 110517735-110517735 on Build GRCh38
rs749237726 Chr.1: 110517721-110517721 on Build GRCh38
rs781334730 Chr.1: 110517675-110517675 on Build GRCh38
rs775722259 Chr.1: 110517835-110517835 on Build GRCh38
rs201318977 Chr.1: 110517707-110517707 on Build GRCh38
rs45442396 Chr.1: 110517678-110517678 on Build GRCh38
rs974039455 Chr.1: 110517833-110517833 on Build GRCh38
rs190117605 Chr.1: 110517695-110517695 on Build GRCh38
rs148047410 Chr.1: 110517771-110517771 on Build GRCh38
rs767085869 Chr.1: 110517668-110517668 on Build GRCh38
rs201622407 Chr.1: 110517724-110517724 on Build GRCh38
rs779739847 Chr.1: 110517775-110517775 on Build GRCh38
rs778875602 Chr.1: 110517797-110517797 on Build GRCh38
rs570945851 Chr.1: 110517711-110517711 on Build GRCh38
rs923643838 Chr.1: 110517668-110517668 on Build GRCh38
rs771519279 Chr.1: 110517802-110517802 on Build GRCh38
rs747047062 Chr.1: 110517684-110517684 on Build GRCh38
rs375331411 Chr.1: 110517739-110517739 on Build GRCh38
rs200678806 Chr.1: 110517740-110517740 on Build GRCh38
rs370455162 Chr.1: 110517758-110517758 on Build GRCh38
rs781330073 Chr.1: 110517769-110517769 on Build GRCh38
rs374428249 Chr.1: 110517687-110517687 on Build GRCh38
rs770577436 Chr.1: 110517826-110517826 on Build GRCh38
rs763352517 Chr.1: 110517837-110517837 on Build GRCh38
rs761479453 Chr.1: 110517654-110517654 on Build GRCh38
rs147012018 Chr.1: 110517702-110517702 on Build GRCh38
rs115895001 Chr.1: 110517699-110517699 on Build GRCh38
rs773832987 Chr.1: 110517646-110517646 on Build GRCh38
rs146624318 Chr.1: 110517670-110517670 on Build GRCh38
rs749047165 Chr.1: 110517779-110517779 on Build GRCh38
rs886190636 Chr.1: 110517674-110517674 on Build GRCh38
rs34314746 Chr.1: 110517708-110517708 on Build GRCh38
rs776600827 Chr.1: 110517613-110517613 on Build GRCh38
rs778020579 Chr.1: 110517680-110517680 on Build GRCh38
rs762947400 Chr.1: 110517627-110517627 on Build GRCh38
rs775327009 Chr.1: 110517712-110517712 on Build GRCh38
rs367703875 Chr.1: 110517801-110517801 on Build GRCh38
rs748376767 Chr.1: 110517774-110517774 on Build GRCh38
rs755353258 Chr.1: 110517671-110517671 on Build GRCh38
rs769786265 Chr.1: 110517624-110517624 on Build GRCh38
rs772753869 Chr.1: 110517807-110517807 on Build GRCh38
rs757471815 Chr.1: 110517764-110517764 on Build GRCh38
rs911249844 Chr.1: 110517804-110517804 on Build GRCh38
rs772803737 Chr.1: 110517725-110517725 on Build GRCh38
rs760266671 Chr.1: 110517812-110517812 on Build GRCh38
rs763636135 Chr.1: 110517638-110517638 on Build GRCh38
rs781216125 Chr.1: 110517692-110517692 on Build GRCh38
rs768589259 Chr.1: 110517794-110517794 on Build GRCh38
rs1339702 Chr.1: 110517681-110517681 on Build GRCh38
rs146859968 Chr.1: 110517625-110517625 on Build GRCh38
rs370709737 Chr.1: 110517757-110517757 on Build GRCh38
rs768535829 Chr.1: 110517722-110517722 on Build GRCh38

View Full Product Details