Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49706070-49706325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPT1C
Entrez Gene ID
126129
Gene Name
carnitine palmitoyltransferase 1C
Gene Aliases
CATL1, CPT1-B, CPT1P, CPTI-B, CPTIC, SPG73
UniGene
Hs.112195
Gene Chromosome Location
Chr.19: 49690672-49713731 on Build GRCh38
Transcripts
NM_001136052.2 NM_001199752.2 NM_001199753.1 NM_152359.2 NR_108072.1 XM_005258505.2 XM_005258506.4 XM_006723009.3 XM_011526438.2 XM_011526439.2 XM_011526440.2 XM_017026265.1 XM_017026266.1 XM_017026267.1 XM_017026268.1 XM_017026269.1 XM_017026270.1 XM_017026271.1 XM_017026272.1 XM_017026273.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5594450 c.1198C>T p.Q400* Chr.19: 49706268-49706268 on Build GRCh38
3707297 c.1171G>A p.A391T Chr.19: 49706241-49706241 on Build GRCh38
3536920 c.1210G>A p.A404T Chr.19: 49706280-49706280 on Build GRCh38
3893068 c.1209G>A p.E403E Chr.19: 49706279-49706279 on Build GRCh38
3536919 c.1129G>A p.E377K Chr.19: 49706073-49706073 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746174740 Chr.19: 49706112-49706112 on Build GRCh38
rs556848512 Chr.19: 49706127-49706127 on Build GRCh38
rs984363196 Chr.19: 49706109-49706109 on Build GRCh38
rs927675700 Chr.19: 49706169-49706169 on Build GRCh38
rs370573255 Chr.19: 49706218-49706218 on Build GRCh38
rs779130037 Chr.19: 49706108-49706109 on Build GRCh38
rs746293416 Chr.19: 49706189-49706189 on Build GRCh38
rs749799725 Chr.19: 49706100-49706100 on Build GRCh38
rs769810756 Chr.19: 49706236-49706236 on Build GRCh38
rs751926613 Chr.19: 49706303-49706303 on Build GRCh38
rs145807606 Chr.19: 49706237-49706237 on Build GRCh38
rs368217937 Chr.19: 49706220-49706220 on Build GRCh38
rs547155396 Chr.19: 49706177-49706177 on Build GRCh38
rs761825439 Chr.19: 49706131-49706131 on Build GRCh38
rs1018765350 Chr.19: 49706266-49706266 on Build GRCh38
rs757810539 Chr.19: 49706153-49706153 on Build GRCh38
rs764179585 Chr.19: 49706275-49706275 on Build GRCh38
rs780577623 Chr.19: 49706301-49706301 on Build GRCh38
rs752905653 Chr.19: 49706145-49706145 on Build GRCh38
rs758822585 Chr.19: 49706190-49706190 on Build GRCh38
rs779355537 Chr.19: 49706110-49706110 on Build GRCh38
rs371325260 Chr.19: 49706228-49706228 on Build GRCh38
rs753809818 Chr.19: 49706150-49706150 on Build GRCh38
rs939103812 Chr.19: 49706171-49706171 on Build GRCh38
rs1053903749 Chr.19: 49706245-49706245 on Build GRCh38
rs201971986 Chr.19: 49706094-49706094 on Build GRCh38
rs770711301 Chr.19: 49706247-49706247 on Build GRCh38
rs374293545 Chr.19: 49706219-49706219 on Build GRCh38
rs748661451 Chr.19: 49706121-49706121 on Build GRCh38
rs372494717 Chr.19: 49706132-49706132 on Build GRCh38
rs61748956 Chr.19: 49706290-49706290 on Build GRCh38
rs764165874 Chr.19: 49706146-49706146 on Build GRCh38
rs760907133 Chr.19: 49706269-49706269 on Build GRCh38
rs767780477 Chr.19: 49706144-49706144 on Build GRCh38
rs763203574 Chr.19: 49706092-49706092 on Build GRCh38
rs755112332 Chr.19: 49706109-49706115 on Build GRCh38
rs376287276 Chr.19: 49706272-49706272 on Build GRCh38
rs771886232 Chr.19: 49706114-49706114 on Build GRCh38
rs772610465 Chr.19: 49706129-49706129 on Build GRCh38
rs892129620 Chr.19: 49706179-49706179 on Build GRCh38
rs768276178 Chr.19: 49706125-49706125 on Build GRCh38
rs775260321 Chr.19: 49706118-49706118 on Build GRCh38
rs774069280 Chr.19: 49706250-49706250 on Build GRCh38
rs758907193 Chr.19: 49706291-49706291 on Build GRCh38
rs201085193 Chr.19: 49706196-49706196 on Build GRCh38
rs181596690 Chr.19: 49706285-49706285 on Build GRCh38
rs761772845 Chr.19: 49706279-49706279 on Build GRCh38
rs746575054 Chr.19: 49706124-49706124 on Build GRCh38
rs762624670 Chr.19: 49706133-49706133 on Build GRCh38
rs757644561 Chr.19: 49706107-49706107 on Build GRCh38
rs376625740 Chr.19: 49706186-49706186 on Build GRCh38
rs948164950 Chr.19: 49706137-49706137 on Build GRCh38
rs572265604 Chr.19: 49706276-49706276 on Build GRCh38

View Full Product Details