Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 124791232-124791505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NKAIN2
Entrez Gene ID
154215
Gene Name
Na+/K+ transporting ATPase interacting 2
Gene Aliases
FAM77B, NKAIP2, TCBA, TCBA1
UniGene
Hs.656604
Gene Chromosome Location
Chr.6: 123803840-124825652 on Build GRCh38
Transcripts
NM_001040214.2 NM_001300737.1 NM_001300738.1 NM_001300740.1 NM_153355.4 XM_011535501.2 XM_011535502.2 XM_011535503.2 XM_017010318.1 XM_017010319.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6536053 c.445T>G p.Y149D Chr.6: 124791354-124791354 on Build GRCh38
3620138 c.486C>T p.F162F Chr.6: 124791350-124791350 on Build GRCh38
6536052 c.490T>G p.Y164D Chr.6: 124791354-124791354 on Build GRCh38
3620139 c.520G>A p.E174K Chr.6: 124791384-124791384 on Build GRCh38
1072882 c.495C>A p.A165A Chr.6: 124791359-124791359 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749567983 Chr.6: 124791395-124791395 on Build GRCh38
rs182153928 Chr.6: 124791376-124791376 on Build GRCh38
rs375002233 Chr.6: 124791339-124791339 on Build GRCh38
rs550796318 Chr.6: 124791382-124791382 on Build GRCh38
rs962539733 Chr.6: 124791481-124791481 on Build GRCh38
rs371264703 Chr.6: 124791332-124791332 on Build GRCh38
rs542163848 Chr.6: 124791333-124791333 on Build GRCh38
rs755923952 Chr.6: 124791343-124791343 on Build GRCh38
rs775360392 Chr.6: 124791428-124791428 on Build GRCh38
rs375289455 Chr.6: 124791312-124791312 on Build GRCh38
rs369014684 Chr.6: 124791326-124791326 on Build GRCh38
rs777080414 Chr.6: 124791311-124791311 on Build GRCh38
rs770172414 Chr.6: 124791319-124791319 on Build GRCh38
rs773640204 Chr.6: 124791392-124791392 on Build GRCh38
rs769171484 Chr.6: 124791306-124791306 on Build GRCh38
rs760662189 Chr.6: 124791430-124791430 on Build GRCh38
rs200968368 Chr.6: 124791291-124791291 on Build GRCh38
rs753505485 Chr.6: 124791347-124791347 on Build GRCh38
rs767605395 Chr.6: 124791337-124791337 on Build GRCh38
rs368496264 Chr.6: 124791422-124791422 on Build GRCh38
rs996608908 Chr.6: 124791381-124791381 on Build GRCh38
rs562018477 Chr.6: 124791356-124791356 on Build GRCh38
rs948839787 Chr.6: 124791293-124791293 on Build GRCh38
rs780197488 Chr.6: 124791357-124791357 on Build GRCh38
rs877058 Chr.6: 124791468-124791468 on Build GRCh38
rs1031218872 Chr.6: 124791330-124791330 on Build GRCh38
rs947773875 Chr.6: 124791270-124791270 on Build GRCh38
rs1021816697 Chr.6: 124791414-124791414 on Build GRCh38
rs767125195 Chr.6: 124791427-124791427 on Build GRCh38
rs755017479 Chr.6: 124791373-124791373 on Build GRCh38
rs368821168 Chr.6: 124791289-124791289 on Build GRCh38
rs373170400 Chr.6: 124791420-124791420 on Build GRCh38
rs759169166 Chr.6: 124791423-124791423 on Build GRCh38
rs753685873 Chr.6: 124791440-124791440 on Build GRCh38
rs146625615 Chr.6: 124791296-124791296 on Build GRCh38
rs747121176 Chr.6: 124791365-124791365 on Build GRCh38
rs993202563 Chr.6: 124791476-124791476 on Build GRCh38
rs528842717 Chr.6: 124791327-124791327 on Build GRCh38
rs772797610 Chr.6: 124791321-124791321 on Build GRCh38
rs1022957765 Chr.6: 124791480-124791480 on Build GRCh38
rs1031904753 Chr.6: 124791462-124791462 on Build GRCh38
rs770741124 Chr.6: 124791417-124791417 on Build GRCh38
rs570691217 Chr.6: 124791433-124791433 on Build GRCh38
rs759079174 Chr.6: 124791335-124791335 on Build GRCh38

View Full Product Details