Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 107931758-107932017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2U1
Entrez Gene ID
113612
Gene Name
cytochrome P450 family 2 subfamily U member 1
Gene Aliases
P450TEC, SPG49, SPG56
UniGene
Hs.109087
Gene Chromosome Location
Chr.4: 107931549-107953461 on Build GRCh38
Transcripts
NM_183075.2 XM_005262717.2 XM_005262720.2 NR_125929.1

Gene Symbol
LOC101929595
Entrez Gene ID
101929595
Gene Name
uncharacterized LOC101929595
Gene Aliases
-
UniGene
Hs.678014
Gene Chromosome Location
Chr.4: 107863479-107932119 on Build GRCh38
Transcripts
NM_183075.2 XM_005262717.2 XM_005262720.2 NR_125929.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6410244 c.140G>A p.G47D Chr.4: 107931783-107931783 on Build GRCh38
3769765 c.268A>C p.R90R Chr.4: 107931911-107931911 on Build GRCh38
4883879 c.264C>A p.S88R Chr.4: 107931907-107931907 on Build GRCh38
6289641 c.351C>T p.I117I Chr.4: 107931994-107931994 on Build GRCh38
6371640 c.293C>A p.P98H Chr.4: 107931936-107931936 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021034246 Chr.4: 107931985-107931985 on Build GRCh38
rs762434691 Chr.4: 107931802-107931802 on Build GRCh38
rs147506864 Chr.4: 107931951-107931951 on Build GRCh38
rs768079432 Chr.4: 107931811-107931811 on Build GRCh38
rs944597665 Chr.4: 107931858-107931858 on Build GRCh38
rs201411336 Chr.4: 107931804-107931804 on Build GRCh38
rs145192019 Chr.4: 107931868-107931868 on Build GRCh38
rs758365928 Chr.4: 107931810-107931810 on Build GRCh38
rs766889023 Chr.4: 107931986-107931986 on Build GRCh38
rs1010706663 Chr.4: 107931829-107931829 on Build GRCh38
rs373291849 Chr.4: 107931927-107931927 on Build GRCh38
rs757245518 Chr.4: 107931777-107931777 on Build GRCh38
rs929055316 Chr.4: 107931816-107931816 on Build GRCh38
rs747138982 Chr.4: 107931845-107931845 on Build GRCh38
rs775828461 Chr.4: 107931852-107931852 on Build GRCh38
rs748573097 Chr.4: 107931956-107931956 on Build GRCh38
rs982216763 Chr.4: 107931922-107931922 on Build GRCh38
rs768868832 Chr.4: 107931878-107931878 on Build GRCh38
rs763547405 Chr.4: 107931979-107931979 on Build GRCh38
rs997228256 Chr.4: 107931940-107931940 on Build GRCh38
rs920319565 Chr.4: 107931813-107931813 on Build GRCh38
rs760339720 Chr.4: 107931994-107931994 on Build GRCh38
rs780202261 Chr.4: 107931838-107931838 on Build GRCh38
rs937661813 Chr.4: 107931787-107931787 on Build GRCh38
rs770267485 Chr.4: 107931965-107931965 on Build GRCh38
rs773635818 Chr.4: 107931973-107931973 on Build GRCh38
rs900437680 Chr.4: 107931883-107931883 on Build GRCh38
rs969327084 Chr.4: 107931915-107931915 on Build GRCh38
rs745929667 Chr.4: 107931807-107931807 on Build GRCh38
rs920309099 Chr.4: 107931991-107931991 on Build GRCh38
rs779042395 Chr.4: 107931798-107931798 on Build GRCh38
rs1053725607 Chr.4: 107931945-107931945 on Build GRCh38

View Full Product Details