Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 94459990-94460239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCT
Entrez Gene ID
1638
Gene Name
dopachrome tautomerase
Gene Aliases
TRP-2, TYRP2
UniGene
Hs.301865
Gene Chromosome Location
Chr.13: 94437304-94550265 on Build GRCh38
Transcripts
NM_001129889.2 NM_001322182.1 NM_001322183.1 NM_001322184.1 NM_001322185.1 NM_001322186.1 NM_001922.4 XM_011521049.1 XM_017020401.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7306812 c.1158C>T p.A386A Chr.13: 94460112-94460112 on Build GRCh38
5478603 c.1109A>G p.N370S Chr.13: 94460161-94460161 on Build GRCh38
4438662 c.1154C>T p.S385L Chr.13: 94460116-94460116 on Build GRCh38
1249835 c.1141G>A p.A381T Chr.13: 94460129-94460129 on Build GRCh38
3814141 c.1092A>T p.Q364H Chr.13: 94460178-94460178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs968888631 Chr.13: 94460030-94460030 on Build GRCh38
rs543257150 Chr.13: 94460098-94460098 on Build GRCh38
rs149962333 Chr.13: 94460076-94460076 on Build GRCh38
rs769800507 Chr.13: 94460170-94460170 on Build GRCh38
rs1036884836 Chr.13: 94460213-94460213 on Build GRCh38
rs146134025 Chr.13: 94460107-94460107 on Build GRCh38
rs770964531 Chr.13: 94460138-94460138 on Build GRCh38
rs373770110 Chr.13: 94460080-94460080 on Build GRCh38
rs370360434 Chr.13: 94460172-94460172 on Build GRCh38
rs752557378 Chr.13: 94460087-94460087 on Build GRCh38
rs759555045 Chr.13: 94460085-94460085 on Build GRCh38
rs930236055 Chr.13: 94460046-94460046 on Build GRCh38
rs777376777 Chr.13: 94460211-94460211 on Build GRCh38
rs779110813 Chr.13: 94460112-94460112 on Build GRCh38
rs759340915 Chr.13: 94460158-94460158 on Build GRCh38
rs376196399 Chr.13: 94460111-94460111 on Build GRCh38
rs773213304 Chr.13: 94460066-94460066 on Build GRCh38
rs762820660 Chr.13: 94460176-94460176 on Build GRCh38
rs751227543 Chr.13: 94460203-94460203 on Build GRCh38
rs763841511 Chr.13: 94460097-94460097 on Build GRCh38
rs760761856 Chr.13: 94460067-94460067 on Build GRCh38
rs374075015 Chr.13: 94460206-94460206 on Build GRCh38
rs369959297 Chr.13: 94460130-94460130 on Build GRCh38
rs748273066 Chr.13: 94460044-94460044 on Build GRCh38
rs748554211 Chr.13: 94460114-94460114 on Build GRCh38
rs776819456 Chr.13: 94460139-94460139 on Build GRCh38
rs943729069 Chr.13: 94460094-94460094 on Build GRCh38
rs762456842 Chr.13: 94460174-94460174 on Build GRCh38
rs977585435 Chr.13: 94460032-94460032 on Build GRCh38
rs932577790 Chr.13: 94460115-94460115 on Build GRCh38
rs772373837 Chr.13: 94460057-94460057 on Build GRCh38
rs757947053 Chr.13: 94460093-94460093 on Build GRCh38
rs772228009 Chr.13: 94460129-94460129 on Build GRCh38
rs764917052 Chr.13: 94460086-94460086 on Build GRCh38
rs764047768 Chr.13: 94460191-94460191 on Build GRCh38
rs138973003 Chr.13: 94460145-94460145 on Build GRCh38

View Full Product Details