Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154765824-154766093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKC1
Entrez Gene ID
1736
Gene Name
dyskerin pseudouridine synthase 1
Gene Aliases
CBF5, DKC, DKCX, NAP57, NOLA4, XAP101
UniGene
Hs.4747
Gene Chromosome Location
Chr.X: 154762742-154777689 on Build GRCh38
Transcripts
NM_001142463.2 NM_001288747.1 NM_001363.4 NR_110021.1 NR_110022.1 NR_110023.1

SNP Detail

SNP ID Chromosome Location
rs199422245 Chr.X: 154765939-154765939 on Build GRCh38
rs371006539 Chr.X: 154766015-154766015 on Build GRCh38
rs782531283 Chr.X: 154765873-154765873 on Build GRCh38
rs782455540 Chr.X: 154765862-154765862 on Build GRCh38
rs781900698 Chr.X: 154765869-154765869 on Build GRCh38
rs374742619 Chr.X: 154766039-154766039 on Build GRCh38
rs199422244 Chr.X: 154765935-154765935 on Build GRCh38
rs121912294 Chr.X: 154765949-154765950 on Build GRCh38
rs782385464 Chr.X: 154766018-154766018 on Build GRCh38
rs372393271 Chr.X: 154765857-154765857 on Build GRCh38
rs781988696 Chr.X: 154766032-154766032 on Build GRCh38
rs1055229197 Chr.X: 154765962-154765962 on Build GRCh38
rs782655180 Chr.X: 154765883-154765883 on Build GRCh38
rs897094414 Chr.X: 154765979-154765979 on Build GRCh38
rs988960799 Chr.X: 154766069-154766069 on Build GRCh38
rs376783942 Chr.X: 154765895-154765895 on Build GRCh38
rs782259834 Chr.X: 154766062-154766062 on Build GRCh38
rs782202263 Chr.X: 154765994-154765994 on Build GRCh38
rs965503303 Chr.X: 154766061-154766061 on Build GRCh38
rs782625316 Chr.X: 154766035-154766035 on Build GRCh38
rs782599928 Chr.X: 154765972-154765972 on Build GRCh38
rs121912301 Chr.X: 154765929-154765929 on Build GRCh38
rs372724285 Chr.X: 154765866-154765866 on Build GRCh38
rs781821680 Chr.X: 154765856-154765856 on Build GRCh38
rs1019814715 Chr.X: 154766042-154766042 on Build GRCh38
rs782236324 Chr.X: 154765894-154765894 on Build GRCh38
rs121912306 Chr.X: 154765949-154765949 on Build GRCh38
rs121912297 Chr.X: 154765931-154765931 on Build GRCh38
rs1036932842 Chr.X: 154765954-154765954 on Build GRCh38
rs782317228 Chr.X: 154765997-154765997 on Build GRCh38

View Full Product Details