Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 9150008-9150281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND5A
Entrez Gene ID
23258
Gene Name
DENN domain containing 5A
Gene Aliases
RAB6IP1
UniGene
Hs.501857
Gene Chromosome Location
Chr.11: 9138825-9265335 on Build GRCh38
Transcripts
NM_001243254.1 NM_015213.3 XM_005252832.1 XM_011519953.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5906878 c.2735+5G>A p.? Chr.11: 9150076-9150076 on Build GRCh38
5739172 c.2649G>T p.K883N Chr.11: 9150167-9150167 on Build GRCh38
3454311 c.2622C>T p.I874I Chr.11: 9150194-9150194 on Build GRCh38
4483351 c.2693C>T p.S898F Chr.11: 9150123-9150123 on Build GRCh38
6235014 c.2607-10C>T p.? Chr.11: 9150219-9150219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756424392 Chr.11: 9150240-9150240 on Build GRCh38
rs777221054 Chr.11: 9150034-9150034 on Build GRCh38
rs774654447 Chr.11: 9150161-9150161 on Build GRCh38
rs77389721 Chr.11: 9150152-9150152 on Build GRCh38
rs778436007 Chr.11: 9150103-9150103 on Build GRCh38
rs751975269 Chr.11: 9150067-9150067 on Build GRCh38
rs758385384 Chr.11: 9150115-9150115 on Build GRCh38
rs924495710 Chr.11: 9150143-9150143 on Build GRCh38
rs7123970 Chr.11: 9150228-9150228 on Build GRCh38
rs746828979 Chr.11: 9150123-9150123 on Build GRCh38
rs760075600 Chr.11: 9150176-9150176 on Build GRCh38
rs373547821 Chr.11: 9150194-9150194 on Build GRCh38
rs370969726 Chr.11: 9150068-9150068 on Build GRCh38
rs754957309 Chr.11: 9150236-9150236 on Build GRCh38
rs945502256 Chr.11: 9150089-9150089 on Build GRCh38
rs767936667 Chr.11: 9150184-9150184 on Build GRCh38
rs371135128 Chr.11: 9150238-9150238 on Build GRCh38
rs768517099 Chr.11: 9150124-9150124 on Build GRCh38
rs199810820 Chr.11: 9150047-9150047 on Build GRCh38
rs964696330 Chr.11: 9150042-9150042 on Build GRCh38
rs141251799 Chr.11: 9150158-9150158 on Build GRCh38
rs749276734 Chr.11: 9150157-9150157 on Build GRCh38
rs775802702 Chr.11: 9150191-9150191 on Build GRCh38
rs770107497 Chr.11: 9150150-9150150 on Build GRCh38
rs749880027 Chr.11: 9150113-9150113 on Build GRCh38
rs781169020 Chr.11: 9150237-9150237 on Build GRCh38
rs764823802 Chr.11: 9150200-9150200 on Build GRCh38
rs779817963 Chr.11: 9150117-9150117 on Build GRCh38
rs781117137 Chr.11: 9150134-9150134 on Build GRCh38
rs765140486 Chr.11: 9150232-9150232 on Build GRCh38
rs766357051 Chr.11: 9150235-9150235 on Build GRCh38
rs763740178 Chr.11: 9150069-9150069 on Build GRCh38
rs753547538 Chr.11: 9150074-9150074 on Build GRCh38
rs748445195 Chr.11: 9150142-9150142 on Build GRCh38
rs773418488 Chr.11: 9150038-9150038 on Build GRCh38
rs756899375 Chr.11: 9150083-9150083 on Build GRCh38
rs749983105 Chr.11: 9150214-9150214 on Build GRCh38
rs766771609 Chr.11: 9150053-9150053 on Build GRCh38
rs897401738 Chr.11: 9150160-9150160 on Build GRCh38
rs112168308 Chr.11: 9150050-9150050 on Build GRCh38

View Full Product Details