Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82024193-82024460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC45
Entrez Gene ID
201255
Gene Name
leucine rich repeat containing 45
Gene Aliases
-
UniGene
Hs.143774
Gene Chromosome Location
Chr.17: 82023252-82031151 on Build GRCh38
Transcripts
NM_144999.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6703908 c.265C>T p.R89C Chr.17: 82024322-82024322 on Build GRCh38
6703907 c.266G>A p.R89H Chr.17: 82024323-82024323 on Build GRCh38
6082522 c.271C>T p.L91L Chr.17: 82024328-82024328 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374049967 Chr.17: 82024289-82024289 on Build GRCh38
rs368332146 Chr.17: 82024351-82024351 on Build GRCh38
rs747449583 Chr.17: 82024363-82024363 on Build GRCh38
rs55732858 Chr.17: 82024401-82024401 on Build GRCh38
rs544515477 Chr.17: 82024369-82024369 on Build GRCh38
rs760714350 Chr.17: 82024322-82024322 on Build GRCh38
rs756084582 Chr.17: 82024229-82024229 on Build GRCh38
rs760683111 Chr.17: 82024389-82024389 on Build GRCh38
rs748514619 Chr.17: 82024253-82024253 on Build GRCh38
rs755233152 Chr.17: 82024334-82024334 on Build GRCh38
rs752968092 Chr.17: 82024348-82024348 on Build GRCh38
rs773811663 Chr.17: 82024375-82024375 on Build GRCh38
rs767352645 Chr.17: 82024316-82024316 on Build GRCh38
rs150032877 Chr.17: 82024315-82024315 on Build GRCh38
rs766929141 Chr.17: 82024248-82024249 on Build GRCh38
rs775029461 Chr.17: 82024280-82024280 on Build GRCh38
rs774183081 Chr.17: 82024296-82024296 on Build GRCh38
rs762563468 Chr.17: 82024285-82024285 on Build GRCh38
rs766533025 Chr.17: 82024323-82024323 on Build GRCh38
rs550077176 Chr.17: 82024385-82024385 on Build GRCh38
rs771717530 Chr.17: 82024378-82024378 on Build GRCh38
rs754111221 Chr.17: 82024342-82024343 on Build GRCh38
rs768169408 Chr.17: 82024373-82024373 on Build GRCh38
rs772496011 Chr.17: 82024261-82024261 on Build GRCh38
rs963884999 Chr.17: 82024303-82024303 on Build GRCh38
rs778220280 Chr.17: 82024353-82024353 on Build GRCh38
rs918524503 Chr.17: 82024388-82024388 on Build GRCh38
rs1035413900 Chr.17: 82024241-82024241 on Build GRCh38
rs747583117 Chr.17: 82024263-82024263 on Build GRCh38
rs779413909 Chr.17: 82024250-82024250 on Build GRCh38
rs1026997205 Chr.17: 82024414-82024414 on Build GRCh38
rs773525050 Chr.17: 82024262-82024262 on Build GRCh38
rs190168749 Chr.17: 82024366-82024366 on Build GRCh38
rs780315700 Chr.17: 82024233-82024233 on Build GRCh38
rs73999342 Chr.17: 82024221-82024221 on Build GRCh38
rs942787717 Chr.17: 82024416-82024416 on Build GRCh38
rs972308295 Chr.17: 82024410-82024410 on Build GRCh38
rs778941313 Chr.17: 82024346-82024346 on Build GRCh38
rs539486708 Chr.17: 82024264-82024264 on Build GRCh38
rs558139324 Chr.17: 82024273-82024273 on Build GRCh38
rs750290073 Chr.17: 82024319-82024319 on Build GRCh38
rs755288221 Chr.17: 82024247-82024247 on Build GRCh38
rs748855212 Chr.17: 82024371-82024371 on Build GRCh38
rs754036133 Chr.17: 82024330-82024330 on Build GRCh38
rs536894217 Chr.17: 82024223-82024223 on Build GRCh38
rs931136527 Chr.17: 82024396-82024396 on Build GRCh38
rs959149685 Chr.17: 82024284-82024284 on Build GRCh38
rs761561906 Chr.17: 82024376-82024376 on Build GRCh38
rs749476456 Chr.17: 82024246-82024246 on Build GRCh38
rs562407195 Chr.17: 82024415-82024415 on Build GRCh38

View Full Product Details