Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52406030-52406279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT82
Entrez Gene ID
3888
Gene Name
keratin 82
Gene Aliases
HB2, Hb-2, KRTHB2
UniGene
Hs.134640
Gene Chromosome Location
Chr.12: 52393951-52406392 on Build GRCh38
Transcripts
NM_033033.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4747254 c.196delC p.R66fs*2 Chr.12: 52406082-52406082 on Build GRCh38
1162238 c.150C>A p.G50G Chr.12: 52406128-52406128 on Build GRCh38
4696911 c.115C>T p.R39W Chr.12: 52406163-52406163 on Build GRCh38
6137394 c.115C>A p.R39R Chr.12: 52406163-52406163 on Build GRCh38
431339 c.46_47delAG p.S16fs*28 Chr.12: 52406231-52406232 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752966130 Chr.12: 52406164-52406164 on Build GRCh38
rs751787099 Chr.12: 52406242-52406242 on Build GRCh38
rs766999588 Chr.12: 52406203-52406203 on Build GRCh38
rs748983586 Chr.12: 52406193-52406193 on Build GRCh38
rs756609221 Chr.12: 52406196-52406196 on Build GRCh38
rs747024167 Chr.12: 52406170-52406170 on Build GRCh38
rs200230668 Chr.12: 52406239-52406240 on Build GRCh38
rs757076531 Chr.12: 52406090-52406090 on Build GRCh38
rs764267511 Chr.12: 52406240-52406240 on Build GRCh38
rs770350865 Chr.12: 52406188-52406188 on Build GRCh38
rs201790528 Chr.12: 52406248-52406248 on Build GRCh38
rs146046365 Chr.12: 52406065-52406065 on Build GRCh38
rs762510631 Chr.12: 52406153-52406153 on Build GRCh38
rs777644287 Chr.12: 52406136-52406136 on Build GRCh38
rs920844901 Chr.12: 52406097-52406097 on Build GRCh38
rs143466157 Chr.12: 52406081-52406081 on Build GRCh38
rs375992302 Chr.12: 52406158-52406158 on Build GRCh38
rs142583496 Chr.12: 52406169-52406169 on Build GRCh38
rs746383199 Chr.12: 52406249-52406249 on Build GRCh38
rs1025241549 Chr.12: 52406047-52406047 on Build GRCh38
rs148287450 Chr.12: 52406100-52406100 on Build GRCh38
rs750337429 Chr.12: 52406160-52406160 on Build GRCh38
rs758682180 Chr.12: 52406237-52406237 on Build GRCh38
rs770264899 Chr.12: 52406146-52406146 on Build GRCh38
rs199864780 Chr.12: 52406101-52406101 on Build GRCh38
rs1021010341 Chr.12: 52406204-52406204 on Build GRCh38
rs766171227 Chr.12: 52406208-52406208 on Build GRCh38
rs139979674 Chr.12: 52406068-52406068 on Build GRCh38
rs538312762 Chr.12: 52406211-52406211 on Build GRCh38
rs767822222 Chr.12: 52406244-52406245 on Build GRCh38
rs772804944 Chr.12: 52406214-52406215 on Build GRCh38
rs746366742 Chr.12: 52406181-52406181 on Build GRCh38
rs558524531 Chr.12: 52406215-52406215 on Build GRCh38
rs757473623 Chr.12: 52406243-52406243 on Build GRCh38
rs150959498 Chr.12: 52406179-52406179 on Build GRCh38
rs757385746 Chr.12: 52406175-52406175 on Build GRCh38
rs760536987 Chr.12: 52406207-52406207 on Build GRCh38
rs763501001 Chr.12: 52406082-52406082 on Build GRCh38
rs1054483167 Chr.12: 52406187-52406187 on Build GRCh38
rs764612955 Chr.12: 52406210-52406210 on Build GRCh38
rs756632417 Chr.12: 52406255-52406255 on Build GRCh38
rs762745172 Chr.12: 52406217-52406217 on Build GRCh38
rs773198894 Chr.12: 52406205-52406205 on Build GRCh38
rs369260868 Chr.12: 52406191-52406191 on Build GRCh38
rs760021179 Chr.12: 52406143-52406143 on Build GRCh38
rs867458757 Chr.12: 52406129-52406129 on Build GRCh38
rs950122678 Chr.12: 52406154-52406154 on Build GRCh38
rs185048433 Chr.12: 52406201-52406201 on Build GRCh38
rs775272060 Chr.12: 52406152-52406152 on Build GRCh38
rs555090994 Chr.12: 52406115-52406115 on Build GRCh38
rs148453926 Chr.12: 52406139-52406139 on Build GRCh38
rs377286730 Chr.12: 52406114-52406114 on Build GRCh38
rs202034583 Chr.12: 52406202-52406202 on Build GRCh38
rs571511455 Chr.12: 52406107-52406107 on Build GRCh38
rs372901802 Chr.12: 52406052-52406052 on Build GRCh38
rs1040508122 Chr.12: 52406213-52406213 on Build GRCh38
rs764947086 Chr.12: 52406075-52406075 on Build GRCh38
rs567549946 Chr.12: 52406162-52406162 on Build GRCh38
rs772259336 Chr.12: 52406128-52406128 on Build GRCh38
rs1017300622 Chr.12: 52406239-52406239 on Build GRCh38
rs142011803 Chr.12: 52406216-52406216 on Build GRCh38
rs752370269 Chr.12: 52406080-52406080 on Build GRCh38
rs536478336 Chr.12: 52406165-52406165 on Build GRCh38
rs751397641 Chr.12: 52406231-52406232 on Build GRCh38
rs201747652 Chr.12: 52406138-52406138 on Build GRCh38
rs755959670 Chr.12: 52406161-52406161 on Build GRCh38
rs761357095 Chr.12: 52406200-52406202 on Build GRCh38
rs766346946 Chr.12: 52406220-52406239 on Build GRCh38
rs763665856 Chr.12: 52406157-52406157 on Build GRCh38

View Full Product Details