Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169132650-169132919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1B1
Entrez Gene ID
481
Gene Name
ATPase Na+/K+ transporting subunit beta 1
Gene Aliases
ATP1B
UniGene
Hs.291196
Gene Chromosome Location
Chr.1: 169106709-169132722 on Build GRCh38
Transcripts
NM_001677.3 NM_013330.4 NM_197972.2 NR_104229.1 XM_005245106.4 XM_017001100.1

Gene Symbol
NME7
Entrez Gene ID
29922
Gene Name
NME/NM23 family member 7
Gene Aliases
CFAP67, MN23H7, NDK 7, NDK7, nm23-H7
UniGene
Hs.706952
Gene Chromosome Location
Chr.1: 169132385-169367963 on Build GRCh38
Transcripts
NM_001677.3 NM_013330.4 NM_197972.2 NR_104229.1 XM_005245106.4 XM_017001100.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6285263 c.1099-8C>T p.? Chr.1: 169132825-169132825 on Build GRCh38
424513 c.1116G>C p.K372N Chr.1: 169132800-169132800 on Build GRCh38
3984448 c.1108T>C p.F370L Chr.1: 169132808-169132808 on Build GRCh38
3477935 c.1131G>T p.*377Y Chr.1: 169132785-169132785 on Build GRCh38
1601200 c.1099-10T>C p.? Chr.1: 169132827-169132827 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992605441 Chr.1: 169132716-169132716 on Build GRCh38
rs201176827 Chr.1: 169132809-169132809 on Build GRCh38
rs774573167 Chr.1: 169132848-169132848 on Build GRCh38
rs781586402 Chr.1: 169132824-169132824 on Build GRCh38
rs780984028 Chr.1: 169132871-169132872 on Build GRCh38
rs545011906 Chr.1: 169132698-169132698 on Build GRCh38
rs887830641 Chr.1: 169132725-169132725 on Build GRCh38
rs764711260 Chr.1: 169132803-169132803 on Build GRCh38
rs763285952 Chr.1: 169132864-169132865 on Build GRCh38
rs376305007 Chr.1: 169132878-169132879 on Build GRCh38
rs369306366 Chr.1: 169132758-169132758 on Build GRCh38
rs777210651 Chr.1: 169132802-169132803 on Build GRCh38
rs913671342 Chr.1: 169132894-169132894 on Build GRCh38
rs748411424 Chr.1: 169132737-169132737 on Build GRCh38
rs984904801 Chr.1: 169132687-169132687 on Build GRCh38
rs756396458 Chr.1: 169132837-169132837 on Build GRCh38
rs756803824 Chr.1: 169132798-169132798 on Build GRCh38
rs375605707 Chr.1: 169132784-169132784 on Build GRCh38
rs896786898 Chr.1: 169132883-169132883 on Build GRCh38
rs770179528 Chr.1: 169132831-169132832 on Build GRCh38
rs548632897 Chr.1: 169132812-169132812 on Build GRCh38
rs188194894 Chr.1: 169132759-169132759 on Build GRCh38
rs77280637 Chr.1: 169132796-169132796 on Build GRCh38
rs773520270 Chr.1: 169132862-169132863 on Build GRCh38
rs992748190 Chr.1: 169132745-169132745 on Build GRCh38
rs767220438 Chr.1: 169132869-169132878 on Build GRCh38
rs781531358 Chr.1: 169132734-169132734 on Build GRCh38
rs768606556 Chr.1: 169132765-169132767 on Build GRCh38
rs771102742 Chr.1: 169132842-169132842 on Build GRCh38
rs531921415 Chr.1: 169132858-169132858 on Build GRCh38
rs748568508 Chr.1: 169132834-169132834 on Build GRCh38
rs116473357 Chr.1: 169132726-169132726 on Build GRCh38
rs936380360 Chr.1: 169132828-169132828 on Build GRCh38
rs755143347 Chr.1: 169132806-169132807 on Build GRCh38
rs763836757 Chr.1: 169132788-169132788 on Build GRCh38
rs1138486 Chr.1: 169132697-169132697 on Build GRCh38
rs758587496 Chr.1: 169132846-169132846 on Build GRCh38
rs3180378 Chr.1: 169132717-169132717 on Build GRCh38
rs770035732 Chr.1: 169132739-169132739 on Build GRCh38
rs1042292639 Chr.1: 169132732-169132732 on Build GRCh38
rs773260598 Chr.1: 169132741-169132741 on Build GRCh38
rs924941800 Chr.1: 169132825-169132825 on Build GRCh38
rs375795890 Chr.1: 169132749-169132749 on Build GRCh38
rs561514028 Chr.1: 169132856-169132856 on Build GRCh38
rs778062810 Chr.1: 169132839-169132839 on Build GRCh38
rs749416691 Chr.1: 169132840-169132840 on Build GRCh38
rs760334412 Chr.1: 169132787-169132787 on Build GRCh38
rs767490707 Chr.1: 169132778-169132778 on Build GRCh38

View Full Product Details