Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153785845-153786090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDH3G
Entrez Gene ID
3421
Gene Name
isocitrate dehydrogenase 3 (NAD(+)) gamma
Gene Aliases
H-IDHG
UniGene
Hs.410197
Gene Chromosome Location
Chr.X: 153785766-153794523 on Build GRCh38
Transcripts
NM_004135.3 NM_174869.2

SNP Detail

SNP ID Chromosome Location
rs782807120 Chr.X: 153786016-153786016 on Build GRCh38
rs782228638 Chr.X: 153785892-153785892 on Build GRCh38
rs782293393 Chr.X: 153785871-153785871 on Build GRCh38
rs782616614 Chr.X: 153786020-153786020 on Build GRCh38
rs781876095 Chr.X: 153785882-153785882 on Build GRCh38
rs782138925 Chr.X: 153785964-153785964 on Build GRCh38
rs782249155 Chr.X: 153786021-153786021 on Build GRCh38
rs782274797 Chr.X: 153785904-153785904 on Build GRCh38
rs782410958 Chr.X: 153785913-153785913 on Build GRCh38
rs781791676 Chr.X: 153785886-153785886 on Build GRCh38
rs782081730 Chr.X: 153786047-153786047 on Build GRCh38
rs782301533 Chr.X: 153786025-153786025 on Build GRCh38
rs782314676 Chr.X: 153786029-153786029 on Build GRCh38
rs782692558 Chr.X: 153785885-153785885 on Build GRCh38
rs900825278 Chr.X: 153785987-153785987 on Build GRCh38
rs201442932 Chr.X: 153786022-153786022 on Build GRCh38
rs189834145 Chr.X: 153785982-153785982 on Build GRCh38
rs782530864 Chr.X: 153786016-153786016 on Build GRCh38
rs782371743 Chr.X: 153785893-153785893 on Build GRCh38
rs782132057 Chr.X: 153785963-153785963 on Build GRCh38
rs369724451 Chr.X: 153785884-153785884 on Build GRCh38
rs782218762 Chr.X: 153785973-153785973 on Build GRCh38
rs782024035 Chr.X: 153785912-153785912 on Build GRCh38
rs781990808 Chr.X: 153785956-153785956 on Build GRCh38
rs782370208 Chr.X: 153785955-153785955 on Build GRCh38
rs1000061564 Chr.X: 153786012-153786012 on Build GRCh38
rs782025533 Chr.X: 153786024-153786024 on Build GRCh38
rs782465724 Chr.X: 153785888-153785888 on Build GRCh38
rs200391024 Chr.X: 153786017-153786017 on Build GRCh38
rs782458117 Chr.X: 153786019-153786019 on Build GRCh38
rs781920782 Chr.X: 153786002-153786002 on Build GRCh38
rs781912650 Chr.X: 153785920-153785920 on Build GRCh38
rs782707752 Chr.X: 153785994-153785994 on Build GRCh38
rs199721612 Chr.X: 153785889-153785889 on Build GRCh38
rs944977292 Chr.X: 153785914-153785914 on Build GRCh38
rs200958121 Chr.X: 153786036-153786036 on Build GRCh38
rs782198097 Chr.X: 153786015-153786016 on Build GRCh38
rs945801138 Chr.X: 153785988-153785988 on Build GRCh38
rs374210146 Chr.X: 153785953-153785953 on Build GRCh38
rs782140197 Chr.X: 153786039-153786040 on Build GRCh38
rs781872253 Chr.X: 153786013-153786013 on Build GRCh38
rs782773577 Chr.X: 153786003-153786003 on Build GRCh38
rs7890126 Chr.X: 153785901-153785901 on Build GRCh38
rs782048919 Chr.X: 153786040-153786041 on Build GRCh38
rs781821977 Chr.X: 153785995-153785995 on Build GRCh38

View Full Product Details