Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44907160-44907433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFAP
Entrez Gene ID
2670
Gene Name
glial fibrillary acidic protein
Gene Aliases
ALXDRD
UniGene
Hs.514227
Gene Chromosome Location
Chr.17: 44905626-44915552 on Build GRCh38
Transcripts
NM_002055.4 XM_017024451.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
137106 c.1288G>A p.D430N Chr.17: 44907358-44907358 on Build GRCh38
1610345 c.1258-8C>T p.? Chr.17: 44907396-44907396 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs531934195 Chr.17: 44907290-44907290 on Build GRCh38
rs373125489 Chr.17: 44907311-44907311 on Build GRCh38
rs758077793 Chr.17: 44907337-44907337 on Build GRCh38
rs745328182 Chr.17: 44907371-44907371 on Build GRCh38
rs1036170201 Chr.17: 44907219-44907219 on Build GRCh38
rs777812616 Chr.17: 44907340-44907340 on Build GRCh38
rs1007142515 Chr.17: 44907199-44907199 on Build GRCh38
rs34579312 Chr.17: 44907195-44907196 on Build GRCh38
rs925118526 Chr.17: 44907329-44907329 on Build GRCh38
rs576556981 Chr.17: 44907184-44907184 on Build GRCh38
rs770321525 Chr.17: 44907301-44907301 on Build GRCh38
rs932960024 Chr.17: 44907180-44907180 on Build GRCh38
rs769443611 Chr.17: 44907372-44907372 on Build GRCh38
rs370608748 Chr.17: 44907318-44907318 on Build GRCh38
rs995613611 Chr.17: 44907274-44907274 on Build GRCh38
rs559313582 Chr.17: 44907243-44907243 on Build GRCh38
rs35560537 Chr.17: 44907194-44907195 on Build GRCh38
rs201709616 Chr.17: 44907317-44907317 on Build GRCh38
rs191737430 Chr.17: 44907181-44907181 on Build GRCh38
rs1018495387 Chr.17: 44907235-44907235 on Build GRCh38
rs111618522 Chr.17: 44907391-44907391 on Build GRCh38
rs113487550 Chr.17: 44907331-44907331 on Build GRCh38
rs540652654 Chr.17: 44907206-44907206 on Build GRCh38
rs561429495 Chr.17: 44907287-44907287 on Build GRCh38
rs765145414 Chr.17: 44907306-44907306 on Build GRCh38
rs779643685 Chr.17: 44907386-44907386 on Build GRCh38
rs765487380 Chr.17: 44907323-44907323 on Build GRCh38
rs780671204 Chr.17: 44907358-44907358 on Build GRCh38
rs377576009 Chr.17: 44907189-44907190 on Build GRCh38
rs550446365 Chr.17: 44907310-44907310 on Build GRCh38
rs756854907 Chr.17: 44907345-44907345 on Build GRCh38
rs369389003 Chr.17: 44907302-44907302 on Build GRCh38
rs544481265 Chr.17: 44907320-44907320 on Build GRCh38
rs267607669 Chr.17: 44907315-44907315 on Build GRCh38
rs183151408 Chr.17: 44907208-44907208 on Build GRCh38
rs11558961 Chr.17: 44907319-44907319 on Build GRCh38
rs772303877 Chr.17: 44907403-44907403 on Build GRCh38
rs773810004 Chr.17: 44907406-44907406 on Build GRCh38
rs891415429 Chr.17: 44907185-44907185 on Build GRCh38
rs753652614 Chr.17: 44907278-44907278 on Build GRCh38
rs768103039 Chr.17: 44907313-44907313 on Build GRCh38
rs572620394 Chr.17: 44907250-44907250 on Build GRCh38
rs776738529 Chr.17: 44907344-44907344 on Build GRCh38
rs543312036 Chr.17: 44907284-44907284 on Build GRCh38
rs748570438 Chr.17: 44907387-44907387 on Build GRCh38
rs761082086 Chr.17: 44907407-44907407 on Build GRCh38
rs267607521 Chr.17: 44907369-44907369 on Build GRCh38

View Full Product Details