Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10284386-10284643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICAM1
Entrez Gene ID
3383
Gene Name
intercellular adhesion molecule 1
Gene Aliases
BB2, CD54, P3.58
UniGene
Hs.643447
Gene Chromosome Location
Chr.19: 10270841-10286615 on Build GRCh38
Transcripts
NM_000201.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
990480 c.1147C>A p.H383N Chr.19: 10284624-10284624 on Build GRCh38
1750615 c.1056G>C p.P352P Chr.19: 10284533-10284533 on Build GRCh38
5926034 c.974G>A p.G325E Chr.19: 10284451-10284451 on Build GRCh38
7220531 c.1095C>T p.N365N Chr.19: 10284572-10284572 on Build GRCh38
990479 c.1022C>T p.T341M Chr.19: 10284499-10284499 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758965302 Chr.19: 10284592-10284592 on Build GRCh38
rs139178890 Chr.19: 10284576-10284576 on Build GRCh38
rs762576033 Chr.19: 10284487-10284487 on Build GRCh38
rs779307629 Chr.19: 10284510-10284510 on Build GRCh38
rs746741810 Chr.19: 10284457-10284457 on Build GRCh38
rs747223390 Chr.19: 10284514-10284514 on Build GRCh38
rs757771096 Chr.19: 10284446-10284446 on Build GRCh38
rs775039982 Chr.19: 10284516-10284536 on Build GRCh38
rs770551007 Chr.19: 10284561-10284561 on Build GRCh38
rs754190930 Chr.19: 10284506-10284506 on Build GRCh38
rs769659570 Chr.19: 10284472-10284472 on Build GRCh38
rs141326678 Chr.19: 10284499-10284499 on Build GRCh38
rs766445571 Chr.19: 10284606-10284606 on Build GRCh38
rs868375367 Chr.19: 10284542-10284542 on Build GRCh38
rs376622574 Chr.19: 10284612-10284612 on Build GRCh38
rs150014298 Chr.19: 10284608-10284608 on Build GRCh38
rs1008944257 Chr.19: 10284454-10284454 on Build GRCh38
rs755868188 Chr.19: 10284615-10284615 on Build GRCh38
rs758480023 Chr.19: 10284512-10284512 on Build GRCh38
rs757757223 Chr.19: 10284509-10284509 on Build GRCh38
rs139290643 Chr.19: 10284584-10284584 on Build GRCh38
rs777679809 Chr.19: 10284625-10284625 on Build GRCh38
rs768543917 Chr.19: 10284412-10284412 on Build GRCh38
rs201383296 Chr.19: 10284533-10284533 on Build GRCh38
rs759319472 Chr.19: 10284500-10284500 on Build GRCh38
rs983667097 Chr.19: 10284526-10284526 on Build GRCh38
rs5495 Chr.19: 10284420-10284420 on Build GRCh38
rs780195275 Chr.19: 10284513-10284513 on Build GRCh38
rs137970890 Chr.19: 10284419-10284419 on Build GRCh38
rs143008699 Chr.19: 10284456-10284456 on Build GRCh38
rs146000551 Chr.19: 10284577-10284577 on Build GRCh38
rs368283911 Chr.19: 10284449-10284449 on Build GRCh38
rs768455781 Chr.19: 10284464-10284464 on Build GRCh38
rs199672020 Chr.19: 10284564-10284564 on Build GRCh38
rs764794802 Chr.19: 10284414-10284414 on Build GRCh38
rs754356021 Chr.19: 10284441-10284441 on Build GRCh38
rs756550665 Chr.19: 10284427-10284427 on Build GRCh38
rs755206701 Chr.19: 10284610-10284610 on Build GRCh38
rs781009982 Chr.19: 10284518-10284518 on Build GRCh38
rs765707240 Chr.19: 10284578-10284578 on Build GRCh38
rs960494429 Chr.19: 10284489-10284489 on Build GRCh38
rs772229913 Chr.19: 10284452-10284452 on Build GRCh38
rs1036180125 Chr.19: 10284622-10284622 on Build GRCh38
rs199692247 Chr.19: 10284413-10284413 on Build GRCh38
rs771319016 Chr.19: 10284515-10284535 on Build GRCh38
rs768938327 Chr.19: 10284516-10284516 on Build GRCh38
rs374392444 Chr.19: 10284431-10284431 on Build GRCh38
rs146134321 Chr.19: 10284465-10284465 on Build GRCh38
rs1027972658 Chr.19: 10284517-10284517 on Build GRCh38
rs762664096 Chr.19: 10284418-10284418 on Build GRCh38
rs146954887 Chr.19: 10284410-10284410 on Build GRCh38
rs964678962 Chr.19: 10284469-10284469 on Build GRCh38
rs370415118 Chr.19: 10284573-10284573 on Build GRCh38
rs772757657 Chr.19: 10284483-10284483 on Build GRCh38
rs13306429 Chr.19: 10284572-10284572 on Build GRCh38
rs1801714 Chr.19: 10284532-10284532 on Build GRCh38
rs772315973 Chr.19: 10284409-10284409 on Build GRCh38
rs377420632 Chr.19: 10284557-10284557 on Build GRCh38

View Full Product Details