Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113218752-113219025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL4A
Entrez Gene ID
8451
Gene Name
cullin 4A
Gene Aliases
-
UniGene
Hs.339735
Gene Chromosome Location
Chr.13: 113208193-113265078 on Build GRCh38
Transcripts
NM_001008895.2 NM_001278513.1 NM_001278514.1 NM_003589.2 XM_011537523.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5624264 c.39C>A p.H13Q Chr.13: 113219019-113219019 on Build GRCh38
5624263 c.339C>A p.H113Q Chr.13: 113219019-113219019 on Build GRCh38
6638069 c.300A>G p.P100P Chr.13: 113218980-113218980 on Build GRCh38
2264189 c.320G>A p.R107H Chr.13: 113219000-113219000 on Build GRCh38
2264186 c.281G>A p.C94Y Chr.13: 113218961-113218961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745437164 Chr.13: 113218905-113218905 on Build GRCh38
rs552346415 Chr.13: 113218869-113218869 on Build GRCh38
rs866118183 Chr.13: 113218969-113218969 on Build GRCh38
rs746688646 Chr.13: 113218925-113218925 on Build GRCh38
rs879145826 Chr.13: 113218848-113218848 on Build GRCh38
rs528329102 Chr.13: 113218859-113218859 on Build GRCh38
rs377053734 Chr.13: 113218892-113218900 on Build GRCh38
rs936864901 Chr.13: 113218992-113218992 on Build GRCh38
rs977312573 Chr.13: 113218844-113218844 on Build GRCh38
rs769673712 Chr.13: 113218973-113218973 on Build GRCh38
rs754897399 Chr.13: 113218828-113218828 on Build GRCh38
rs899842215 Chr.13: 113218891-113218891 on Build GRCh38
rs372753362 Chr.13: 113218937-113218937 on Build GRCh38
rs1028827446 Chr.13: 113218838-113218838 on Build GRCh38
rs781705519 Chr.13: 113218898-113218898 on Build GRCh38
rs920277692 Chr.13: 113218804-113218804 on Build GRCh38
rs756498004 Chr.13: 113219005-113219005 on Build GRCh38
rs991371583 Chr.13: 113218824-113218824 on Build GRCh38
rs996046452 Chr.13: 113218837-113218837 on Build GRCh38
rs201258343 Chr.13: 113218888-113218889 on Build GRCh38
rs771581101 Chr.13: 113218906-113218906 on Build GRCh38
rs766541599 Chr.13: 113218986-113218986 on Build GRCh38
rs978202220 Chr.13: 113218971-113218971 on Build GRCh38
rs537956282 Chr.13: 113218900-113218900 on Build GRCh38
rs386775045 Chr.13: 113218890-113218900 on Build GRCh38
rs566756725 Chr.13: 113218846-113218846 on Build GRCh38
rs912583870 Chr.13: 113218874-113218874 on Build GRCh38
rs751631994 Chr.13: 113218896-113218896 on Build GRCh38
rs765121980 Chr.13: 113218864-113218864 on Build GRCh38
rs752550286 Chr.13: 113218883-113218884 on Build GRCh38
rs958574596 Chr.13: 113218817-113218817 on Build GRCh38
rs527534380 Chr.13: 113218860-113218860 on Build GRCh38
rs967180353 Chr.13: 113218890-113218890 on Build GRCh38
rs369798527 Chr.13: 113218899-113218899 on Build GRCh38
rs932674631 Chr.13: 113218914-113218914 on Build GRCh38
rs200825752 Chr.13: 113218999-113218999 on Build GRCh38
rs761615029 Chr.13: 113218952-113218952 on Build GRCh38
rs201003509 Chr.13: 113219000-113219000 on Build GRCh38
rs773192332 Chr.13: 113218977-113218977 on Build GRCh38
rs768261236 Chr.13: 113218933-113218933 on Build GRCh38
rs779732455 Chr.13: 113218912-113218912 on Build GRCh38
rs199860261 Chr.13: 113218981-113218981 on Build GRCh38

View Full Product Details