Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70370014-70370275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX19A
Entrez Gene ID
55308
Gene Name
DEAD-box helicase 19A
Gene Aliases
DDX19-DDX19L, DDX19L
UniGene
Hs.656037
Gene Chromosome Location
Chr.16: 70346829-70373383 on Build GRCh38
Transcripts
NM_001320522.1 NM_001320525.1 NM_001320526.1 NM_001320527.1 NM_018332.4 NR_135296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5696966 c.1048G>T p.A350S Chr.16: 70370250-70370250 on Build GRCh38
3511365 c.1072C>T p.Q358* Chr.16: 70370274-70370274 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746734483 Chr.16: 70370178-70370178 on Build GRCh38
rs543499435 Chr.16: 70370136-70370136 on Build GRCh38
rs752248645 Chr.16: 70370253-70370253 on Build GRCh38
rs760980950 Chr.16: 70370201-70370201 on Build GRCh38
rs777635223 Chr.16: 70370176-70370176 on Build GRCh38
rs749800407 Chr.16: 70370058-70370058 on Build GRCh38
rs769657427 Chr.16: 70370187-70370187 on Build GRCh38
rs903429848 Chr.16: 70370045-70370045 on Build GRCh38
rs1031357995 Chr.16: 70370190-70370190 on Build GRCh38
rs910386576 Chr.16: 70370239-70370239 on Build GRCh38
rs531712900 Chr.16: 70370061-70370061 on Build GRCh38
rs377735304 Chr.16: 70370210-70370210 on Build GRCh38
rs1013006067 Chr.16: 70370117-70370117 on Build GRCh38
rs774464255 Chr.16: 70370191-70370191 on Build GRCh38
rs551502746 Chr.16: 70370119-70370119 on Build GRCh38
rs776901149 Chr.16: 70370233-70370233 on Build GRCh38
rs758110086 Chr.16: 70370173-70370173 on Build GRCh38
rs995884178 Chr.16: 70370142-70370142 on Build GRCh38
rs749080479 Chr.16: 70370208-70370208 on Build GRCh38
rs145142990 Chr.16: 70370151-70370151 on Build GRCh38
rs745846027 Chr.16: 70370186-70370186 on Build GRCh38
rs987758668 Chr.16: 70370204-70370204 on Build GRCh38
rs765722278 Chr.16: 70370247-70370247 on Build GRCh38
rs766764627 Chr.16: 70370207-70370207 on Build GRCh38
rs149127218 Chr.16: 70370179-70370179 on Build GRCh38
rs567958200 Chr.16: 70370238-70370238 on Build GRCh38
rs145537687 Chr.16: 70370146-70370146 on Build GRCh38
rs773440467 Chr.16: 70370196-70370196 on Build GRCh38
rs566046959 Chr.16: 70370057-70370057 on Build GRCh38
rs896470839 Chr.16: 70370197-70370197 on Build GRCh38
rs748230424 Chr.16: 70370192-70370192 on Build GRCh38
rs1040092867 Chr.16: 70370130-70370130 on Build GRCh38
rs894602506 Chr.16: 70370089-70370089 on Build GRCh38
rs777484981 Chr.16: 70370207-70370207 on Build GRCh38
rs796215650 Chr.16: 70370052-70370052 on Build GRCh38
rs780941777 Chr.16: 70370180-70370180 on Build GRCh38

View Full Product Details