Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 23437029-23437290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD4
Entrez Gene ID
9583
Gene Name
ectonucleoside triphosphate diphosphohydrolase 4
Gene Aliases
LALP70, LAP70, LYSAL1, NTPDase-4, UDPase
UniGene
Hs.444389
Gene Chromosome Location
Chr.8: 23429152-23457731 on Build GRCh38
Transcripts
NM_001128930.2 NM_004901.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3899032 c.1187G>A p.R396Q Chr.8: 23437121-23437121 on Build GRCh38
4419539 c.1060A>G p.K354E Chr.8: 23437248-23437248 on Build GRCh38
5991440 c.1173C>G p.D391E Chr.8: 23437135-23437135 on Build GRCh38
6013095 c.1202C>T p.P401L Chr.8: 23437106-23437106 on Build GRCh38
1236569 c.1088C>T p.P363L Chr.8: 23437220-23437220 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755215378 Chr.8: 23437121-23437121 on Build GRCh38
rs149781931 Chr.8: 23437085-23437085 on Build GRCh38
rs774046300 Chr.8: 23437254-23437254 on Build GRCh38
rs750946718 Chr.8: 23437241-23437242 on Build GRCh38
rs747617396 Chr.8: 23437162-23437162 on Build GRCh38
rs761851175 Chr.8: 23437103-23437103 on Build GRCh38
rs749197277 Chr.8: 23437252-23437252 on Build GRCh38
rs751787221 Chr.8: 23437054-23437054 on Build GRCh38
rs752707254 Chr.8: 23437126-23437126 on Build GRCh38
rs774139817 Chr.8: 23437202-23437202 on Build GRCh38
rs1005445834 Chr.8: 23437203-23437203 on Build GRCh38
rs757878495 Chr.8: 23437072-23437072 on Build GRCh38
rs779156362 Chr.8: 23437223-23437223 on Build GRCh38
rs776281230 Chr.8: 23437158-23437158 on Build GRCh38
rs762708504 Chr.8: 23437116-23437116 on Build GRCh38
rs200990091 Chr.8: 23437091-23437091 on Build GRCh38
rs751797992 Chr.8: 23437119-23437119 on Build GRCh38
rs767686644 Chr.8: 23437207-23437207 on Build GRCh38
rs889661283 Chr.8: 23437231-23437231 on Build GRCh38
rs754704722 Chr.8: 23437238-23437238 on Build GRCh38
rs142918666 Chr.8: 23437058-23437058 on Build GRCh38
rs766147341 Chr.8: 23437199-23437199 on Build GRCh38
rs1012994702 Chr.8: 23437149-23437149 on Build GRCh38
rs985094298 Chr.8: 23437140-23437140 on Build GRCh38
rs371867310 Chr.8: 23437217-23437217 on Build GRCh38
rs150653551 Chr.8: 23437059-23437059 on Build GRCh38
rs947609879 Chr.8: 23437092-23437092 on Build GRCh38
rs200546697 Chr.8: 23437122-23437122 on Build GRCh38
rs759682393 Chr.8: 23437204-23437204 on Build GRCh38
rs146688903 Chr.8: 23437148-23437148 on Build GRCh38
rs752796827 Chr.8: 23437211-23437211 on Build GRCh38
rs759326188 Chr.8: 23437260-23437260 on Build GRCh38
rs777325275 Chr.8: 23437251-23437251 on Build GRCh38
rs371638792 Chr.8: 23437153-23437153 on Build GRCh38
rs202109130 Chr.8: 23437086-23437086 on Build GRCh38
rs746173135 Chr.8: 23437229-23437229 on Build GRCh38
rs766169270 Chr.8: 23437118-23437118 on Build GRCh38
rs35942025 Chr.8: 23437219-23437219 on Build GRCh38
rs756644544 Chr.8: 23437129-23437129 on Build GRCh38
rs147868072 Chr.8: 23437111-23437111 on Build GRCh38
rs773166801 Chr.8: 23437113-23437113 on Build GRCh38
rs139756981 Chr.8: 23437066-23437066 on Build GRCh38
rs142298900 Chr.8: 23437132-23437132 on Build GRCh38
rs773326250 Chr.8: 23437184-23437184 on Build GRCh38
rs771777272 Chr.8: 23437074-23437074 on Build GRCh38
rs781045414 Chr.8: 23437240-23437240 on Build GRCh38
rs140786682 Chr.8: 23437078-23437078 on Build GRCh38
rs762945610 Chr.8: 23437197-23437197 on Build GRCh38
rs372875344 Chr.8: 23437220-23437220 on Build GRCh38
rs367917989 Chr.8: 23437067-23437067 on Build GRCh38
rs368240094 Chr.8: 23437189-23437189 on Build GRCh38
rs1015440521 Chr.8: 23437209-23437209 on Build GRCh38
rs755054888 Chr.8: 23437056-23437056 on Build GRCh38
rs34073541 Chr.8: 23437087-23437087 on Build GRCh38
rs769410229 Chr.8: 23437177-23437177 on Build GRCh38
rs770836670 Chr.8: 23437253-23437253 on Build GRCh38
rs144647572 Chr.8: 23437139-23437139 on Build GRCh38
rs778203028 Chr.8: 23437130-23437130 on Build GRCh38
rs61731462 Chr.8: 23437247-23437247 on Build GRCh38
rs927488658 Chr.8: 23437128-23437128 on Build GRCh38
rs2272641 Chr.8: 23437248-23437248 on Build GRCh38
rs554223103 Chr.8: 23437063-23437063 on Build GRCh38
rs746194550 Chr.8: 23437082-23437082 on Build GRCh38
rs367737939 Chr.8: 23437105-23437105 on Build GRCh38
rs865800658 Chr.8: 23437221-23437221 on Build GRCh38
rs73551669 Chr.8: 23437143-23437143 on Build GRCh38

View Full Product Details