Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47716572-47716845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHD2
Entrez Gene ID
30846
Gene Name
EH domain containing 2
Gene Aliases
PAST2
UniGene
Hs.744963
Gene Chromosome Location
Chr.19: 47713344-47743134 on Build GRCh38
Transcripts
NM_014601.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5471070 c.105G>A p.P35P Chr.19: 47716717-47716717 on Build GRCh38
3536371 c.156C>G p.D52E Chr.19: 47716768-47716768 on Build GRCh38
5519689 c.31C>T p.R11W Chr.19: 47716643-47716643 on Build GRCh38
4186250 c.103C>T p.P35S Chr.19: 47716715-47716715 on Build GRCh38
6652422 c.144G>T p.P48P Chr.19: 47716756-47716756 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763612750 Chr.19: 47716752-47716752 on Build GRCh38
rs201716618 Chr.19: 47716701-47716701 on Build GRCh38
rs11550017 Chr.19: 47716636-47716636 on Build GRCh38
rs757569293 Chr.19: 47716717-47716717 on Build GRCh38
rs753444147 Chr.19: 47716643-47716643 on Build GRCh38
rs865806381 Chr.19: 47716806-47716806 on Build GRCh38
rs374903022 Chr.19: 47716593-47716593 on Build GRCh38
rs200498127 Chr.19: 47716750-47716750 on Build GRCh38
rs558812716 Chr.19: 47716740-47716740 on Build GRCh38
rs767110542 Chr.19: 47716630-47716630 on Build GRCh38
rs939377555 Chr.19: 47716728-47716728 on Build GRCh38
rs753491573 Chr.19: 47716692-47716692 on Build GRCh38
rs372776185 Chr.19: 47716778-47716778 on Build GRCh38
rs978169042 Chr.19: 47716741-47716741 on Build GRCh38
rs752294943 Chr.19: 47716635-47716635 on Build GRCh38
rs779218399 Chr.19: 47716813-47716813 on Build GRCh38
rs376620177 Chr.19: 47716716-47716716 on Build GRCh38
rs774772728 Chr.19: 47716733-47716733 on Build GRCh38
rs750209017 Chr.19: 47716777-47716777 on Build GRCh38
rs150872006 Chr.19: 47716667-47716667 on Build GRCh38
rs61746407 Chr.19: 47716672-47716672 on Build GRCh38
rs760139473 Chr.19: 47716623-47716623 on Build GRCh38
rs772524049 Chr.19: 47716819-47716819 on Build GRCh38
rs771094107 Chr.19: 47716727-47716727 on Build GRCh38
rs377445271 Chr.19: 47716753-47716753 on Build GRCh38
rs748297307 Chr.19: 47716658-47716658 on Build GRCh38
rs756884374 Chr.19: 47716696-47716696 on Build GRCh38
rs933165812 Chr.19: 47716799-47716799 on Build GRCh38
rs369448629 Chr.19: 47716734-47716734 on Build GRCh38
rs34140460 Chr.19: 47716781-47716781 on Build GRCh38
rs764869867 Chr.19: 47716768-47716768 on Build GRCh38
rs760084581 Chr.19: 47716671-47716671 on Build GRCh38
rs757488835 Chr.19: 47716805-47716805 on Build GRCh38
rs781221558 Chr.19: 47716657-47716657 on Build GRCh38
rs188300386 Chr.19: 47716637-47716637 on Build GRCh38
rs539106529 Chr.19: 47716719-47716719 on Build GRCh38
rs777875700 Chr.19: 47716800-47716800 on Build GRCh38
rs146201317 Chr.19: 47716632-47716632 on Build GRCh38
rs377376274 Chr.19: 47716785-47716785 on Build GRCh38
rs765059260 Chr.19: 47716700-47716700 on Build GRCh38
rs371592171 Chr.19: 47716644-47716644 on Build GRCh38
rs763515230 Chr.19: 47716689-47716689 on Build GRCh38
rs369298767 Chr.19: 47716612-47716612 on Build GRCh38
rs199754199 Chr.19: 47716669-47716669 on Build GRCh38
rs754918575 Chr.19: 47716795-47716795 on Build GRCh38
rs886341237 Chr.19: 47716591-47716591 on Build GRCh38
rs147756934 Chr.19: 47716756-47716756 on Build GRCh38
rs752062746 Chr.19: 47716614-47716614 on Build GRCh38
rs11550016 Chr.19: 47716755-47716755 on Build GRCh38
rs760203223 Chr.19: 47716681-47716681 on Build GRCh38
rs746091692 Chr.19: 47716818-47716818 on Build GRCh38
rs899245616 Chr.19: 47716604-47716604 on Build GRCh38
rs563303087 Chr.19: 47716598-47716598 on Build GRCh38
rs751447069 Chr.19: 47716793-47716793 on Build GRCh38
rs758807685 Chr.19: 47716704-47716704 on Build GRCh38
rs776021238 Chr.19: 47716822-47716822 on Build GRCh38
rs371409023 Chr.19: 47716668-47716668 on Build GRCh38
rs772577292 Chr.19: 47716735-47716735 on Build GRCh38
rs563066551 Chr.19: 47716648-47716648 on Build GRCh38
rs764409449 Chr.19: 47716599-47716599 on Build GRCh38
rs763435241 Chr.19: 47716605-47716605 on Build GRCh38
rs144624643 Chr.19: 47716804-47716804 on Build GRCh38
rs773429484 Chr.19: 47716660-47716660 on Build GRCh38
rs142388414 Chr.19: 47716655-47716655 on Build GRCh38

View Full Product Details