Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48625561-48625828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXB9
Entrez Gene ID
3219
Gene Name
homeobox B9
Gene Aliases
HOX-2.5, HOX2, HOX2E
UniGene
Hs.463350
Gene Chromosome Location
Chr.17: 48621156-48626473 on Build GRCh38
Transcripts
NM_024017.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5682789 c.491G>C p.S164T Chr.17: 48625779-48625779 on Build GRCh38
6328221 c.507G>A p.R169R Chr.17: 48625763-48625763 on Build GRCh38
6254039 c.509C>A p.P170Q Chr.17: 48625761-48625761 on Build GRCh38
6681720 c.489A>T p.G163G Chr.17: 48625781-48625781 on Build GRCh38
4690986 c.455G>T p.R152I Chr.17: 48625815-48625815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199882905 Chr.17: 48625804-48625804 on Build GRCh38
rs373579346 Chr.17: 48625746-48625746 on Build GRCh38
rs540664499 Chr.17: 48625619-48625619 on Build GRCh38
rs934473479 Chr.17: 48625593-48625593 on Build GRCh38
rs1051389016 Chr.17: 48625588-48625588 on Build GRCh38
rs371142398 Chr.17: 48625708-48625708 on Build GRCh38
rs200925253 Chr.17: 48625780-48625780 on Build GRCh38
rs552239806 Chr.17: 48625625-48625625 on Build GRCh38
rs768068469 Chr.17: 48625724-48625724 on Build GRCh38
rs924731822 Chr.17: 48625587-48625587 on Build GRCh38
rs112409771 Chr.17: 48625637-48625637 on Build GRCh38
rs528673550 Chr.17: 48625638-48625638 on Build GRCh38
rs746897612 Chr.17: 48625777-48625777 on Build GRCh38
rs199501425 Chr.17: 48625623-48625623 on Build GRCh38
rs762490822 Chr.17: 48625723-48625723 on Build GRCh38
rs143822435 Chr.17: 48625651-48625651 on Build GRCh38
rs376332007 Chr.17: 48625750-48625750 on Build GRCh38
rs548773900 Chr.17: 48625641-48625641 on Build GRCh38
rs914359802 Chr.17: 48625756-48625756 on Build GRCh38
rs1050067617 Chr.17: 48625793-48625793 on Build GRCh38
rs192619342 Chr.17: 48625707-48625707 on Build GRCh38
rs201721789 Chr.17: 48625733-48625733 on Build GRCh38
rs952260532 Chr.17: 48625694-48625694 on Build GRCh38
rs770582746 Chr.17: 48625712-48625712 on Build GRCh38
rs769492183 Chr.17: 48625719-48625719 on Build GRCh38
rs186639828 Chr.17: 48625658-48625658 on Build GRCh38
rs570824497 Chr.17: 48625703-48625703 on Build GRCh38
rs375169682 Chr.17: 48625715-48625715 on Build GRCh38
rs1037658834 Chr.17: 48625603-48625603 on Build GRCh38
rs758406359 Chr.17: 48625762-48625762 on Build GRCh38
rs144939547 Chr.17: 48625768-48625768 on Build GRCh38
rs775211289 Chr.17: 48625805-48625805 on Build GRCh38
rs1027800446 Chr.17: 48625674-48625674 on Build GRCh38
rs1007896574 Chr.17: 48625642-48625642 on Build GRCh38
rs1054319490 Chr.17: 48625594-48625594 on Build GRCh38
rs961143104 Chr.17: 48625652-48625652 on Build GRCh38
rs893041036 Chr.17: 48625600-48625600 on Build GRCh38
rs534521996 Chr.17: 48625720-48625720 on Build GRCh38
rs112687528 Chr.17: 48625752-48625752 on Build GRCh38
rs200252541 Chr.17: 48625728-48625728 on Build GRCh38
rs148177888 Chr.17: 48625711-48625711 on Build GRCh38
rs537701030 Chr.17: 48625654-48625654 on Build GRCh38
rs543888333 Chr.17: 48625698-48625698 on Build GRCh38
rs933049056 Chr.17: 48625743-48625743 on Build GRCh38
rs745570627 Chr.17: 48625801-48625801 on Build GRCh38
rs369814321 Chr.17: 48625742-48625742 on Build GRCh38
rs144092219 Chr.17: 48625782-48625782 on Build GRCh38
rs199659441 Chr.17: 48625725-48625725 on Build GRCh38

View Full Product Details