Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 160427678-160427913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTTG1
Entrez Gene ID
9232
Gene Name
pituitary tumor-transforming 1
Gene Aliases
EAP1, HPTTG, PTTG, TUTR1
UniGene
Hs.350966
Gene Chromosome Location
Chr.5: 160421807-160428744 on Build GRCh38
Transcripts
NM_001282382.1 NM_001282383.1 NM_004219.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3853348 c.508C>T p.P170S Chr.5: 160427852-160427852 on Build GRCh38
1065217 c.385G>A p.D129N Chr.5: 160427729-160427729 on Build GRCh38
1065216 c.380G>T p.S127I Chr.5: 160427724-160427724 on Build GRCh38
3827596 c.516A>C p.P172P Chr.5: 160427860-160427860 on Build GRCh38
7386429 c.402C>G p.H134Q Chr.5: 160427746-160427746 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201530656 Chr.5: 160427845-160427845 on Build GRCh38
rs890511710 Chr.5: 160427763-160427763 on Build GRCh38
rs752402652 Chr.5: 160427889-160427889 on Build GRCh38
rs764721500 Chr.5: 160427887-160427888 on Build GRCh38
rs1054748669 Chr.5: 160427768-160427768 on Build GRCh38
rs201000365 Chr.5: 160427808-160427808 on Build GRCh38
rs751144118 Chr.5: 160427830-160427830 on Build GRCh38
rs760841598 Chr.5: 160427786-160427786 on Build GRCh38
rs769793739 Chr.5: 160427756-160427756 on Build GRCh38
rs749845133 Chr.5: 160427733-160427733 on Build GRCh38
rs370916098 Chr.5: 160427833-160427833 on Build GRCh38
rs1009503207 Chr.5: 160427703-160427703 on Build GRCh38
rs777820619 Chr.5: 160427767-160427767 on Build GRCh38
rs370650306 Chr.5: 160427755-160427755 on Build GRCh38
rs952386445 Chr.5: 160427862-160427862 on Build GRCh38
rs756000817 Chr.5: 160427849-160427849 on Build GRCh38
rs757986269 Chr.5: 160427734-160427734 on Build GRCh38
rs1057851 Chr.5: 160427848-160427848 on Build GRCh38
rs375357049 Chr.5: 160427702-160427702 on Build GRCh38
rs377676384 Chr.5: 160427818-160427818 on Build GRCh38
rs745929107 Chr.5: 160427773-160427773 on Build GRCh38
rs751007105 Chr.5: 160427812-160427812 on Build GRCh38
rs780145186 Chr.5: 160427885-160427885 on Build GRCh38
rs1027890997 Chr.5: 160427856-160427856 on Build GRCh38
rs779569934 Chr.5: 160427737-160427737 on Build GRCh38
rs777537997 Chr.5: 160427852-160427852 on Build GRCh38
rs561528753 Chr.5: 160427704-160427704 on Build GRCh38
rs772167069 Chr.5: 160427697-160427697 on Build GRCh38
rs754698420 Chr.5: 160427752-160427752 on Build GRCh38
rs765863164 Chr.5: 160427822-160427822 on Build GRCh38
rs765650582 Chr.5: 160427809-160427809 on Build GRCh38
rs377175957 Chr.5: 160427713-160427713 on Build GRCh38
rs751273574 Chr.5: 160427750-160427750 on Build GRCh38
rs775846230 Chr.5: 160427699-160427699 on Build GRCh38
rs1042131472 Chr.5: 160427830-160427830 on Build GRCh38
rs764494898 Chr.5: 160427708-160427708 on Build GRCh38
rs754610499 Chr.5: 160427831-160427831 on Build GRCh38
rs896135688 Chr.5: 160427815-160427815 on Build GRCh38
rs749294575 Chr.5: 160427859-160427859 on Build GRCh38
rs772230778 Chr.5: 160427776-160427776 on Build GRCh38
rs757166662 Chr.5: 160427869-160427869 on Build GRCh38
rs781094535 Chr.5: 160427754-160427754 on Build GRCh38
rs776918740 Chr.5: 160427800-160427800 on Build GRCh38
rs1802499 Chr.5: 160427794-160427794 on Build GRCh38
rs1018150562 Chr.5: 160427821-160427821 on Build GRCh38
rs775556311 Chr.5: 160427783-160427783 on Build GRCh38
rs1001035771 Chr.5: 160427858-160427858 on Build GRCh38
rs762272902 Chr.5: 160427719-160427719 on Build GRCh38
rs765779103 Chr.5: 160427722-160427722 on Build GRCh38

View Full Product Details