Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 154091268-154091529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OPRM1
Entrez Gene ID
4988
Gene Name
opioid receptor mu 1
Gene Aliases
LMOR, M-OR-1, MOP, MOR, MOR1, OPRM
UniGene
Hs.2353
Gene Chromosome Location
Chr.6: 154010496-154246867 on Build GRCh38
Transcripts
NM_000914.4 NM_001008503.2 NM_001008504.3 NM_001008505.2 NM_001145279.3 NM_001145280.3 NM_001145281.2 NM_001145282.2 NM_001145283.2 NM_001145284.3 NM_001145285.2 NM_001145286.2 NM_001145287.2 NM_001285522.1 NM_001285523.1 NM_001285524.1 NM_001285526.1 NM_001285527.1 NM_001285528.1 NR_104348.1 NR_104349.1 NR_104350.1 NR_104351.1 XM_011535851.2 XM_011535853.2 XM_011535856.2 XM_011535862.2 XM_017010903.1 XM_017010904.1 XM_017010905.1 XM_017010906.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1286777 c.1006G>T p.V336F Chr.6: 154091314-154091314 on Build GRCh38
6105821 c.1022T>A p.L341Q Chr.6: 154091330-154091330 on Build GRCh38
137474 c.1319G>A p.R440Q Chr.6: 154091348-154091348 on Build GRCh38
5895861 c.1108C>T p.Q370* Chr.6: 154091416-154091416 on Build GRCh38
136576 c.1105C>T p.R369C Chr.6: 154091413-154091413 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762824513 Chr.6: 154091480-154091480 on Build GRCh38
rs200638619 Chr.6: 154091388-154091388 on Build GRCh38
rs201410932 Chr.6: 154091473-154091473 on Build GRCh38
rs77013544 Chr.6: 154091348-154091348 on Build GRCh38
rs200695585 Chr.6: 154091397-154091397 on Build GRCh38
rs761403456 Chr.6: 154091356-154091356 on Build GRCh38
rs771106503 Chr.6: 154091344-154091344 on Build GRCh38
rs747061106 Chr.6: 154091337-154091337 on Build GRCh38
rs202194691 Chr.6: 154091442-154091442 on Build GRCh38
rs199649616 Chr.6: 154091333-154091333 on Build GRCh38
rs201007162 Chr.6: 154091477-154091477 on Build GRCh38
rs1052445344 Chr.6: 154091469-154091469 on Build GRCh38
rs753872489 Chr.6: 154091308-154091308 on Build GRCh38
rs201496848 Chr.6: 154091448-154091448 on Build GRCh38
rs752930536 Chr.6: 154091319-154091319 on Build GRCh38
rs542134149 Chr.6: 154091334-154091334 on Build GRCh38
rs17181227 Chr.6: 154091484-154091484 on Build GRCh38
rs765079742 Chr.6: 154091395-154091395 on Build GRCh38
rs376756256 Chr.6: 154091447-154091447 on Build GRCh38
rs746718852 Chr.6: 154091455-154091455 on Build GRCh38
rs1021061437 Chr.6: 154091431-154091431 on Build GRCh38
rs199668737 Chr.6: 154091350-154091350 on Build GRCh38
rs772985784 Chr.6: 154091374-154091374 on Build GRCh38
rs751700008 Chr.6: 154091487-154091487 on Build GRCh38
rs757710134 Chr.6: 154091321-154091321 on Build GRCh38
rs751785041 Chr.6: 154091351-154091351 on Build GRCh38
rs200260230 Chr.6: 154091453-154091453 on Build GRCh38
rs778895453 Chr.6: 154091316-154091316 on Build GRCh38
rs201516315 Chr.6: 154091408-154091408 on Build GRCh38
rs184553578 Chr.6: 154091441-154091441 on Build GRCh38
rs893899910 Chr.6: 154091295-154091295 on Build GRCh38
rs954272197 Chr.6: 154091456-154091456 on Build GRCh38
rs765963522 Chr.6: 154091382-154091382 on Build GRCh38
rs772717842 Chr.6: 154091454-154091454 on Build GRCh38
rs770310912 Chr.6: 154091459-154091459 on Build GRCh38
rs776230437 Chr.6: 154091466-154091466 on Build GRCh38
rs778134755 Chr.6: 154091424-154091424 on Build GRCh38
rs976919553 Chr.6: 154091438-154091438 on Build GRCh38
rs199570196 Chr.6: 154091488-154091488 on Build GRCh38
rs755338736 Chr.6: 154091358-154091358 on Build GRCh38
rs201429844 Chr.6: 154091321-154091321 on Build GRCh38
rs200206841 Chr.6: 154091434-154091434 on Build GRCh38
rs373795454 Chr.6: 154091298-154091298 on Build GRCh38
rs1025223733 Chr.6: 154091376-154091376 on Build GRCh38
rs200735555 Chr.6: 154091423-154091423 on Build GRCh38
rs759581216 Chr.6: 154091392-154091392 on Build GRCh38
rs867865977 Chr.6: 154091407-154091407 on Build GRCh38
rs754712053 Chr.6: 154091310-154091310 on Build GRCh38
rs529479862 Chr.6: 154091493-154091493 on Build GRCh38
rs202118803 Chr.6: 154091426-154091426 on Build GRCh38
rs79216711 Chr.6: 154091381-154091381 on Build GRCh38
rs199984546 Chr.6: 154091414-154091414 on Build GRCh38
rs200512398 Chr.6: 154091347-154091347 on Build GRCh38
rs200226734 Chr.6: 154091444-154091444 on Build GRCh38
rs200468658 Chr.6: 154091301-154091301 on Build GRCh38
rs1010052797 Chr.6: 154091413-154091413 on Build GRCh38
rs961097298 Chr.6: 154091486-154091486 on Build GRCh38
rs949954608 Chr.6: 154091309-154091309 on Build GRCh38
rs79668187 Chr.6: 154091476-154091476 on Build GRCh38
rs73014297 Chr.6: 154091387-154091387 on Build GRCh38
rs202115383 Chr.6: 154091450-154091450 on Build GRCh38
rs201538512 Chr.6: 154091419-154091419 on Build GRCh38
rs200663220 Chr.6: 154091371-154091371 on Build GRCh38

View Full Product Details