Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 106095394-106095663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRDM1
Entrez Gene ID
639
Gene Name
PR domain 1
Gene Aliases
BLIMP1, PRDI-BF1
UniGene
Hs.436023
Gene Chromosome Location
Chr.6: 106046729-106109939 on Build GRCh38
Transcripts
NM_001198.3 XM_006715550.3 XM_011536062.2 XM_011536063.2 XM_017011187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6950668 c.232G>A p.G78R Chr.6: 106095663-106095663 on Build GRCh38
5344987 c.335G>A p.R112H Chr.6: 106095658-106095658 on Build GRCh38
1595713 c.313G>T p.E105* Chr.6: 106095636-106095636 on Build GRCh38
1439493 c.227G>A p.R76H Chr.6: 106095658-106095658 on Build GRCh38
7240857 c.329G>T p.G110V Chr.6: 106095652-106095652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1037373113 Chr.6: 106095462-106095462 on Build GRCh38
rs755652034 Chr.6: 106095600-106095603 on Build GRCh38
rs970480598 Chr.6: 106095534-106095534 on Build GRCh38
rs569250456 Chr.6: 106095446-106095446 on Build GRCh38
rs1018141207 Chr.6: 106095643-106095643 on Build GRCh38
rs531707133 Chr.6: 106095517-106095517 on Build GRCh38
rs886702927 Chr.6: 106095450-106095450 on Build GRCh38
rs372322351 Chr.6: 106095589-106095589 on Build GRCh38
rs534036483 Chr.6: 106095629-106095629 on Build GRCh38
rs568523900 Chr.6: 106095597-106095597 on Build GRCh38
rs957953172 Chr.6: 106095605-106095605 on Build GRCh38
rs894851261 Chr.6: 106095479-106095479 on Build GRCh38
rs763908201 Chr.6: 106095638-106095638 on Build GRCh38
rs1006442388 Chr.6: 106095458-106095458 on Build GRCh38
rs947179931 Chr.6: 106095566-106095566 on Build GRCh38
rs922926380 Chr.6: 106095449-106095449 on Build GRCh38
rs989397336 Chr.6: 106095612-106095612 on Build GRCh38
rs771366017 Chr.6: 106095575-106095575 on Build GRCh38
rs976175431 Chr.6: 106095432-106095432 on Build GRCh38
rs934406880 Chr.6: 106095475-106095475 on Build GRCh38
rs943619698 Chr.6: 106095424-106095424 on Build GRCh38
rs1052817528 Chr.6: 106095498-106095498 on Build GRCh38
rs745623405 Chr.6: 106095454-106095455 on Build GRCh38
rs775342433 Chr.6: 106095623-106095623 on Build GRCh38
rs1025173636 Chr.6: 106095514-106095517 on Build GRCh38
rs192908756 Chr.6: 106095552-106095552 on Build GRCh38
rs773856990 Chr.6: 106095587-106095587 on Build GRCh38
rs776311086 Chr.6: 106095640-106095640 on Build GRCh38

View Full Product Details